Overview of energy-dispersive X-ray spectroscopy technology
Elemental analysis is the fundamental application of energy-dispersive X-ray spectroscopy (EDS, EDX, or XEDS). EDS adds vital compositional information to electron microscopy images, enabling a comprehensive morphological and chemical overview of your sample.
Modern Scanning Transmission Electron Microscopy (S/TEM) X-ray analysis employs energy-dispersive X-ray spectroscopy (EDX) technology. In this process, the electron beam ejects atomic core-shell electrons in the sample. Recombination of a higher shell electron into the now empty core state creates X-rays imprinted with this atom’s characteristic energy. These X-rays are absorbed by a nearby semiconductor detector, generating electron-hole pairs that are separated and collected as charge pulses.
Fast electronics reliably detect and quantify these pulses, allowing for precise energy spectrum creation. Pulse-processing electronics measure the area under each pulse and then each pulse is “binned” into an energy spectrum. These spectra of characteristic peaks are automatically labeled by the computer.
While EDX is user-friendly and dependable, its limitations include a small analysis volume and poor total X-ray collection efficiency (~1%). To bring about a transformative advancement, a significant increase in the net X-ray count rate, far beyond incremental improvements, was identified as necessary. Addressing this challenge involved exploring all potential avenues for enhancement.
How does EDS work?
When a sample is excited by a source like an electron microscope's electron beam, it dissipates some of the absorbed energy by ejecting a core-shell electron. The ejected electron's place is filled by a higher energy outer-shell electron, and the difference in energy is released as an X-ray. These X-rays carry a unique spectrum corresponding to their atom of origin. EDS spectra are linked to specific sample positions and corresponding elements.
The peak positions in the spectrum indicate the element, while signal intensity reflects its concentration. Integrating an EDS detector with an electron microscope enables atomic-level compositional analysis.
Result quality depends on signal strength and spectrum cleanliness. Signal strength, especially for trace element detection and dose minimization, is contingent on a robust signal-to-noise ratio, enabling faster recording and artifact-free outcomes. Spectrum cleanliness is influenced by the composition of the electron column and affects the presence of unwanted peaks. 
Benefits of EDS analysis for materials science
  • Sensitive to low concentrations—minimum detection limits below 0.1% in the best cases
  • Affords a high degree of relative precision—typically 2–4%
  • Non-destructive in most situations
  • Usually requires minimal sample preparation effort and time
  • Delivers complete analyses of complex samples quickly, often in under a minute
Electroplated bimetallic metal foams imaged with ChemiSEM Technology. Sample courtesy of Prof. Dr. Peter Broekmann, Department of Chemistry and Biochemistry, University of Bern.
EDS analysis instruments
Axia ChemiSEM
An SEM EDS instrument that is flexible, easy to use, and offers instantaneous, quantitative elemental analysis
The Thermo Scientific Axia ChemiSEM distinguishes itself from traditional SEMs by continuously collecting EDS data in the background. It employs special algorithms to concurrently process SEM and EDS signals, enabling real-time presentation of sample morphology and quantitative elemental composition. It continuously processes EDS data, offering live updates on acquired elemental information. Users can toggle elements on and off to focus on specific areas of interest.
Quattro ESEM
An environmental scanning electron microscope (ESEM) for the study of materials in their natural state
The Thermo Scientific Quattro ESEM combines versatile imaging and analytics performance with a specialized environmental mode (ESEM) for studying samples in their natural state. It suits academic, industrial, and government labs with varied user backgrounds and research disciplines while still supporting the ability to run unique in situ experiments.
The Quattro ESEM is equipped with a field emission gun (FEG) for exceptional resolution and features three vacuum modes (high vacuum, low vacuum, and ESEM) to accommodate a wide range of samples, including those prone to outgassing or incompatible with vacuum conditions.
Apreo ChemiSEM
High-performance imaging with integrated chemical analysis and structural characterization
The new Thermo Scientific Apreo ChemiSEM System supports your materials science research by simplifying the imaging process, making it accessible to both expert users and newcomers alike. With innovative features like Smart Frame Integration (SFI) and newly developed autofocus and autostigmation functions, it can help you easily obtain clear, high-resolution images as well as results for elemental analysis (EDS) and diffraction (EBSD).
Prisma E SEM
Scanning electron microscope for industrial research and development with environmental scanning electron microscope capability
The Thermo Scientific Prisma E Scanning Electron Microscope (SEM) integrates various imaging and analytical features with advanced automation, offering a comprehensive solution. Ideal for industrial research, quality control, and failure analysis, it offers high resolution, sample versatility, and a user-friendly interface. The Prisma E SEM is the successor to the highly acclaimed Thermo Scientific Quanta SEM.  
Phenom Desktop SEM
Desktop SEM instruments boast enhanced ease of use, democratizing SEM technology. Additionally, they have the added benefit of a reduced frame size. While traditional “floor model” SEM instrumentation can require a specialized room or facility, desktop instrumentation is far more robust. Some have even taken their desktop SEMs on the road, providing on-site electron microscopy analysis via mobile laboratories.
Spectra Ultra
Scanning transmission electron microscope for imaging and spectroscopy of beam sensitive materials
To truly optimize TEM and STEM imaging, EDX and EELS may require acquisition of different signals at different accelerating voltages. The rules may vary from sample to sample but, it is generally accepted that: 1) the best imaging is done at the highest possible accelerating voltage above which visible damage will occur, 2) EDX, especially when mapping, benefits from lower voltages with increased ionization cross-sections, thus yielding better signal-to-noise ratio maps for a given total dose, and 3) EELS works best at high voltages to avoid multiple scattering, which degrades the EELS signal with increasing sample thickness.
Spectra 200
High throughput TEM and STEM microscope for all materials science applications
The Thermo Scientific Spectra 200 (S)TEM, is a high-throughput, aberration-corrected, (scanning) transmission electron microscope for all materials science applications designed to offer an exceptional level of mechanical stability and high imaging quality though passive and (optional) active vibration isolation.
The system is housed in a fully re-designed enclosure with a built-in on-screen display for convenient specimen loading and removal. For the first time, full modularity and upgradeability can be offered between uncorrected and single-corrected configurations with variable heights, allowing maximum flexibility for different room configurations.
Spectra 300
High resolution TEM and STEM microscope for all materials science and semiconductor applications
The Thermo Scientific Spectra 300 (S)TEM is a high resolution, aberration corrected, scanning transmission electron microscope for all materials science applications designed to offer an exceptional level of mechanical stability and high imaging quality though passive and (optional) active vibration isolation. The system is housed in a fully redesigned enclosure with a built-in on-screen display for convenient specimen loading and removal. For the first time, full modularity and upgradeability can be offered between uncorrected and single-corrected configurations with variable heights, allowing maximum flexibility for different room configurations.  
Talos F200X TEM
TEM microscope for high resolution TEM and STEM with accurate chemical quantification
The Thermo Scientific Talos F200X STEM is a scanning transmission electron microscope that combines outstanding high-resolution STEM and TEM imaging with exceptional energy dispersive X-ray spectroscopy (EDS) signal detection. 2D/3D chemical characterization with compositional mapping is performed by 4 in-column SDD Super-X detectors with remarkable cleanliness. The Talos F200X scanning transmission electron microscope allows for fast, precise EDS analysis in all dimensions, along with high-resolution TEM and STEM (HRTEM and HRSTEM) imaging with fast navigation for dynamic microscopy.
Talos F200S TEM
TEM microscope for high productivity, high resolution TEM and STEM characterization with chemical quantification
The Thermo Scientific Talos F200S (S)TEM is a (scanning) transmission electron microscope that combines outstanding high-resolution STEM and TEM imaging with exceptional energy dispersive X-ray spectroscopy (EDS). The Talos F200S (S)TEM features great versatility and high through in STEM imaging and allows for precise EDS analysis and high-resolution TEM (HRTEM) for dynamic microscopy.
Talos F200i TEM
TEM and STEM analysis for high throughput, high resolution chemical characterization and dynamic observations
The Thermo Scientific Talos F200i (S)TEM is a 20-200 kV field emission (scanning) transmission electron microscope designed for performance and productivity across a wide range of materials science samples and applications. Its standard X-Twin pole piece gap—giving high flexibility in applications—combined with a reproducibly performing electron column opens opportunities for high-resolution 2D and 3D characterization, in situ dynamic observations, and diffraction applications.
Talos F200C TEM
TEM and STEM analysis for high contrast imaging of beam sensitive materials
The Thermo Scientific Talos F200C TEM is a 20-200 kV thermionic (scanning) transmission electron microscope designed for performance and productivity across a wide range of samples and applications, such as 2D and 3D imaging of cells, organelles, asbestos, polymers, and soft materials, both at ambient and cryogenic temperatures.
Talos 12 TEM
Versatile TEM and STEM microscope for 2D and 3D visualization of beam sensitive samples and materials using high contrast
The Thermo Scientific Talos 12 TEM is a multidisciplinary 120 kV thermionic (scanning) transmission electron microscope designed to fit your needs. The user-friendly Talos 12 TEM offers exceptional performance for a diverse range of scientific imaging and experimentation needs.  It can be used for 2D and 3D imaging of cells, cell organelles, asbestos, polymers, and soft materials, both at ambient and cryogenic temperatures. 
For Research Use Only. Not for use in diagnostic procedures.