CD133 Antibody (63-1338-42) in Flow
Staining of normal human peripheral blood cells with Anti-Human CD34 APC (cat. 17-0349) and Mouse IgG1 K Isotype Control Super Bright 600 (cat. 63-4714) (left) or Anti-Human CD133 Super Bright 600 (right). Cells expressing intermediate levels of CD45 within the lymphocyte gate were used for analysis.
Staining of normal human peripheral blood cells with Anti-Human CD34 APC (cat. 17-0349) and Mouse IgG1 K Isotype Control Super Bright 600 (cat. 63-4714) (left) or Anti-Human CD133 Super Bright 600 (right).  Cells expressing intermediate levels of CD45 within the lymphocyte gate were used for analysis.
Related Products
Related Images