Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Columns and Consumables / 5 Key Takeaways from LC Columns Tips and Tricks Webinar
5 Key Takeaways from LC Columns Tips and Tricks Webinar
By Hai Ning Pee, APJ Regional Marketing Manager 01.16.2025
5 Key Takeaways from LC Columns Tips and Tricks Webinar
The webinar, hosted by Hai Ning, Regional Marketing Manager for APJ, featured Adham Ahmed, a global product manager with over 15 years of experience in chromatographic applications. Adham provided insights into LC column attributes for small molecules, setting up new columns, troubleshooting techniques and column care tips to prolong column life. Below are some key takeaways:
  1. LC Column Attributes and Setup: Understanding LC column attributes (particle size, surface area, carbon load) and proper setup (checking COA, column care guide, solvent compatibility, and conditioning) are crucial for optimal performance and interaction with analytes.
  2. Root Cause Identification and Troubleshooting: Effective troubleshooting requires identifying whether issues originate from the column or the system and addressing common problems like insufficient conditioning, system leaks, or system blockages.
  3. Retention Time Consistency: Consistent retention times depend on proper column conditioning, system maintenance, and addressing factors such as possible leakage and system errors.
  4. Impact of Mobile Phase: The age and pH of the mobile phase can significantly affect column performance and retention time. Using a pH outside the recommended range can cause irreversible damage to the stationary phase.
  5. Column Maintenance: Regular column washing, effective sample preparation, and the use of fresh mobile phase solvents and additives are essential for maintaining column performance. Consult the column care guide or technical support team for further assistance.
Register for our On-Demand Webinar: LC Columns Tips and Tricks to gain valuable insights to optimize your separations, extend column lifetime and effective troubleshooting strategies.
Automated production line filling and transporting medical injection pens in a factory.
Stability-Indicating HPLC-UV Method for Semaglutide Impurity Analysis
As demand for GLP-1 receptor agonists like semaglutide conti...
Read More
Hand placing a missing piece into a white jigsaw puzzle.
Blueprint for QC Ready mRNA Mapping
mRNA therapeutics demand analytical methods that are fast, r...
Read More
Brewer and scientist examining hops and beer with chromatography graph overlay.
How Bitter Acid Analysis Helps Brewers Better Control Beer Flavors
Tasty beer starts with knowing what’s in your hops. Explor...
Read More
AAV-SEC Analysis: Top 3 Takeaways From the Latest Article
AAV-SEC Analysis: Top 3 Takeaways From the Latest Article
What is the best column for separating AAV size variants? Th...
Read More
Hai Ning Pee
Advancing PFAS Detection in Pharmaceutical Manufacturing with LC-HRAM-MS
Enhanced Security Through Centralized Data Management: A guide for Chromatography and Mass Spectrometry Users