Did you know that there are significant differences in hydrolytic glass?
When we talk about glass types 70, 51, or 33, the number indicates the amount of free silanol groups on the surface of your glass. Free silanol groups tend to interact with polar substances such as NH, NH2 or OH groups (drugs of abuse: non-derivetized), and with halogenated species (PCBs, Fuarans and Dioxins). They also interact with pesticides containing sulfur or phosphor in the molecular structure – to name merely a few examples. Derivatization or salinized glass vials — which can show weird effects in LCMS — are not always an option. This might not matter if you are looking into the percentage of analytes. However, when talking about femtogram, this becomes a different story. If you are missing the last points in your lower end of the calibration curve, then you have your answer.
Considering a lack of awareness on this topic, the sensitivity of the instrument is of paramount importance, considering something as trivial as the vials could make or break your detection. If you see issues with your detection limits, consider the vials you use. If the analyte doesn’t reach your detector, it can’t be detected. It as simple as that.
On the topic of glass quality: Have you ever heard about Chromcol GOLD vials? These ultra-inert vials with the lowest free silanol groups on the surface are an excellent solution to your analytical challenges.
Improve your detection limits down to the last femtograms: www.thermofisher.com/inertvials
When “Fairy Lights” Turn Toxic: The Hidden Hazard of Chlorinated Paraffins
Are you looking forward to the holidays and a lot of cozy ev...
Read More
MOSH/MOAH Analysis: Technology, Regulation, and the Road Ahead
Mineral oil hydrocarbons (MOSH) and mineral oil aromatic hyd...
Read More
New European Wine Scandal on PFAS in Wine
Introduction PFAS, also known as “eternity chemicals...
Read More
Unveiling the Advanced Workflow for PAH Analysis in Smoke Flavored Foods
Introduction Polycyclic aromatic hydrocarbons (PAHs) are a p...
Read More