Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Pesticide Analysis / A Commitment Realized: New Optimized and Standardized Solution for Polar Pesticides Analysis Launches
A Commitment Realized: New Optimized and Standardized Solution for Polar Pesticides Analysis Launches
By AnalyteGuru Staff, 11.20.2019
anionic-pesticide-explorer-prod-cat-1000x1000
These were not empty words. At the 9th International Symposium on Recent Advances in Food Analysis (RAFA) which was held in early November, we launched the Thermo Scientific™ Anionic Pesticides Explorer, an integrated sample-to-results analytical workflow. This workflow has been extensively optimized and validated with a range of representative sample types — including wheat flour, grape, leek and baby food — to offer high-throughput detection, identification and quantification of anionic polar pesticides compliant with EU SANTE/11813/2017 method validation and ongoing quality control guideline criteria in a complete off-the-shelf package.
The instrumentation comprises the Thermo Scientific™ Dionex Integrion High-Performance Ion Chromatography system coupled with the Thermo Scientific™ TSQ Altis Triple Quadrupole MS. It comes complete with optimized sample preparation, software, necessary consumables, suitability check standard solution and a detailed deployment guide to enable food testing laboratories to overcome the challenge of reliable quantitation and identification of multiple polar anionic pesticides.
Richard mentioned many benefits of using IC/MS for this analysis, and the key takeaways that still apply to this workflow, are the robustness gained by using high capacity and high matrix tolerance anion exchange columns and productivity gains achieved as there is no need to add a derivatization step.
If this solution has caught your attention and you wish to learn more, there is a very detailed and well-written application note titled, “Multi-residue analysis of polar anionic pesticides in food samples using a compact ion chromatography system coupled with tandem mass spectrometry (IC-MS/MS).” There is also a really engaging on-demand webinar, “A New Integrated, Sample-to-Result Analytical Workflow for the Sensitive and Reliable Analysis of Polar Anionic Pesticides and Metabolites” that I recommend that you watch.
upcoming events for clinical research and toxicology
Upcoming LC-MS Events to Watch this Fall
Mass spectrometry professionals in clinical research, forens...
Read More
Thermo Fisher Scientific's Edward Goucher and Alexandra Graham pictured beside KCL's Stefania Boccuzzi and Vincenzo Abbate
Advancing Drug Analysis with VeriSpray PaperSpray Mass Spectrometry
At the January 2025 CFSRE Seized Drugs Symposium, a Thermo F...
Read More
MOSH MOAH image
MOSH/MOAH Analysis: Technology, Regulation, and the Road Ahead
Mineral oil hydrocarbons (MOSH) and mineral oil aromatic hyd...
Read More
Pesticides
Pesticide Residue Analysis in the 21st Century: Innovations in Sample Preparation and Detection Using the Thermo Scientific TSQ 9610 GC-MS/MS
Introduction The accurate detection of pesticide residues in...
Read More
AnalyteGuru Staff
Predicting the Future for Pharmaceutical Impurities Roundtable
Automate Sample Preparation for PFAS Analysis in Drinking Water