Anionic Pesticides Explorer
Anionic Pesticides Explorer
Thermo Scientific™
Anionic Pesticides Explorer
Discover the productivity and robustness of the Anionic Pesticides Explorer workflow for the multi-residue determination of anionic pesticides in complex food matrices.
Have Questions?
Catalog NumberColumn TypeFor Use With (Equipment)Includes
AP-IALT-01Thermo Scientific™ Dionex™ IonPac™ AG19-4μm anion guard column, 2 x 50 mm and Thermo Scientific™ Dionex™ IonPac™ AS19-4μm separation column, 2 x 250 mmIC-MS SystemsUser Guide, Training and Testing Performance with QC test mix
Catalog number AP-IALT-01
Price (USD)
-
Request A Quote
Column Type:
Thermo Scientific™ Dionex™ IonPac™ AG19-4μm anion guard column, 2 x 50 mm and Thermo Scientific™ Dionex™ IonPac™ AS19-4μm separation column, 2 x 250 mm
For Use With (Equipment):
IC-MS Systems
Includes:
User Guide, Training and Testing Performance with QC test mix
Expand the scope of your routine pesticide analysis in food using the Thermo Scientific™ Anionic Pesticides Explorer, an integrated sample-to-results analytical workflow. It enables laboratories to overcome the challenge of reliable quantitation and identification of multiple polar anionic pesticides including glyphosate, glufosinate, fosetyl, chlorate, and others; all in a single analysis and without the need for derivatization. This analytical workflow is delivered as a robust, validated off-the-shelf solution offering fast implementation and optimum performance for routine testing labs.
The Anionic Pesticides Explorer provides a fully integrated analytical workflow for the complete analysis of 14 anionic pesticides and their metabolites, increasing productivity and reducing costs. Extraction is performed using a modified version of the Quick Polar Pesticides Method (QuPPe) with a combination of freezing, dilution and solid phase extraction clean-up to reduce matrix co-extractives before IC-MS/analysis. High performance ion chromatography with post-column eluent suppression, coupled to a high-sensitivity triple quadrupole mass spectrometer (IC-MS/MS), all controlled by the Thermo Scientific™ Chromeleon™ Chromatography Data System, providing robust and sensitive quantitation and identification of all targeted anionic polar pesticides.
The Anionic Pesticides Explorer has been extensively optimized and validated with a range of representative sample types including wheat flour, grape, leek and baby food. It includes all instruments, software, necessary consumables, suitability check standard solution, and a detailed deployment guide for reaching higher productivity and maintaining the workflow performance during routine analysis.
For routine laboratories, the Anionic Pesticide Explorer offers:
  • Compliance with regulations and method performance guidelines.
  • Increased productivity, ensured by consolidation of multiple methods into one analysis.
  • Robustness, underpinned by the high capacity and high matrix tolerance of anion exchange columns enabling the routine analysis of analytes at low concentration in complex samples.
Specifications
Column TypeThermo Scientific™ Dionex™ IonPac™ AG19-4μm anion guard column, 2 x 50 mm and Thermo Scientific™ Dionex™ IonPac™ AS19-4μm separation column, 2 x 250 mm
For Use With (Equipment)IC-MS Systems
IncludesUser Guide, Training and Testing Performance with QC test mix
Liquid Chromatography SystemThermo Scientific™ Dionex™ Integrion™ HPIC™ system, RFIC model including electrolytic eluent generation capabilities, column oven and detector temperature control, RFID consumables tracking plus an additional six-port valve, eluent generation consumables, an electrolytic suppressor, CD Conductivity Detector, and IC-MS interface kit with two auxiliary pumps (Thermo Scientific™ AXP, and AXP-MS pumps). Thermo Scientific™ Dionex™ AS-AP Autosampler with 1.5mL vial trays, and 1.5mL polypropylene vials with pre-cut septa.
Mass Spectrometer InterfaceThermo Scientific™ TSQ Altis™ Triple Quadrupole Mass Spectrometer, including Thermo Scientific™ EASY-Max NG™ Source, syringes, 6 port valve
MethodsThermo Scientific™ Chromeleon™ pre-optimized eWorkflow for acquisition and processing method
Sample PreparationModified Quick Polar Pesticides Method (QuPPe) with SPE clean-up option to reduce matrix effects
SoftwareThermo Scientific Chromeleon Chromatography Data System
TypeSoftware
Unit SizeEach