The American Society for Testing and Materials (ASTM) and the European Standards (EN) have published analytical methods that are widely adopted to characterize impurities in pure biodiesel (B100) by using gas chromatography (GC) coupled to flame ionization detection (FID):
| Standard methods | Relevance |
| Determination of residual free and total glycerol and mono-, di-, triglyceride content | Unreacted glycerides and free glycerol are critical for engine injector clogging and fouling compounds formation |
| Standard methods | Relevance |
| Determination of total FAMEs (Fatty Acid Methyl Esters) and linolenic acid methyl ester (C18:3) content | FAME content is a measure the reaction yield and allows accurate calculation of the cetane index of biodiesel |
| Standard methods | Relevance |
| Determination of residual methanol | Small amount of residual methanol can reduce the flash point of biodiesel, potentially affecting the safe operation of fuel pumps, lifetime of seals and elastomers, and resulting in poor combustion |
Biodiesel analyzer: comprehensive quality check with one automated system
Each of the standard methods requires specific GC conditions, calibration set up and sample preparation procedures, typically approached with different GC systems and manual sample handling. Especially the analysis of glycerol and unreacted mono-, di-, triglycerides requires complex and time consuming sample derivatization and complex calibration set up.
To face the high throughput demand of analytical laboratories, a multimethod approach for the analysis of biodiesel samples can be achieved by operating a Thermo Scientific™ TriPlus™ RSH SMART robotic autosampler in Clone Mode configuration, allowing it to serve two Thermo Scientific TRACE™ 1610 GC systems. The TriPlus actss as two independent autosamplers, so that different samples can be automatically injected into two GCs — one dedicated to glycerol analysis due to the high oven temperature that has to be reached — and one dedicated to methanol and FAME determination.
The automation capability and versatility of the TriPlus RSH SMART autosampler is used to execute derivatization protocol as well as calibration setup with internal standard addition, for all the standard methods, followed by injection into the GC systems.
This comprehensive solution for biodiesel analysis is ideal for laboratories performing quality control and production process monitoring, and looking to improve productivity with confident results.
Automation for calibration standards, sample derivatization, and internal standard addition, reduces the risk of errors and cross-contaminations, delivering highly precise and reliable analytical results. Automation also allows for unattended operations, saving analysts valuable time as well as limiting analysts’ exposure to the toxic reagents typically used in biodiesel sample derivatization protocols.
Additional resources
- Thermo Scientific AN001897 – Biodiesel quality assessment: an automated approach for analysis of free and total glycerol content in biodiesel (B100), according to the EN 14105 and ASTM D6584 methods.
- Thermo Scientific AN001898 – Biodiesel quality assessment: characterization of residual methanol in finished biodiesel (B100) by headspace sampling according to EN 14110.
- Thermo Scientific AN001899 – Biodiesel quality assessment: determination of esters and linolenic acid methyl ester content in biodiesel (B100) by GC-FID, according to EN 14103
- Thermo Scientific EB000396 – Guide to automated sample preparation workflows for GC and GC-MS