Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Pharmaceutical / Decoding Amino Acids Using Ion Chromatography
Decoding Amino Acids Using Ion Chromatography
By Wai-Chi Man, Product Marketing Manager, IC/SP, Thermo Fisher Scientific 10.30.2023
Sometimes, you need the little guys to see the bigger picture. Amino acids are the little guys, the peptides are the bigger ones, and then the mega protein structural picture. To understand their structures, we must identify their sub-components. Like in troubleshooting, the problem arises, and then we put all the pieces together to solve it (or to see the story)!
There are many ways to analyze amino acids. The most recommended way is using post-column derivatization with the Pickering Pinnacle PCX amino acid analyzer solution, suggested by the European Pharmacopeia (EP) Monograph. It uses post-column ninhydrin derivatization. It requires some colorful staining and corrosive chemicals and can stain and corrode the instrument if it has a metal flow path. Some laboratories use inert flow path ion chromatography (IC) systems to avoid any corrosion. See the case study from Reading Scientific Services Limited (RSSL) about their amino acids application and setup. We know precolumn derivatization is highly susceptible to interference from the sample matrix, and complex sample matrices can reduce derivatization efficiency, causing high variability in amino acid recovery.
Although there are other amino acid methods, we often go with what is familiar to us and what we have around the laboratory. Sometimes it works, and sometimes not. It can divert and create other problems and challenges. I am a big fan of the ‘KISS’ method (Keep it stupidly simple). For many years we have demonstrated high-performance anion exchange (HPAE) chromatography for carbohydrate analysis with no derivatization as a simple and easy-to-use technique. Like carbohydrates, amino acids can easily be reduced and oxidized (Redox reaction). Hence, they can be analyzed by HPAE chromatography, just like carbohydrates, as demonstrated in our manual for Amino Acid Analysis (AAA) – Direct, MAN031481). With HPAE, with integrated pulsed amperometry detection (IPAD), the system can separate the amino acids and carbohydrates in complex matrices even in fermentation broth (See application update 152 and application note 150). The capability of AAA-Direct to analyze amino acids can be demonstrated in peptide samples, as types of hydrolysates, in Technical Note 50.
The use of AAA for total protein determinations provides advantages over colorimetric methods. Complete degradation of proteins to amino acids enables for the accounting of the composition completely, without bias from the specificity of dye-binding typical for colorimetric assays (BCA, Bradford, Lowry, etc.). The compositional analysis also provides information about the identity and purity of the protein not achieved using dye-binding assays. Direct detection of amino acids using AAA-Direct results in a lower cost in materials and labor. It also eliminates the risk of exposure to the hazardous chemicals used for pre- or post-column derivatization and helps to eliminate hazardous waste stream created by pre- or post-column derivatization techniques. Accurate total protein measurements can be achieved using AAA-Direct, with results equivalent to other AAA techniques (Application Note 163).
If you are studying proteins and peptides, you may want to know how to decode the amino acids. Why not KISS your analysis with AAA-Direct using HPAE-PAD? There is no messy derivatization, and it is much safer for you, your instrument and your laboratory.
Learn more
Webpage: Ion Chromatography Columns Selection
RSSL Case study: RSSL expands GMP services for the pharmaceutical industry
AAA – Direct manual
Application update 152: An Improved Gradient Method for the AAA-Direct™ Separation of Amino Acids
Application note 150: Determination of Amino Acids in Cell Cultures and Fermentation Broths
Technical note 50: Determination of the Amino Acid Content of Peptides by AAA-Direct
Application note 163: Determination of Protein Concentrations Using AAA-Direct™
On LinkedIn? Visit our LinkedIn page #IonChromatography
Brewer and scientist examining hops and beer with chromatography graph overlay.
How Bitter Acid Analysis Helps Brewers Better Control Beer Flavors
Tasty beer starts with knowing what’s in your hops. Explor...
Read More
Amino acid chromatogram showing labeled peaks over a 7-minute separation time.
How to Cut HPLC Sample Preparation Time for Derivatizing Amino Acids
Derivatizing amino acids doesn’t have to take hours to do....
Read More
freedom to fragment
Freedom to Fragment: Introduction
The importance of fragmentation in mass spectrometry 1. What...
Read More
IC Golden Jubilee blog header image
What is the Ion Chromatography Golden Jubilee? Celebrating 50 Years of IC Innovation
The year 2025 marks the golden jubilee of ion chromatography...
Read More
Wai-Chi Man
Wai-Chi Man is Product Marketing Manager, IC/SP, for Thermo Fisher Scientific.
Is There a Better Way to Determine Anionic Impurities in Sulfuric Acid?
A Novel Automated Approach to Detecting Chloride in Drinking Water with Discrete Analysis