Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Combustion Ion Chromatography / Enhancing PFAS Analysis in Food Contact Materials Using Combustion Ion Chromatography
Enhancing PFAS Analysis in Food Contact Materials Using Combustion Ion Chromatography
By Jingli Hu, Application Staff Scientist, Thermo Fisher Scientific 07.25.2025
Per- and polyfluoroalkyl substances (PFAS) have been used in food contact materials (FCM) for decades to provide grease and water repellency. However, due to their persistence, bioaccumulative nature, and toxicity, PFAS pose significant concerns for human health and environmental pollution. As a result, several U.S. states have enacted legislation to limit or ban the use of PFAS in food packaging.
Food packaging
The Thermo Scientific application proof note: Screening of PFAS in food contact materials: Utilizing a new combustion-ion chromatography system for total organic fluorine (TOF) analysis, describes a method to measure TOF in food contact materials (FCM) using an innovative combustion-ion chromatography (C-IC) system. This system integrates an ion chromatography system with a combustion/adsorption module to offer enhanced combustion efficiency. The application proof note addresses topics that include:
  • State regulations: Twelve U.S. states have enacted or will enact legislation to limit PFAS in food packaging, with California banning all plant fiber-based food packaging containing PFAS above 100 ppm TOF.
  • A new, innovative C-IC system: The Thermo Scientific Cindion Combustion Ion Chromatography System improves combustion efficiency with a z-fold combustion tube, reducing combustion times and saving bench space.
  • Efficient analysis: The system measures TOF by subtracting total inorganic fluorine (TIF) from total fluorine (TF), with TF measured via combustion and TIF via direct injection into the IC.
  • Enhanced workflow: The C-IC system supports a versatile 2-in-1 configuration, enabling seamless switching between C-IC and standalone IC, improving efficiency and convenience.
The C-IC method provides a robust tool for manufacturers to ensure compliance with state regulations on PFAS in food contact materials. Additionally, the Cindion C-IC system is shown to meet the rigorous demands of PFAS analysis in FCM. With its advanced features and integrated software, it offers enhanced performance, efficiency, and a more streamlined workflow, making it an invaluable asset for analytical laboratories.
Visit the Cindion combustion ion chromatography system page to learn how the Cindion C-IC system can elevate your analytical capabilities and ensure regulatory compliance.
Visit us onĀ LinkedIn: #PFAS #FoodPackaging #CIC #EnvironmentalProtection
pfas image
Understanding PFAS in Pharmaceutical Manufacturing
Introduction to PFAS Per- and polyfluoroalkyl substances (PF...
Read More
wine bottle image
New European Wine Scandal on PFAS in Wine
Introduction PFAS, also known as “eternity chemicals&#...
Read More
US EPA Method 1621_1200x600
Understanding PFAS and Combustion-Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) comprise more tha...
Read More
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
In industrial processes, accurately determining halogens and...
Read More
Jingli Hu
Jingli Hu is a Senior Applications Chemist at Thermo Fisher Scientific in Sunnyvale, CA. She specializes in ion chromatography (IC) applications, focusing on food, environmental, and biopharma markets.
GC Orbitrap Guru Series #4: Scott Borden: Deep Breaths in Breathomics
Freedom to Fragment: Introduction