Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Metabolomics / Not Using IC-MS in Metabolomics to Fill the Gaps? See What You're Missing
Not Using IC-MS in Metabolomics to Fill the Gaps? See What You’re Missing
By Gemma Ellison, Thermo Fisher Scientific 03.23.2020
To state the obvious, researching metabolomics is complex! As a still relatively new branch of “omics,” researching and understanding the metabolism of living organisms in a wide range of conditions, including in health and disease, offers up complex matrices, large numbers of analytes to map and low levels of detection required; plus the sheer amount of data involved. There is no doubt advances in mass spectrometry, particularly high-resolution accurate mass instruments, have helped shape the study of metabolites allowing sensitivity in the femtomolar and attomolar levels.
Most research scientists will be familiar with liquid chromatography methods to separate out compounds of interest then hyphenating these techniques to a mass spectrometer to harness the additional resolving power and sensitivity required.
Gas chromatography is another tool in metabolomics profiling. Although not many metabolites are truly volatile, derivatization can be used to detect, for example, amino acids using GC-MS, however, derivatization comes with its own issues as detailed in my previous blog post. LC-MS would be the overarching technique with many citations in the literature. However, LC-MS does also have some limitations in its capabilities.
This diagram demonstrates the analyte areas generally covered using HILIC or reverse-phase liquid chromatography in terms of polarity and ionic properties.
Any analyte which is highly polar and highly ionic, such as sugar-phosphate isomers or those in the TCA (tricarboxylic acid) cycle, such as citrate, isocitrate, malate, and glutamate, are better suited to ion chromatography. Couple this to mass spectrometry and you have separation designed for these compounds with all the sensitivity and resolving power of a mass spectrometer.
Ion Chromatography coupled to mass spectrometry is a growing technology in many fields due to the fact it can fill in the gaps left by LC-MS analysis. IC-MS is used in targeted and untargeted metabolomic profiling. Analyzing small polar metabolites has been a key part of research into metabolic disturbances in oral cancers. Methods have been developed to analyze mono and diphosphate sugars in under nine minutes. With the advances in HRAM MS systems, you can calculate the empirical formula of the target compound from the molecular ion. If you have gaps in your metabolomics profiling, can IC-MS help you reach further?
You can contact us to speak with an expert on this subject. Become a member of our metabolomics community.
On LinkedIn? Visit our LinkedIn page #metabolomics #icms #lcms
Colorful 3D molecular structures and lab glassware in a vibrant scientific illustration.
One Injection, Two Answers: Introducing Squad Metabolomics on Orbitrap Mass Spectrometers
Metabolomics researchers are often forced to choose between ...
Read More
Excedion Pro Banner
Discover. Innovate. Exceed. — Introducing the Thermo Scientific Orbitrap Excedion Pro Mass Spectrometer
Mass spectrometry is entering a new era of precision, perfor...
Read More
Thermo Fisher Wins Lab Innovation Award for New Stellar Mass Spectrometer
Thermo Fisher Wins Award for Stellar Mass Spectrometer
Celebrating innovation in mass spectrometry We are delighted...
Read More
Unveiling the Power of the Stellar Mass Spectrometer in Metabolomics
Unveiling the Power of the Stellar Mass Spectrometer in Metabolomics
In the intricate world of metabolomics, understanding the co...
Read More
Gemma Ellison
An ICP MS Expedition through the Elements: Part 2
What Do You Know About High-Temperature Static Headspace?