Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Biopharmaceutical / Five Key Advantages of the Thermo Scientific LC-MSn Solution for Ultra-Sensitive GLP-1 Quantitation
Five Key Advantages of the Thermo Scientific LC-MSn Solution for Ultra-Sensitive GLP-1 Quantitation
By Scott M. Peterman, PhD, Sr. Product Marketing Manager, Life Science Mass Spectrometry, Thermo Fisher Scientific 09.25.2025
Bioanalytical analysis using LC-MSn requires precise, accurate, and robust methods to confidently evaluate pharmacokinetics and toxicokinetics. This is particularly challenging for peptide-based biologics like glucagon-like peptide-1 (GLP-1), which have low circulating levels and require high sensitivity in complex biological matrices. The Thermo Scientific LC-MSn solution, featuring the Stellar mass spectrometer acquiring parallel reaction monitoring (PRM) MS2 and MS3, offers a new paradigm for routine bioanalysis of GLP-1, improving quantitative performance almost 16-fold compared to triple quadrupole mass spectrometers (QQQ MS).
GLP-1 image
Here are five key advantages of the Thermo Scientific LC-MSn solution for GLP-1 quantitation:
     1. Enhanced quantitative sensitivity
The Stellar mass spectrometer enables researchers to achieve quantitative sensitivity down to 0.03 ng/mL using both simple and complex sample preparation methods. This is a significant improvement over more common SRM/MRM methods, which often struggle to reach such low levels due to the limitations of MS2 quantitation of one product ion.
     2.  Parallel reaction monitoring (PRM) acquisition improved specificity with MS2 acquisition
The Stellar MS boosts measured signal-to-noise (S/N) ratios 3X at 0.05 pg/mL GLP-1 spiking levels through co-adding multiple MS2 product ion signal without adding noise. This enhances data quality and quantitative performance, allowing for more accurate and reliable results at LOD and LOQ levels.
     3. Reach next-level performance with SPS MS3 acquisition
Moving from PRM-based MS2 to MS3 increases the specificity and selectivity necessary for highly confident quantitation in complex biological matrices. Extending the MS3 performance using synchronous precursor selection (SPS) boosts the sensitivity needed to reach the same LOQ using more cost-effective sample preparation steps as MS2 acquisition with SPE. By co-isolating the three MS2 product ions prior to dissociation, more of the original precursor signal is measured at the MS3 level. Acquiring SPS MS3 using PRM also enables post-acquisition summation of multiple product ions realizing the same benefits as that described when using PRM-MS2.
     4. Simplified data acquisition with dynamic AGC
Dynamic automatic gain control (AGC) manages ion accumulation times to maintain a stable charge density across a range of GLP-1 expression levels. This ensures high data quality by improving ion statistics and maintaining robust and reproducible PRM acquisition. The user defines the targeted charge density, LC peak width, and number of data points, while AGC automatically adjusts ion fill times to ensure optimal data acquisition.
     5. Comprehensive workflow management with Chromeleon 7.4 CDS software
Chromeleon 7.4 software provides a unique software ecosystem managing the entire LC-MSn workflow, from data acquisition to customizable reporting. It handles unregulated and regulated data acquisition, data processing, and system suitability testing, ensuring compliance with stringent regulatory requirements. The software also facilitates collaboration and cross-study management with full traceability and verification throughout the data lifecycle.
Conclusion
The Thermo Scientific LC-MSn solution, featuring Chromeleon CDS software and the Stellar mass spectrometer, offers unparalleled sensitivity, specificity, and quantitative performance for GLP-1 quantitation. By leveraging advanced techniques such as PRM acquisition, SPS MS3 acquisition, and dynamic AGC, researchers can achieve reliable and accurate results with simplified sample preparation methods. Chromeleon 7.4 software further enhances the workflow by providing comprehensive data management and compliance capabilities that transitions from method creation to analytical quantitation.
For more information visit Chromeleon Software for Targeted Quantitation
For more information visit Stellar mass spectrometer
Visit us on LinkedIn: #GLP-1, #biologics, #PRM, #bioanalytical
Upgrade Smarter: Trade In Your Current LC-MS System and Boost Lab Performance Without Breaking Budget
Is your current mass spectrometer limiting your lab’s capa...
Read More
Redefining sensitivity and selectivity in steroid quantification
Advancing Steroid Quantification: Unlocking Sensitivity and Selectivity with the Thermo Scientific Stellar Mass Spectrometer
Quantifying steroids in biological matrices has always prese...
Read More
Going beyond 4D to improve analytical performance for targeted quantitative analysis
Why You Need a Verification-Class Mass Spectrometer: Going Beyond 4d to Improve Analytical Performance for Targeted Quantitative Analysis
Part 4: Going beyond 4D to improve analytical performance fo...
Read More
Why You Need a Verification-Class Mass Spectrometer: Solving the Top 5 Pain Points of Translational Metabolomics Researchers
Why You Need a Verification-Class Mass Spectrometer: Solving the Top 5 Pain Points of Translational Metabolomics Researchers
Part 3: Solving the top 5 pain points of translational metab...
Read More
Scott M. Peterman, PhD
Scott Peterman, Ph.D., is Senior Product Marketing Manager for Life Science Mass Spectrometry of the Chromatography and Mass Spectrometry Division of Thermo Fisher Scientific.
A Greener HPLC Guide: How ACT Labels Help Promote Lab Sustainability
Beyond Blood: Sheffield Teaching Hospitals NHS Foundation Trust Develops HRAM-Powered Method for Post-Mortem Toxicology Analysis