Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Orbitrap / GC-MS/MS: A Confirming Method for Food Safety Analysis
GC-MS/MS: A Confirming Method for Food Safety Analysis
By , 11.23.2020
It’s a very uncertain world. Who knows what the future holds? Having certainty is important in everything we do, especially in analytical testing because the implications of being uncertain can be far reaching. Such dilemmas can include:
  • Is that compound really there?
  • What if I report a false positive?
  • Have I missed something?
  • How can I get confirmation?
All these need accurate answers, with speed and efficiency.
In analytical testing, instrumental methods often rely on triple quadrupole GC and LC-MS methods. These methods usually provide the sensitivity needed to meet regulatory requirements, but when they don’t, because of matrix interferences or when a confirming method is needed, orthogonal technology with enhanced specificity and sensitivity is needed. One lab that has taken the GC Orbitrap route is the Laboratori de l’Agencia de Salut Publica de Barcelona, who adopted a Q Exactive GC Orbitrap GC-MS/MS approach to confirming suspect triple quadrupole GC-MS results.
According to Dr. Nuria Cortes-Francisco, “the concern we had with our triple quadrupole methods was matrix interferences, where in some cases we were not confident in our results because the confirming ion ratio requirements weren’t met. Based on triple quadrupole method requirements, you would say it’s a negative, but with an obvious peak, we were concerned that the negative was false. In these cases, we confirm our triple quadrupole results using the Q Exactive GC Orbitrap GC-MS/MS system. With a triple quadrupole system, you only obtain the nominal mass so you can’t be sure if it’s the target compound or matrix interferences.”
Confidence and certainty in the results of food safety and environmental testing is of utmost importance. The Q Exactive GC Orbitrap GC-MS/MS system brings together the power of high-chromatographic resolution and HRAM Orbitrap MS to provide high-specificity, high-sensitivity target compound analyses when GC-MS triple quadrupole methods produce inconclusive results or cannot achieve required detection limits. When a confirming method is needed, the Q Exactive GC Orbitrap GC-MS/MS system is the orthogonal technology to apply.
Read more in this case study
Fairy light background image
When “Fairy Lights” Turn Toxic: The Hidden Hazard of Chlorinated Paraffins
Are you looking forward to the holidays and a lot of cozy ev...
Read More
GC Orbitrap Guru Series #4: Scott Borden: Deep Breaths in Breathomics
Hey there GC Orbitrap enthusiasts! Welcome to blog number 4 ...
Read More
Dr Araceli Rivera-Pérez
GC Orbitrap Guru Series #3: Dr Araceli Rivera-Pérez
Hey everyone! The response to my second blog with Brooklynn ...
Read More
Dr Kerstin Krätschmer
GC Orbitrap Guru Series: Dr. Kerstin Krätschmer
It’s been 10 years! Yes, in 2015 “gas chromatography...
Read More
Why Your Lab Should Upgrade Its HPLC or UHPLC System
What Would You Change If You Could Go Back in Time