Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Gas Chromatography / Helium Shortage 3.0
Helium Shortage 3.0
By Dr. Daniela Cavagnino, Product Marketing Manager, GC/Autosampler, Thermo Fisher Scientific 02.17.2020
“Helium Shortage 3.0 will likely ease in the second half of 2020, but that does not mean it’s going away anytime soon — in fact it will remain until 2021.” [www.gasworld.com]
helium-tanksDuring his closing keynote speech at gasworld’s MENA Industrial Gases Conference 2019 in Dubai, Phil Kornbluth, President of Kornbluth Helium Consulting, provided an update on the global helium business today and the status of its latest market imbalance, Helium Shortage 3.0.
“The shortage should ease a little during the second half of 2020,” he explained while pointing out that “ease doesn’t mean go away. It just means it won’t be quite so severe.”
There is a positive note in the key takeaways from Phil Kornbluth regarding helium availability, at least for the current year, however, we cannot forget the heavy implication of the increased cost of helium and much longer delivery times still impacting semiconductor and medical device industries, as well as analytical laboratories, especially in countries fully relying on imports for helium, like Korea.
For gas chromatographers, helium represents the most-used carrier gas for many applications. Within the gases which can be used as a mobile phase in a capillary GC or GC-MS, helium is offering the highest inertness combined with high diffusivity, a measure of the diffusion speed of a solute vapor in a given gas. The diffusion speed of the solute in the carrier gas determines the speed of the gas chromatographic process, allowing optimum higher flow rates and making the analysis time acceptable.
The shortage of helium is a recurrent problem many laboratories have faced for a few years now and need to overcome.
A possible alternative carrier gas in a capillary GC system is hydrogen, due to its diffusivity comparable with helium. Hydrogen is actually even preferable to helium because its viscosity is about half, permitting lower inlet pressure for a given gas velocity. Since the lower is the inlet pressure and the higher is the optimum gas velocity, hydrogen permits to maintain the optimum separation efficiency at higher flow rates, providing shorter analysis time. Additionally, the availability of hydrogen generators provides a safe, steady, and renewable supply of hydrogen for proper instrument operation.
So, why are laboratories not simply getting rid of helium in favor of hydrogen?
Moving from helium to hydrogen to run GC and GC-MS systems is the approach many laboratories are taking with the benefits of faster analyses and reduced cost of operation.
Nevertheless, there are still two important arguments against the use of hydrogen: one concerns safety and the other is related to many existing validated methods based on helium which must be re-optimized and re-validated in the case of swapping carrier gas.helium-saver-modules
Modern technology allows laboratories that need to maintain helium as a carrier gas, to significantly reduce helium consumption, alleviating the pain of costs and supply shortage.
The Thermo Scientific iConnect Helium Saver Split Splitless (SSL) injector module (pictured on the right, U.S. Patent 8,371,152) is a proprietary technological innovation available on the Thermo Scientific Trace 1300 series GC, offering helium gas-saving while keeping the existing validated analytical methods.
Unlike other gas-saving solutions, the Helium Saver SSL Module is always ready and works while your instrument is running, as well as when it is idle, providing real 24/7 savings operation.
How much helium can I save?
Considering that the majority of the carrier gas consumption is due to splitting the sample during the injection and keeping the injector purged during the analysis, think about how much helium you can save if those operations are performed with nitrogen, feeding helium only for the limited flow through the capillary column for the separation process. Learn more about how the Helium Saver SSL works by watching this video.
If you are wondering how much helium you can save, just consider that a cylinder of helium can last from 3.5 to 14 years, depending on how continuously the GC or GC-MS is used. Try the Helium Saver Calculator to discover how much you can save based on your method and sample-throughput.
Thanks to the Trace 1300 GC modular design, the iConnect Helium Saver is a plug and play module, easily installable by the user at any time. Saving helium has never been easier and more effective while keeping your methods intact, validation-effective, and regulation-compliant.
Additional resources
    1. AN 10441: Helium conservation in volatile organic compound analysis using U.S. EPA Method 8260
    2. AN 10522: Optimized GC-MS solution for semivolatiles (SVOC) analysis in environmental samples in co…
    3. PN 10410: Helium Carrier Gas Conserving Inlet for Gas Chromatography
Fairy light background image
When “Fairy Lights” Turn Toxic: The Hidden Hazard of Chlorinated Paraffins
Are you looking forward to the holidays and a lot of cozy ev...
Read More
wine bottle image
New European Wine Scandal on PFAS in Wine
Introduction PFAS, also known as “eternity chemicals&#...
Read More
Chromeleon CDS: Hints and tips to help optimize your GC-MS workflows
Chromeleon CDS: Hints and Tips to Help Optimize your GC-MS Workflows
Gas Chromatography-Mass Spectrometry (GC-MS) is a workhorse ...
Read More
Pesticides
Pesticide Residue Analysis in the 21st Century: Innovations in Sample Preparation and Detection Using the Thermo Scientific TSQ 9610 GC-MS/MS
Introduction The accurate detection of pesticide residues in...
Read More
Dr. Daniela Cavagnino
Dr. Daniela Cavagnino is Product Marketing Manager for GC and Sampling Solutions. She started her career in gas chromatography at Thermo Fisher Scientific spending several years in the R&D laboratories working on GC technology innovation. Then, she conveyed her technical background into product management and marketing management roles with more than 20 years’ experience in promoting GC/GCMS technology and applications in several different market segments.
Dioxin Analysis Meets Speed and Compliance NofaLab Laboratories
An ICP-MS Expedition through the Elements: Part 1