Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Orbitrap / You Try to Eat Healthier, But How to Avoid a Bad Taste in Your Mouth?
You Try to Eat Healthier, But How to Avoid a Bad Taste in Your Mouth?
By Petra Gerhards, Regional Marketing Manager GC and GC/MS, EMEA, Thermo Fisher Scientific 01.04.2023
We are all familiar with the high-calorie dishes during the holidays. And many of them are made with meat, adding fat as well. That’s why a lot of people go vegan in January. But are you sure you want to do a vegan January? You may have the best of intentions, but how do you get past the bad taste of some of the products?
How do off-flavors form?
Most of today’s vegan products are plant-based, but that doesn’t always mean they taste good. When it comes down to salt, fat and other content, there are many published articles that question the healthiness of vegan foods. This is, mostly, because plants don’t contain a lot of fat. However, fat is the carrier for flavors. The food industry spends billions of dollars to make a vegan burger taste “umami,” which describes the taste of meat.
“Umami, which is also known as monosodium glutamate, is one of the core fifth tastes: sweet, sour, bitter and salty. Umami means ‘essence of deliciousness’ in Japanese, and its taste is often described as the meaty, savory deliciousness that deepens flavor.” [1]
There are millions of different vegan foods that made it to market during the last decade. However, their unpleasant, intrinsic flavor and odor have been the major obstacles limiting consumers’ acceptance. These off-flavors are often described as “green,” “grassy,” “beany,” “fatty” and “bitter.”
Highlighting off-flavor volatiles
This review highlights the presence and formation of common off-flavor volatiles (aldehydes, alcohols, ketones, pyrazines, furans) and non-volatiles (phenolics, saponins, peptides, alkaloids) from a variety of plant-based foods, including legumes (e.g., lentil, soy, pea), fruits (e.g., apple, grape, watermelon) and vegetables (e.g., carrot, potato, radish). These compounds are formed through various pathways, including lipid oxidation, ethanol fermentation and Maillard reaction (and Strecker degradation). [2]
Since off-flavors are a big topic, there are industries that spend millions of dollars to cover up for these issues. Functional ingredients and masking are common techniques to deal with off-flavors. [3] But it is not easy to betray human tastebuds. A masked rancid taste might end up as a “coconut flavor,” which is also not wanted in a vegan burger.
Now, you might think that 99 percent cocoa-based chocolate is vegan, and you are right. But free of off-flavors? The answer is no!
“It was found that 2-methoxyphenol, 3- and 4-methylphenol, 3- and 4-ethylphenol and 3-propylphenol are responsible for the ham-like smoky off-flavor. The moldy, musty off-flavor is mainly caused by geosmin and the coconut-like off-flavor by massoialactone.” [4]
The list of off-flavors is endless. A brand of orange juice can be considered “good” in terms of following food regulations; however, it may render a bitter aftertaste. This might come from oxidation. The oxygen penetrates the packaging into the juice and is responsible for the bitterness. So other industries, such as packaging, are involved in testing for off-flavors as well.
Contamination in vegan food
Off-flavors aren’t the only issue. A human doesn’t eat what tastes strange or bad. But some things you can’t taste, and they also are not healthy. Recently, there were very high concentrations of MOSH/MOAH (mineral oil saturated hydrocarbons/mineral oil aromatic hydrocarbons) in Germany in pesto, olive oil, chocolate Santa Clauses and vegan cold cuts. If you would like to learn more, click on the related blog post.
The difficulty with off-flavors is always the concentration — the peaks are very small. When it comes to sensitivity, the human nose detects off-flavors to a higher degree compared to a GC-MS. This is why chemists around the world spend a lot of time trying to find out how to detect off-flavors to the highest level of sensitivity.
If this is one of the issues you are trying to resolve, join one of our free webinars on off-flavors:
On-demand webinar: Off-flavor analysis in food — in German:
On-demand webinar: Off-flavor analysis in food — English:
If you would like to learn how to detect off-flavors immediately, read this blog post:
Conclusion
Resolutions to eat healthier are great! But some “healthy” options taste better than others. Consuming products that are not heavily processed— or leading a simple vegetarian lifestyle— can help you avoid having to deal with weird off-flavors.
The Thermo Fisher Scientific team wishes you all the best for 2023, and we look forward to seeing you in the community for more interesting topics this year.
Resources
[1] https://dictionary.cambridge.org/dictionary/english/umami
[2] https://www.researchgate.net/publication/360818318_Surmounting_the_off-flavor_challenge_in_plant-bas…
[3] https://www.firmenich.com/taste-and-beyond/nutrition-and-wellbeing/meat-dairy-analogs
[4] https://www.laborpraxis.vogel.de/fehlaromen-in-schokolade-auf-der-spur-a-020b42de7bbd765bb81ee07d564…
Fairy light background image
When “Fairy Lights” Turn Toxic: The Hidden Hazard of Chlorinated Paraffins
Are you looking forward to the holidays and a lot of cozy ev...
Read More
GC Orbitrap Guru Series #4: Scott Borden: Deep Breaths in Breathomics
Hey there GC Orbitrap enthusiasts! Welcome to blog number 4 ...
Read More
Dr Araceli Rivera-Pérez
GC Orbitrap Guru Series #3: Dr Araceli Rivera-Pérez
Hey everyone! The response to my second blog with Brooklynn ...
Read More
Dr Kerstin Krätschmer
GC Orbitrap Guru Series: Dr. Kerstin Krätschmer
It’s been 10 years! Yes, in 2015 “gas chromatography...
Read More
Petra Gerhards
Petra Gerhards, Dipl-Ing, is Regional Marketing Manager of GC and GC-MS for EMEA at Thermo Fisher Scientific. She has more than 29 years of experience in the fields of GC-MS, SPE and LC-MS. Since joining the regional team she has contributed to workflow solutions combining vials and closures with SPE solutions, GC-MS and LC-MS. She works with KOL's on data for regional specific marketing campaigns, organizes in-house seminars and works on customer specific solutions. Her main expertise is in the field of doping and drugs-of-abuse analysis.
Could the Sodium-Ion Battery Eventually Displace Lithium?
Biopharmaceutical Characterization: A Biopharmaceutical Basics Overview