Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Isotope Ratio MS / How to Minimize Sample Preparation and Optimize Sr Isotope Ratio Analysis
How to Minimize Sample Preparation and Optimize Sr Isotope Ratio Analysis
By Dr. Claudia Bouman, Product Manager, Multicollector ICP-MS and Noble Gas MS, Thermo Fisher Scientific, Bremen, Germany, 04.16.2024
Strontium has 4 isotopes, 84Sr, 86Sr, 87Sr and 88Sr. One of them is non-natural (87Sr) and is produced by the radioactive decay of 87Rb. In addition, elemental strontium behaves very similar to calcium. So we have two characteristics, age and chemical composition, that are helping scientists understand various processes on Earth. Besides applications in geosciences, strontium isotope ratios also are used in archeology, forensics and food authentication, where they provide a link to the land where food was grown or grazed.
Thorough laboratory work is required to extract strontium from a sample before it can be taken to the mass spectrometer for analysis. Classically, this has been done by digestion and purification using ion exchange methods, which is labor-intensive and requires a very clean environment to keep blank levels to a minimum.
What if you could reduce lab work and minimize sample preparation for your strontium isotope ratio analysis?
The new Thermo Scientific Neoma MS/MS MC-ICP-MS  can make this happen. The MS/MS option of the Neoma MC-ICP-MS consists of a patented hexapole collision-reaction cell with a novel pre-cell mass filter. The collision reaction cell allows for the separation of the isobaric interference of 87Rb+ from 87Sr+ using SF6 gas (Sr reacts with F, whereas Rb doesn’t), while the pre-cell mass filter allows a certain mass window to enter the collision-reaction cell thereby reducing the potential for spectral and matrix interferences.
The Journal of Analytical Atomic Spectrometry has published the first paper on radiogenic Sr isotope ratio measurements with the Neoma MS/MS MC-ICP-MS, demonstrating the potential of the new MS/MS technology for analyzing samples without the need for extensive sample preparation. The authors analyzed Sr isotope ratios in a series of geological reference materials and found both precision and accuracy to be equivalent to those obtained by MC-ICP-MS with prior Sr extraction and sample purification. Along with mass fractionation following exponential law, the new Neoma MS/MS MC-ICP-MS opens the door to a wider suite of isotope applications while minimizing sample preparation.
Ready to break through the noise with the unique MS/MS technology?
Download our infographic to learn more about how the pre-cell mass filter works.
On LinkedIn? Visit our LinkedIn page #IsotopeRatioMS #IsotopeAnalysis #Strontium
mannequin wearing necklace
Cheap, Trendy – and Toxic? High Levels of Cadmium in Necklaces Have Been Reported Lately
Valentines Day is an opportunity to show your partner (or yo...
Read More
From Complexity to Clarity: Smarter PFAS Identification Workflows in Compound Discoverer Software
Per- and polyfluoroalkyl substances (PFAS) have become a maj...
Read More
Dr. Magnus Ericsson
Inside Laboratoire Anti-Dopage Français (LADF): An Interview with Director Dr. Magnus Ericsson
At the forefront of global anti-doping efforts, the Laborato...
Read More
Advancements in Thorium Isotope Analysis: The Role of Neoma MS/MS MC-ICP-MS
Thorium (Th) isotope analysis is important across various sc...
Read More
Dr. Claudia Bouman
Claudia Bouman is Product Manager at Thermo Fisher Scientific in Bremen, Germany, responsible for the Multicollector ICP-MS and Noble Gas MS product line. Claudia graduated from the University of Utrecht and completed her Ph.D. in Isotope Geochemistry at the VU in Amsterdam, Netherlands. In 2002, Claudia joined the Bremen factory to work as an Application Specialist. Since then, she had various roles in Product Management, Sales Support and Marketing.
Tired of HPLC Downtime? Time for a Technology Boost?
Enabling Science, Industry to Protect the Planet