Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Environmental / IC-MS/MS Now Approved for Polar Pesticide Analysis in Food by an EU Reference Laboratory
IC-MS/MS Now Approved for Polar Pesticide Analysis in Food by an EU Reference Laboratory
By AnalyteGuru Staff, 03.11.2021
Every week I come across a new article relating to pesticides. Just this week I was reading about a peer-reviewed study in Germany that shows glyphosate and dozens of other pesticides can travel through air into our national parks and cities miles away from the site of spraying. This means pesticides are not only entering the food chain, but also polluting sites of natural beauty and entering the air we breathe.
The pesticide crisis continues. Earlier in 2021, Greta Thunberg criticized the UK government for allowing the use of a potent ‘bee-killing’ pesticide, which the European Union had already banned. In France, President Macron vows to push forward the phasing out of pesticides during his presidency term. With mounting pressure coming from a vast number of environmental charities and the EU Farm to Fork Strategy, which commits to reducing pesticide use by 50 percent by 2030 as part of the European Green Deal.
While it is encouraging to have these targets, pesticides are a problem now and analysis of pesticides in our food is challenging, due mainly to the diverse sample matrices of all manner of food types.
Glyphosate is one of the most commonly used pesticides and one of the most difficult to analyze. Highly polar and ionic, and insoluble in organic solvents and no naturally occurring chromophore, glyphosate doesn’t loan itself well to traditional HLPC.
The good news is that glyphosate can readily be analyzed by ion chromatography, and recently an updated IC-MS/MS method was included in an EU reference laboratory single residue method (EURL-SRM).
Even better news: The method is utilizing the Thermo Scientific„¢ Dionex„¢ IonPac„¢ AS19 column, and the coverage of polar pesticides is higher than any other method.
The method is titled:
€Quick Method for the Analysis of Highly Polar Pesticides in Food Involving Extraction with Acidifie…
Method 11 (M11) version 12 gives the €œProposed IC-MS/MS conditions for Glyphosate, AMPA, N-Acetyl-Glyphosate, Ethephon, HEPA, Glufosinate, MPPA, Fosetyl-Al, Cyanuric acid, Bromide, Chlorate, Perchlorate, Phosphonic acid and TFA and utilizes Thermo Scientific„¢ Dionex„¢ Eluent Generation technology.€
The previous version, 11.1 utilized the Dionex„¢ IonPac„¢ AS11 column, but with LC-MS technology.
The inclusion of IC-MS/MS and updated column chemistry with the IonPac„¢ AS19 means that this is an approved and recommended technique and gives the most coverage of the separation of the polar pesticides listed and meets EU regulations in SANTE/11945/2015. The use of eluent generation technology means users can benefit from ultra-pure, highly reproducible eluents made inline by the system €” meaning no more manually prepared, tricky buffer solutions.
With the IC-MS/MS method, no inline prefilters are needed, which are commonplace for HPLC methods, and the method comes with a section of handy hints to help get you started.
In this video interview, Ann-Kathrin Wachtler from CVUA Stuttgart, a European reference laboratory for pesticides analysis, talks about the challenges of polar pesticide analysis, their initial reluctance to use ion chromatography as an unknown technology to their lab, and how IC helped overcome the challenges they faced leading to the inclusion in the EURL-SRM.
For more on pesticide analysis, including the anionic pesticide explorer solution, visit thermofisher.com/pesticides.
Analyteguru.com.jpg
Scientists are looking at data charts on a computer with test tubes beside them.
Ion Chromatography Turns 50: Chromeleon Software’s Tribute to a Lifelong Partner
This year marks a special milestone: the 50th anniversary of...
Read More
Pesticides
Pesticide Residue Analysis in the 21st Century: Innovations in Sample Preparation and Detection Using the Thermo Scientific TSQ 9610 GC-MS/MS
Introduction The accurate detection of pesticide residues in...
Read More
IC Golden Jubilee blog header image
What is the Ion Chromatography Golden Jubilee? Celebrating 50 Years of IC Innovation
The year 2025 marks the golden jubilee of ion chromatography...
Read More
New Column for Fast Analysis of Haloacetic Acids, Bromate and Dalapon Using Ion Chromatography Coupled with Mass Spectrometry
New Column for Fast Analysis of Haloacetic Acids, Bromate and Dalapon Using Ion Chromatography Coupled with Mass Spectrometry
Haloacetic acids (HAAs) are disinfection byproducts (DBPs) f...
Read More
AnalyteGuru Staff
Organic Acid Determinations in Beer: Leveraging High-Pressure IC for Enhanced Resolution and Quantitation
Reagent-Free IC Keeps Your Analysis Simple