Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Industrial / Is There a Better Way to Determine Anionic Impurities in Sulfuric Acid?
Is There a Better Way to Determine Anionic Impurities in Sulfuric Acid?
By Gary He, Product Marketing Manager, Gallery Discrete Analyzers, IC Industrial markets, Sample Prep, Thermo Fisher Scientific 10.25.2023
4 things to know about two new analytical methods
Did you know that sulfuric acid is produced and consumed more than any other chemical in the world? It’s true! Sulfuric acid is also one of the oldest known industrial chemicals. That’s because this strong inorganic acid has qualities that make it very useful for a large number of industries, including fertilizers, inorganic chemical manufacturing, petroleum refining, pharmaceuticals, soaps and detergents, textiles, pulp and paper, metals, car batteries, and pigments and dyes.
However, determining impurities in sulfuric acid comes with its share of challenges. Here are four things to know about identifying sulfuric acid:
1. Trace impurities in sulfuric acid matter
Trace ionic impurities in concentrated sulfuric acid can have a direct impact on the yield and reliability of chemical and semiconductor material processes, such as water cleaning.
2. Determining impurities is no cake walk
The determination of anionic impurities in concentrated acids has been an analytical challenge for years. Why? The titration method is cumbersome, tedious and uses hazardous chemicals.
3. Ion chromatography (IC) is a promising alternative, but presents challenges for sulfuric acid
IC is a promising alternative technique to determine low levels of anionic impurities. However, while low concentrations of contaminating anions in weaker acids such as hydrofluoric acid and phosphoric acid can be determined using online ion-exclusion sample preparation, that approach cannot be used for a strong acid like sulfuric acid. In sulfuric acid, the high concentration of sulfate limits the amount that can be injected into the anion-exchange column, thus limiting method sensitivity.
4. Two methods offer a fresh approach to determining impurities in sulfuric acid
In recent work by a team at Thermo Fisher Scientific, two experts borrow knowledge from determining anionic contamination in another strong acid, nitric acid. The team developed a similar approach for sulfuric acid whereby the concentrated sulfuric acid sample is diluted, and a volume that has been empirically determined to avoid overloading a high-capacity column is injected. Here’s a breakdown of the two methods:
  • Method 1 uses a Thermo Scientific™ Dionex™ IonPac™ AS19 column with a hydroxide eluent to accurately determine fluoride, chloride, bromide, nitrite, and nitrate. This method is ideal for separating a wide range of inorganic anions with high sensitivity.
  • Method 2 uses a Thermo Scientific™ Dionex™ IonPac™ AS23-4μm column with a carbonate/bicarbonate eluent to determine phosphate. The carbonate eluent is ideal for determining phosphate in sulfuric acid because it elutes before sulfate.
The team published the two methods and evaluated key performance parameters (including response linearity for each analyte, limits of detection, and accuracy) in a new application note.
To learn more about the methods and the team’s findings, read the application note.
Temperature regulation graphic image
How to Get More Accurate HPLC Quantitation of Semi-Volatiles
Struggling to accurately measure semi-volatile analytes? Str...
Read More
freedom to fragment
Freedom to Fragment: Introduction
The importance of fragmentation in mass spectrometry 1. What...
Read More
Stay Ahead in Developing Green Energy Solutions with FAME Analysis for Jet Fuel
Biofuels, particularly biodiesel, are an attractive, cleaner...
Read More
New Column for Fast Analysis of Haloacetic Acids, Bromate and Dalapon Using Ion Chromatography Coupled with Mass Spectrometry
New Column for Fast Analysis of Haloacetic Acids, Bromate and Dalapon Using Ion Chromatography Coupled with Mass Spectrometry
Haloacetic acids (HAAs) are disinfection byproducts (DBPs) f...
Read More
Gary He
Gary He is a Product Marketing Manager at Thermo Fisher Scientific, overseeing the Gallery discrete analyzers, Dionex ion chromatography (IC) in industrial markets, and sample prep. In this role, he leads marketing programs aimed at fostering the sustainable adoption of technology in well-established applications like water and inorganic contaminants analyses. Additionally, Gary focuses on advancing growth applications by implementing IC workflows in areas such as clean energy and lithium-ion battery testing.
New Dimensions in Isotope Ratio Analysis: Site Specificity
Decoding Amino Acids Using Ion Chromatography