Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Battery Material Analysis / Lithium-Ion Battery Production: The Analytical Techniques Used
Lithium-Ion Battery Production: The Analytical Techniques Used
By Simon Nelms, 05.09.2023
The surge in demand for high-performance batteries is driving significant investment in every stage of production. Laboratories associated with battery research and manufacturing need material analysis solutions in each step of the process, all the way from raw material production to recycling spent batteries.  To meet the analysis requirements, a wide range of instruments, software and related technologies is required.
Analytical techniques across the battery supply chain
Research
This work involves development of next-generation battery materials and improvements in existing lithium battery technologies.
Lithium-ion battery
Processes includeAnalytical needAnalytical techniques
Novel cathode active materials researchElectrolyte, cathode and anode material degradation product studiesExploration of new methods and improvement of existing techniques for battery material recyclingDevelopment of next-generation batteries (e.g., solid state, sodium) Material composition and purity characterizationFailure analysis      ICP-OESICP-MSIC/IC-MSLC/LC-MSGC-GC-MSHR-ICP-MSGD-MSOEA 
Raw materials
This step of the supply chain involves mining, processing and refining raw materials, such as lithium, manganese, nickel, cobalt and iron salts, graphite and silicon.
Processes includeAnalytical needAnalytical techniques
Mined material processingRefined product purity analysisQuality assurance and control  Bulk process control and quality control (QC)Mineral, ore and brine analysis for lithium and impurity element analysisConfirmation of material quality ICP-OESICP-MSIC/IC-MSOEA 
Cell components and assembly
In this stage, a variety of processes, including development of novel chemistries, anodes, cathodes, electrolytes and separators, as well as on-line inspection are required. This step also covers mixing of materials, coating, drying, calendering and stacking of battery jelly rolls, and packaging components into pouch, prismatic or cylindrical forms.
Processes includeAnalytical needAnalytical techniques
Manufacturing of battery cells and/or cell arraysElectrolyte composition confirmation        Ensure quality of designProcess controlIn-line inspectionFailure analysis and material rejectionComposition measurementDetection and quantification of elemental impuritiesDegradation studiesResearch and routine QA/QC measurementICP-OESICP-MSGC/GC-MSIC/IC-MS       
Battery testing
During this phase, manufacturers need to ensure that the finished battery cells meet specification before they are assembled into battery packs.
Processes includeAnalytical needAnalytical techniques
Failure analysis and rejectionBattery aging, environmental and electrical studies  Analysis of degradation products:Cathode material compoundsElectrolytesSeparator degradation productsICP-OESICP-MSLC/LC-MSGC/GC-MSIC/IC-MS
Battery recycling
This final stage of the battery supply chain incorporates bulk recycling and recovery of cathode and anode materials (the so-called ‘black mass’) and includes recycling of both end-of-life batteries and manufacturing scrap from failed cells to retrieve and refine the material for use in new batteries.
Processes includeAnalytical needAnalytical techniques
Material recoveryInspection and reuseBattery recyclingTesting  InspectionCharacterization    ICP-OESICP-MSHPLCIC/IC-MSLC/LC-MSGC/GC-MS
 What are the analytical techniques?
ICP-OES – Inductively coupled plasma optical emission spectrometry
ICP-MS – Inductively coupled plasma mass spectrometry
HPLC – High performance liquid chromatography
LC-MS – Liquid chromatography mass spectrometry
IC – Ion chromatography
GC – Gas chromatography
GC-MS – Gas chromatography mass spectrometry
IC-MS – Ion chromatography mass spectrometry
HR-ICP-MS – High resolution inductively coupled plasma mass spectrometry
GD-MS – Glow discharge mass spectrometry
OEA – Organic elemental analyzer
Why these analytical techniques?
Ion chromatography – By using ion chromatography, researchers and scientists can confirm the anionic composition and purity of electrolyte solutions and gain insights into degradation mechanisms in lithium-ion batteries to ensure product quality during manufacturing.
Gas chromatography – Gas chromatography-mass spectrometry (GC-MS) systems can reach lower detection limits and obtain richer information with sensitive detection and accurate mass-fragment and molecular ion identification. By identifying and quantifying compounds formed during electrolyte aging, GC-MS systems can provide new insight into Li-ion battery degradation.
Mass spectrometry – When studying lithium-ion battery components, detection using mass spectrometry (MS) dramatically extends the capabilities of ion and liquid chromatography (IC and HPLC) systems and provides higher sensitivity, accurate quantitation, peak confirmation and evaluation of chromatography peak purity, improved resolution of complex samples with seamless integration of MS data into IC and HPLC workflows.
Elemental analysis – Long before lithium-ion batteries are produced, laboratories use elemental analysis to determine the composition and purity of lithium-rich brine and lithium ores which will eventually be refined into battery component materials. The refined materials also require elemental analysis to ensure their composition and purity before use, since the presence of impurities significantly degrades the performance and lifetime of the battery.
Liquid chromatography – Whether you’re analyzing electrolyte solvents or working to achieve the next breakthrough in battery recycling, liquid chromatography solutions can improve your productivity, deliver a maximum return on your investment, and when coupled with mass spectrometry provide confident insights to drive business decisions.
Additional Information
To learn more about battery research and manufacturing, visit our website.
Automated production line filling and transporting medical injection pens in a factory.
Stability-Indicating HPLC-UV Method for Semaglutide Impurity Analysis
As demand for GLP-1 receptor agonists like semaglutide conti...
Read More
pfas image
Understanding PFAS in Pharmaceutical Manufacturing
Introduction to PFAS Per- and polyfluoroalkyl substances (PF...
Read More
Cutting-Edge Triple Quad ICP-MS for Clinical Trace Element Analysis
Cutting-Edge Triple Quad ICP-MS for Clinical Trace Element Analysis
Unlocking the power of triple quadrupole ICP-MS for clinical...
Read More
The Benefits of Charged Aerosol Detection (CAD) in High-Performance Liquid Chromatography (HPLC)
Episode 1: The Benefits of Charged Aerosol Detection (CAD) in High-Performance Liquid Chromatography (HPLC)
High-Performance Liquid Chromatography (HPLC) is a cornersto...
Read More
Simon Nelms
HPLC Purification: When to Use Analytical, Semi-Preparative & Preparative Methods
Top 7 Benefits of Using HPLC for Manufacturing Quality Control