Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Mass Spectrometry / Modern Technologies for High-Performance LC-MS Data Transfers
Modern Technologies for High-Performance LC-MS Data Transfers
By Chris Knowles, Product marketing manager: Chromatography and mass spectrometry software 10.01.2025
High-resolution LC-MS workflows generate massive files that can quickly bottleneck traditional transfer methods. The primary challenge is not just the sheer volume of data, or the storage and processing power required, but also how it is collected, formatted, structured and annotated.
Image depicting data transfer
To address this challenge, advanced protocols and orchestration platforms must enable secure, global, multi-gigabit transfers without manual intervention.
This blog explores the technologies that facilitate efficient and reliable data transfers in LC-MS environments, focusing on WAN accelerators, on-premises transfer appliances, cloud-native streaming, and why the Thermo Scientific Ardia Platform might be the perfect choice for the challenges faced in today’s labs.
WAN accelerators
WAN accelerators improve data transfer performance over high-latency, high-loss networks by replacing or optimizing traditional transport protocols.
IBM Aspera FASP uses a UDP-based protocol to sustain high throughput, automatically recover from packet loss, and encrypt all data in motion.
While not a WAN accelerator in the strictest sense, Globus GridFTP extends standard FTP with features like parallel data streams, third-party transfers, and strong authentication, making it highly effective for moving large scientific datasets across global networks.
Check out this post from Globus:  GridFTP: A Brief History of Fast File Transfer
On-premises transfer
The purpose of an on-premises data transfer solution is to detect new files and initiate validated transfers to central storage without manual oversight. 
The key benefit is helping to integrate extensive historical datasets with current databases, making legacy data usable with modern tools, including AI. This integration is crucial for leveraging older data, ensuring it can be analyzed alongside new data to uncover valuable insights and improve reproducibility
Cloud-native streaming
Cloud-native streaming directly into object storage, like Amazon S3 (Simple Storage Service) or Microsoft Azure Blob Service enables compute services to start processing immediately upon file arrival. Serverless event triggers and lifecycle policies further automate tiered storage and archival.
This approach not only accelerates data processing but also facilitates the integration of diverse datasets, ensuring that both new and legacy data can be efficiently managed and analyzed. By leveraging cloud-native streaming, laboratories can enhance data accessibility and scalability, supporting advanced analytics and AI/ML applications.
However, it is important to consider the associated costs and data security risks. Cloud storage and processing can incur significant expenses, and robust security measures must be implemented to protect sensitive data from breaches and unauthorized access.
Would you trust cloud services to securely store and manage your clinical data?
Is your lab ready to move all your data to the cloud and pay a subscription for the benefits?
Thermo Scientific Ardia platform
The Thermo Scientific Ardia Platform offers a robust data management and orchestration engine. It routes RAW files from instruments to processing nodes and archives, logs every action for audit readiness, and scales across hybrid environments.
The Ardia platform has dedicated tools specific to LC-MS as a physical server with cloud-based technologies for scaling end to end processing and connecting labs around the world.
Proteomics users are finding significant success with this platform as an end-to-end solution that leverages the latest technologies, including AI for advanced data analysis and robust security measures to protect sensitive data.
More information
Thermo Scientific Ardia platform for proteomics.
Don’t miss our on-demand webcast: “Cloud computing in the analytical lab: The strategic risks, challenges and opportunities to consider.” Register now to access the webcast and learn how to future-proof your lab with cloud technology.
Check out this recent white paper Centralising analytical data from mass spectrometry in drug discovery and development | Scientific Computing World
Visit us on LinkedIn: #ArdiaPlatform #DataManagement #Informatics
Scientist holding a test tube beside industrial lab equipment and glassware.
Monitoring mAb Manufacturing the Easy Way with Switchable 2D‑LC-UV-MS
Why are switchable 2D-LC workflows fit for mAb manufacturing...
Read More
Upgrade Smarter: Trade In Your Current LC-MS System and Boost Lab Performance Without Breaking Budget
Is your current mass spectrometer limiting your lab’s capa...
Read More
Pre-configured LC_MS workflow solutions
Maximize Performance with Specialized LC-MS Workflows — and Save Up to 40%!
When it comes to clinical research, precision, reliability, ...
Read More
Automated LC-MS Sample Cleanup Using Turbulent Flow Chromatography
Too much interfering matrix? Discover how to streamline your...
Read More
Chris Knowles
Chris Knowles began his career with a PhD in mammalian cell line engineering and over a decade as a Principal Scientist and applications specialist in biopharma and bioprocessing. With deep expertise in software solutions for LC-MS, MS/MS, and advanced mass spectrometry techniques, he focuses on biologics production and analytics, including emerging areas like gene therapy, oligonucleotide therapeutics, and viral vectors. Now, as a Product Marketing Manager for BioPharma Software at Thermo Fisher Scientific, Chris is dedicated to helping customers overcome analytical challenges by delivering powerful, compliant, and user-friendly software solutions.
Scaling Up Biopharmaceutical Characterization with the Orbitrap Ascend BioPharma Tribrid Mass Spectrometer
How to Get More Accurate HPLC Quantitation of Semi-Volatiles