Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / GC-MS / Moving to Hydrogen or Staying With Helium? A Solution to Support Both Choices
Moving to Hydrogen or Staying With Helium? A Solution to Support Both Choices
By Dr. Daniela Cavagnino, Product Marketing Manager, GC/Autosampler, Thermo Fisher Scientific 09.26.2022
Disruption in the helium supply chain continues to pressure analytical laboratories to find alternative solutions for their carrier gas requirements. The choice of the most suitable carrier gas for a specific GC or GC-MS application should keep into consideration some basilar concepts, with the ultimate purpose to separate the analytes of interest in a relatively short time and preserving the sample integrity for quantitative analysis.
Disruption in the helium supply chain continues to pressure analytical laboratories to find alternative solutions for their carrier gas requirements. The choice of the most suitable carrier gas for a specific GC or GC-MS application should keep into consideration some basilar concepts, with the ultimate purpose to separate the analytes of interest in a relatively short time and preserving the sample integrity for quantitative analysis.
Gas viscosity and diffusivity impact the separation efficiency and, consequently, corresponding optimum linear velocity, as represented by the Golay equation for the most common carrier gases: helium, hydrogen and nitrogen (Figure 1).
Figure 1: Golay curves for carrier gasses.
Other considerations are, of course, the inertness of the carrier gas, to avoid possible reaction with the sample or the stationary phase; the purity of the gas, limiting contaminants like oxygen or water, which are detrimental for the stationary phase; or organic impurities responsible for a high signal background.
Finally, it must be compatible with the detector in use. For example, different considerations need to be done if using a GC standalone or a GC-MS.
Helium has been — and still is — the most used carrier gas, combining high separation efficiency with high inertness and extended compatibility, however, its shortage and price increase is forcing laboratories to find alternatives when possible. Hydrogen is definitely the best alternative, offering high separation efficiency in a shorter analysis time, while nitrogen has a much lower diffusivity, leading to much longer analysis time. Nitrogen is a good alternative if some resolution can be sacrificed, but it is not an option when working with a GC-MS, since it is ionized very easily.
Migrating to hydrogen appears to be the most convenient approach to overcoming the helium shortage crisis: It offers high speed of analysis at a low cost … and it is sustainable!
However, hydrogen is flammable, which poses safety risk concerns in the laboratory. Additionally, it is not an inert gas. Possible reactions have to be considered, especially in a hot injector or when working with a mass spectrometer detector.
Obviously, there are practical benefits in maintaining helium as a carrier gas, such as maintaining existing validated methods, maintaining unchanged GC-MS analytical performance and avoiding concerns about safety.
When helium is the best (or only) option, reducing consumption becomes mandatory.
Thermo Fisher Scientific developed a new option for its iConnect Split/Splitless inlet when mounted on the Thermo Scientific™ TRACE™ 1600 Series GC, the Thermo Scientific™ HeSaver-H2Safer™1. It offers several benefits, whether saving on helium consumption is required or hydrogen is a viable choice for the laboratory.
In the HeSaver mode, HeSaver-H2Safer extends the lifetime of a He cylinder 350% under 24/7 GC operation2 by limiting the maximum flow rate to fewer mL/min during analysis — just enough to feed the column for the chromatographic separation.
Using the H2Safer mode ensures safe operations without the need to install a H2 sensor, thanks to the limited maximum flow rate — even in case of column breakage. The fixed and limited consumption of hydrogen is also advantageous for optimum use of the hydrogen gas generators.
Whether you choose to migrate to hydrogen, or you prefer to reduce helium consumption, the HeSaver-H2Safer option is the solution for intelligent carrier gas management.
Learn more about how it works
  1. TN001218 – Addressing gas conservation challenges when using helium or hydrogen as GC carrier gas
  2. Helium Saver Calculator | Thermo Fisher Scientific – IT
Related Content
  • Helium Shortage and Price Spikes: Which is More Concerning?
  • Video – A better way to manage gas consumption with the HeSaver-H2Safer technology
  • What Good is Conserving Helium Carrier Gas Only When Your GC System is Idle?
Ocean to the laboratory; story of microplastics
From the Ocean to the Laboratory: How Scalable Pyrolysis-GC/MS Reveals the Hidden Story of Microplastics
Plastics have become part of everything we touch — from th...
Read More
Fairy light background image
When “Fairy Lights” Turn Toxic: The Hidden Hazard of Chlorinated Paraffins
Are you looking forward to the holidays and a lot of cozy ev...
Read More
MOSH MOAH image
MOSH/MOAH Analysis: Technology, Regulation, and the Road Ahead
Mineral oil hydrocarbons (MOSH) and mineral oil aromatic hyd...
Read More
Pesticides
Pesticide Residue Analysis in the 21st Century: Innovations in Sample Preparation and Detection Using the Thermo Scientific TSQ 9610 GC-MS/MS
Introduction The accurate detection of pesticide residues in...
Read More
Dr. Daniela Cavagnino
Dr. Daniela Cavagnino is Product Marketing Manager for GC and Sampling Solutions. She started her career in gas chromatography at Thermo Fisher Scientific spending several years in the R&D laboratories working on GC technology innovation. Then, she conveyed her technical background into product management and marketing management roles with more than 20 years’ experience in promoting GC/GCMS technology and applications in several different market segments.
Do All Your Yogurts Taste the Same? If Not, You Need a GC Orbitrap
How to Avoid Frustration When Looking for HPLC Information