Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Chromatography Data System / Reduce Sample Waste in the Lab Using Real-Time Data Evaluation with Automated Reactions
Reduce Sample Waste in the Lab Using Real-Time Data Evaluation with Automated Reactions
By Peter Zipfell, Product Marketing Manager for Chromatography and Mass Spectrometry software at Thermo Fisher Scientific 10.10.2025
In the ever-evolving landscape of laboratory analysis, avoiding data loss and ensuring accurate results are of vital importance, but laboratories face many challenges to ensure precious sample is not wasted and analytical sequences run correctly the first time, keeping throughput as high as possible. These challenges can be associated with examples such as an analytical run coming to an abrupt halt, resulting in sample re-extraction, the need for repeat analysis due to a poor injection or a sample falling outside acceptable tolerances. In particular, unknown samples with analyte concentrations exceeding the upper limit of quantification (ULOQ), can compromise quantification accuracy and increase the risk of carryover to subsequent injections. To address these issues, software can provide automated system suitability testing and in-run decision making for more right first-time analyses.
A set of test tubes filled with blue liquid in a rack on a laboratory bench, with a scientist in a lab coat blurred in the background.
Enhancing data integrity with SST/IRC
The System Suitability Testing (SST) and Intelligent Run Control (IRC) functionality in Thermo Scientific Chromeleon software is designed to enable real-time monitoring and evaluation of key operational and data processing parameters. This capability allows laboratories to define specific thresholds and automatically respond to deviations, preventing potential issues such as carryover, sample waste and quantification inaccuracies. By automating the evaluation of critical operational conditions and data processing results, Chromeleon software ensures that both chromatography and MS systems operate consistently and within expected parameters, ultimately leading to more accurate and reliable analytical results.
Support for MS instruments
The SST/IRC functionality can be applied to Thermo Scientific mass spectrometers, including single quadrupole, triple quadrupole and HRAM MS. This versatility makes it an invaluable tool for clinical research and forensic toxicology laboratories, where maintaining the integrity and reliability of LC-MS datasets is crucial. The ability to define and execute automated responses to test case outcomes ensures that the system operates within expected parameters, saving precious sample and time for data review.
Real-time decision making
One of the standout features of the SST/IRC functionality is its ability to make real-time decisions based on predefined thresholds. For instance, if an unknown sample has very high analyte concentrations, the system can automatically trigger the “AutoDilution” function, adjusting the injection volume to ensure accurate quantification. This intelligent response helps mitigate the risk of carryover and maintains the integrity of subsequent injections.
Advancing laboratory performance
The SST/IRC capabilities in Chromeleon software provide a robust framework for maintaining the integrity and reliability of LC-MS datasets. By automating the evaluation of critical operational conditions and data processing results, you can enable real-time decision-making to address potential issues promptly. This not only safeguards the quality of the data but also enhances laboratory efficiency, reducing sample waste and minimizing data loss.
The integration of SST/IRC functionality represents a significant advancement in laboratory data management. As laboratories continue to face increasing demands for accuracy and efficiency, tools like SST/IRC will play a crucial role in ensuring reliable and high-quality analytical results. For more information on how Chromeleon software can enhance your laboratory operations, you can register for our latest webinar, Safeguard dataset quality in LC-MS analysis with system suitability testing and intelligent run control (SST/IRC) or visit Chromeleon Software-Targeted MS Quantitation.
Visit us on LinkedIn: #Chromeleon #ChromatographyDataSystem #MassSpectrometry #TargetedQuantitation
Thermo Fisher Scientific's Edward Goucher and Alexandra Graham pictured beside KCL's Stefania Boccuzzi and Vincenzo Abbate
Advancing Drug Analysis with VeriSpray PaperSpray Mass Spectrometry
At the January 2025 CFSRE Seized Drugs Symposium, a Thermo F...
Read More
Sheffield teaching hospital NHS foundation
Beyond Blood: Sheffield Teaching Hospitals NHS Foundation Trust Develops HRAM-Powered Method for Post-Mortem Toxicology Analysis
In a recent study published in the Journal of Analytical Tox...
Read More
Decoupling Acquisition From Data Review: Unlocking Flexibility and Efficiency in Chromatography and Mass Spectrometry Workflows
In a recent blog post we discussed how centralized data mana...
Read More
Automated LC-MS Sample Cleanup Using Turbulent Flow Chromatography
Too much interfering matrix? Discover how to streamline your...
Read More
Peter Zipfell
For over 25 years, he has worked extensively with analytical hardware and software transitioning from LC-MS method development through HPLC technical support to chromatography data systems. His focus has always been to support our customers by delivering software solutions across multiple industries to enable them to improve their current workflows and increase productivity.
Generating Quantitative Methods Automatically From Spectral Library Data
Ion Chromatography Turns 50: Chromeleon Software’s Tribute to a Lifelong Partner