Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Biopharmaceutical / Sample Handling Made Easy!
Sample Handling Made Easy!
By Petra Gerhards, Regional Marketing Manager GC and GC/MS, EMEA, Thermo Fisher Scientific 01.08.2020
Do you feel like there’s never enough time in the workday? If that’s the case, there are ways to reduce your stress with Thermo Fisher Scientific’s easy-to-use, U.S. Environmental Protection Agency (EPA), convenient kits and best-in-class They are suitable for any samples that require the removal of particulates and microorganisms.
Are you performing environmental analysis? Check out our
Common Problems in Sample Handling and How to Solve Them
Samples need to be taken, transported and stored before they can be analyzed in a lab. The most important part of analyzing down to the last fg is to use the right glass quality. The first hydrolytic class, as described in the Pharmacopeia, chapter 3.2., is used for glass vials of all kinds used in labs. However, there are differences. The difference is the amount of free silanol groups on the surface of the glass. These EPA storage vials provide a highly inert surface with only 33% free silanol groups (51% for brown glass).
It is also important to use glass that is not too thin. Storage vials are used for stirring, shaking, freezing and heating. If a vial brakes, it can contaminate the fridge, lab, or even employees.
A lot of samples are filtered before going into further steps of sample preparation. This can be a filtration for particles or microorganisms. However, it is important to use plastic materials that are tested for extractables and leachables. If this is not the case, a high background can result in GCMS or ion suppression in LCMS. This means a loss of sensitivity and round-robin tests are not passed. At the end of the day, labs can lose reputation with their customers as a result of low-quality glass in the lab.
Another big issue with filters is poor design of the filter itself. If there is an uneven distribution over the membrane, a lot of sample still sits on the filter and only a low amount goes through. In a worst-case scenario, the filters can burst.
The use of wrong membranes can also lead to bad results.
Using Thermo Scientific Titan3 filters can prevent these problems. They have an engineered design for the distribution of samples over the membrane and can stand up to 15 bar pressure. A special burst ring provides bursting. In addition to those benefits, the filters are color-coded to prevent wrong usage.
Get your free samples today at www.thermofisher.com/filter
More information is available at www.thermofisher.com/syringefilters
Sample Preparation Strategies Demystified: Choosing the Right Fit for Your Toxicology Workflow
Smarter sample preparation for LC-MS: from simplicity to sel...
Read More
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
In industrial processes, accurately determining halogens and...
Read More
Unveiling the Advanced Workflow for PAH Analysis in Smoke Flavored Foods
Unveiling the Advanced Workflow for PAH Analysis in Smoke Flavored Foods
Introduction Polycyclic aromatic hydrocarbons (PAHs) are a p...
Read More
Benefits of Automating Your Antibody QC Analytical Workflows
Want a less stressed QC lab?  Discover how automation c...
Read More
Petra Gerhards
Petra Gerhards, Dipl-Ing, is Regional Marketing Manager of GC and GC-MS for EMEA at Thermo Fisher Scientific. She has more than 29 years of experience in the fields of GC-MS, SPE and LC-MS. Since joining the regional team she has contributed to workflow solutions combining vials and closures with SPE solutions, GC-MS and LC-MS. She works with KOL's on data for regional specific marketing campaigns, organizes in-house seminars and works on customer specific solutions. Her main expertise is in the field of doping and drugs-of-abuse analysis.
Automate Sample Preparation for PFAS Analysis in Drinking Water
Orbitrap Mass Spectrometry and Artificial Intelligence