Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Ion Chromatography / Solving Hydrogen Peroxide Analysis Problems
Solving Hydrogen Peroxide Analysis Problems
By Wai-Chi Man, Product Marketing Manager, IC/SP, Thermo Fisher Scientific 07.18.2020
One of my earliest memories of hydrogen peroxide was at school in chemistry class where we put it on the scab of a classmate and watched it fizz and disappear. It was fascinating! Back then, health and safety were a little different than they are today! The older I got the more I realized how often hydrogen peroxide is used, mainly as an oxidizer, bleaching agent, and antiseptic, and it is found in many products used from households to hospitals.
The use of hydrogen peroxide is extensive. In our homes, we can find it in mouthwashes where it acts as an antiseptic and teeth whitening agent at a low concentration. It’s widely used in the hairdressing industry to bleach hair. Hair bleaching kits have become more available for home use, always supplied with a pair of disposable gloves (ever think about what comes through those gloves?).
Other uses for hydrogen peroxide include bleaching paper and flour. It’s also found in glow sticks to form chemiluminescence, rocket propellant, laundry detergents, disinfectant, and antiseptics used in hospitals to clean surgical equipment. Many surfaces are cleaned with products that contain hydrogen peroxide due to its efficacy against viruses, yeasts, bacteria, and spores.
Now that I have established that hydrogen peroxide is used everywhere, you may wonder I’m spending time demonstrating this point. Well, hydrogen peroxide decomposes to water, which is much more environmentally friendly and less pungent than some other oxidizers and bleaching agents:
2 H2O2 → 2 H2O + O2
However, hydrogen peroxide is thermodynamically unstable and very reactive!
At high concentrations it can corrode many surfaces, and when in contact with organic compounds it can form explosive mixtures. Hydrogen peroxide vapours inhaled can irritate and damage your nasal path and throat. If our skin comes into close contact with the high concentration it will oxidize our skin! Ever accidentally get a little on your fingers (at low concentration) and your skin turns all white then rinse off with water? It slowly goes back to normal but if it is very concentrated it will corrode your skin. For such a small molecule, there are a lot of advantages but it can also cause harm if used inappropriately at an increased concentration. So, how can we monitor the concentration? How do you solve an analysis problem like hydrogen peroxide?
Are you currently finding yourself in a need to disinfect more surfaces like most of us are during this global pandemic? Want to know how to detect the residue amount of hydrogen peroxide from surfaces? With electrochemistry using high-pressure anion exchange (HPAE) with pulsed amperometry detection (PAD), partnered with the right working electrode (conventional or disposable), you can measure the current generated by the hydrogen peroxide. This then allows us to quantify it in order to better understand if it’s being used in an appropriate, safe concentration.
To find out more about hydrogen peroxide analysis > http://tools.thermofisher.com/content/sfs/posters/PN-70967-Hydrogen-Peroxide-Detection-IC-Electroche…
1975 text image
Innovations of 1975 and the Birth of the First Commercialized Suppressed Ion Chromatograph
The year 1975 was a remarkable period in the history of tech...
Read More
US EPA Method 1621_1200x600
Understanding PFAS and Combustion-Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) comprise more tha...
Read More
Pesticides
Pesticide Residue Analysis in the 21st Century: Innovations in Sample Preparation and Detection Using the Thermo Scientific TSQ 9610 GC-MS/MS
Introduction The accurate detection of pesticide residues in...
Read More
IC Golden Jubilee blog header image
What is the Ion Chromatography Golden Jubilee? Celebrating 50 Years of IC Innovation
The year 2025 marks the golden jubilee of ion chromatography...
Read More
Wai-Chi Man
Wai-Chi Man is Product Marketing Manager, IC/SP, for Thermo Fisher Scientific.
If You Had the Chance, What Would You Ask About Isotope Fingerprints?
Breath Biopsy™ – A Non-Invasive Tool for the Early Detection of Disease and Cancer