Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Biopharmaceutical / Speeding Up Development of Biosimilars With Improved Analytical Tools
Speeding Up Development of Biosimilars With Improved Analytical Tools
By AnalyteGuru Staff, 06.26.2020
Biosimilar Market Space and Challenges
Biotherapeutics have grown to be an indispensable tool in medicine today. Advances in the development of biologics have offered lifesaving treatments to patients with deadly diseases and quality of life improvements by managing chronic diseases. It sounds too good to be true, so what is the downside? Biologics are often associated with high costs and limited patient access. This is where the market opportunities open for biosimilars. To date, there are 26 biosimilars approved by the Food and Drug Administration and 53 approved by the European Medicines Agency, with thousands in development. Biosimilars can offer competition in the market to create pricing pressure and expand patient access to innovative medicines.
A biosimilar is a biologic that is highly similar to the approved reference biologic on the market. The FDA defines a biosimilar as “a biological product that is highly similar to the reference product, notwithstanding minor differences in clinically inactive components.” However, it’s not as simple as it sounds. The challenge lies in the fact that a biologic is produced in living cells and, consequently, it is a highly complex heterogeneous mixture of products with slight variations. For this reason, it is very important to identify a robust, controlled process and then follow that manufacturing process every time to control natural process variability in biologics.
To demonstrate biosimilarity, a manufacturer must provide sufficient data and information to regulatory agencies to prove there are no clinically meaningful differences from the reference product. The necessary overlap between a biosimilar and reference product is the amino acid sequence and the drug’s mechanism of action. The biosimilar host cell line, processes, and formulation typically diverge from the reference product. Analytical tools must be used to design quality controls to ensure biosimilarity.
For biosimilar approval, regulatory bodies require detailed therapeutic protein characterization including:
    • Batch-to-batch comparisons
    • Stability studies
    • Impurity profiling including aggregation, post-translational modifications, glycoprofiling
    • Determination of related proteins and excipients
The Key to Success- Biosimilar Analytical Strategy
Analytical strategy is key for creating and generating data showing comparability of a biosimilar. This analytical strategy can provide important clinical similarity data streamlining and speeding up the development process. Fully understanding the effects of the manufacturing process on the product at early stages of development allows for generation of suitable methods for product monitoring in later stages. Currently developing a biosimilar is a multi-year process generating considerable development costs, with each biosimilar costing $150-$200 million to develop. Early process understanding can help better define risks and allow for better risk management strategies. For the success of biosimilars, it is important for this process to quickly identify successes and where changes are needed in the process.
Identifying analytical methodologies providing characterization and comparability data in a shorter time can reduce the overall product development and development costs. High-resolution accurate mass (HRAM) mass spectrometry techniques can bring fast answers during biosimilar development and allow for stringent process control throughout development and into manufacturing. Learn more in this recent application note using mass spectrometry to quickly compare biosimilars to reference products on an intact protein level.
Comparing biosimilars using intact mass analysis under denaturing and native conditions
To learn more, visit: Biosimilar Analysis Workflows
On LinkedIn? Visit our LinkedIn page #biopharmaceutical
Innovation in Biopharma analysis image
Scaling Up Biopharmaceutical Characterization with the Orbitrap Ascend BioPharma Tribrid Mass Spectrometer
Biopharmaceutical pipelines are expanding rapidly, with incr...
Read More
GLP-1 image
Five Key Advantages of the Thermo Scientific LC-MSn Solution for Ultra-Sensitive GLP-1 Quantitation
Bioanalytical analysis using LC-MSn requires precise, accura...
Read More
ACT platinum cover image
A Greener HPLC Guide: How ACT Labels Help Promote Lab Sustainability
Labs use up to 10 times more energy than offices. Here’s h...
Read More
How Can Data Sharing Accelerate my Drug Research
How Can Data Sharing Accelerate my Drug Research?
In today’s fast-paced world of drug research, big data...
Read More
AnalyteGuru Staff
A Fully Automated Method for MCPD- and GE-esters and the Importance of Glass Quality of the Used Autosampler Vials
Re-defining Productivity for Small Molecule, High-resolution Mass Spectrometry Analysis