Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Environmental / Testing Orthophosphate in Drinking Water? Discrete Analyzers Bring Welcome Automation
Testing Orthophosphate in Drinking Water? Discrete Analyzers Bring Welcome Automation
By Gary He, Product Marketing Manager, Gallery Discrete Analyzers, IC Industrial markets, Sample Prep, Thermo Fisher Scientific 03.03.2023
In a world continuously faced with environmental challenges, analyzing drinking water for contaminants is essential to protect public health.
If your lab analyzes drinking water — and if you’re seeking better ways to automate and simplify the process while meeting regulatory compliance requirements — here are eight things you need to know:
1. Compliance with standards and regulations is non-negotiable
In the United States, the quality of drinking water is regulated and supervised by the U.S. Environmental Protection Agency (EPA). The water quality parameters of health concerns and their analysis are regulated under the 40 CFR Part 141 National Primary Drinking Water Regulations.
2. Drinking water contaminants can be wide-ranging
Analysis of drinking water is an important method for identifying contaminants such as inorganic anions, cations, heavy metals, organic pollutants, and nutrients.
3. Orthophosphate is a common additive used by water utilities
Water utility companies treat drinking water by adding orthophosphate to prevent the corrosion that can cause metals, particularly copper and lead, to dissolve and leak into water systems. Lead contamination occurs when water comes in contact with lead pipes, and lead contamination in drinking water is a huge problem for municipalities with an older infrastructure.
4. Discrete analyzers are a smart way to measure orthophosphate
In 2016, the EPA approved the first discrete analyzer method for testing orthophosphate in drinking water, which uses the Thermo Scientific Gallery discrete analyzer platform. The Thermo Scientific drinking water method: Drinking Water Orthophosphate for the Thermo Scientific Gallery Discrete Analyzer is the resource that describes the EPA-approved alternative method for testing drinking water for orthophosphate.
5. EPA’s original method (2016) is based on ascorbic acid reduction
The Thermo Scientific drinking water method is based on the well-established and EPA-approved reference method SM 4500-P E: Phosphorous by Ascorbic Acid Method. Standard Methods for the Examination of Water and Wastewater, which detects orthophosphate using the ascorbic acid reduction method.
In this method, orthophosphate reacts with ammonium molybdate and antimony potassium tartrate in an acidic medium to form an antimony-phospho-molybdate complex. The complex is subsequently reduced by ascorbic acid to form an intensely blue complex that is measured photometrically at 880 nm. The method is easily automated using the Gallery discrete analyzers, which allow simultaneous analysis of multiple parameters in the same sample aliquot.
6. Ready-to-use Gallery system reagents, test procedures help to further streamline analyses
The Thermo Scientific Gallery and Gallery Plus Aqua Master discrete analyzers provide equivalent performance and are therefore also applicable to the Thermo Scientific method. The Gallery Aqua Master discrete analyzers add additional capabilities, including automatic spiking, flexible test, and quality control (QC) parameter configuration, and flexible results reporting with versatile features for configuring report templates.
7. New application note shows correlation between two methods
In Thermo Fisher’s application note (001747), you can read the performance study results that showed the automated method meets the QC acceptance criteria in the reference method. The Thermo Scientific orthophosphate method showed a good correlation to the well-established EPA-approved reference method 4500-P E: Phosphorous by Ascorbic Acid Method. The study also demonstrated that when performed using the Gallery discrete analyzers, the method is suitable for analyzing different types of water samples. Read the app note, Automated method to detect orthophosphate in drinking water using the Thermo Scientific Gallery discrete analyzer.
8. Discrete analyzers and methods can dramatically increase efficiency
The Thermo Scientific discrete analyzer method offers laboratories an automated, high-throughput option for regulated testing for orthophosphate in drinking water. Walk-away automation simplifies drinking water analyses and reduces errors. Ready-to-use Gallery system reagents are convenient, and likewise decrease the costs and errors associated with manual processes. The Gallery and Gallery Plus Aqua Master discrete analyzers provide equivalent performance and can also be applied to the Thermo Scientific method.
Related information
To learn more about the Gallery Aqua Master, check out these online resources:
  • Read the application note
  • Read the solution brochure
  • Explore the informational web page
  • Blog post: Analyzing Total Oxidized Nitrogen (TON) in Drinking Water? Simplify Your Method
  • Blog post: Turnkey Wet Chemistry Analysis: Top 7 Questions Answered
Sealed vial with vapor above liquid, laboratory glassware blurred in background.
VOC Analysis in Drinking Water: Achieving ppt-Level Sensitivity with ITEX Dynamic Headspace GC-MS
The quality of drinking water is under closer scrutiny than ...
Read More
Food packaging
Enhancing PFAS Analysis in Food Contact Materials Using Combustion Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) have been used in...
Read More
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
In industrial processes, accurately determining halogens and...
Read More
IC Golden Jubilee blog header image
What is the Ion Chromatography Golden Jubilee? Celebrating 50 Years of IC Innovation
The year 2025 marks the golden jubilee of ion chromatography...
Read More
Gary He
Gary He is a Product Marketing Manager at Thermo Fisher Scientific, overseeing the Gallery discrete analyzers, Dionex ion chromatography (IC) in industrial markets, and sample prep. In this role, he leads marketing programs aimed at fostering the sustainable adoption of technology in well-established applications like water and inorganic contaminants analyses. Additionally, Gary focuses on advancing growth applications by implementing IC workflows in areas such as clean energy and lithium-ion battery testing.
How Much Microplastic Do You Eat in a Day?
How the Solid-State Battery Could Power the Future of EV Industry