Get credit towards a new discrete analyzer when you trade in your current system
Trade in your current discrete analyzer or flow injection analyzer system and get a credit toward the purchase of a Thermo Scientific Gallery or Gallery Plus Discrete Analyzer, whether you originally purchased from us or another manufacturer.
Which Discrete Analyzer Is Most Suitable for My Lab? A Discrete Analyzer Selection Guide
Use this guide to view the ten factors to consider before making your automated discrete analyzer purchase decision.
Wastewater surveillance of COVID-19
Learn about strategies to consolidate chemical reference parameters and population biomarker analysis to support the wastewater-based epidemiology.
How to automate traditional wet chemistry workflow
As the world is continuously faced with environmental challenges, compliance standards and regulations must be met along with them. Soil analysis, drinking water analysis, sewage water and wastewater analysis are all required to protect the environment and our public health. Environmental contaminants to be analyzed may include inorganic anions, cations, heavy metals, organic pollutants and nutrients. The United States Environmental Protection Agencies (USEPA) Safe Drinking Water Act (SDWA) and the Clean Water Act (CWA) are the two important environmental laws governing water regulations. Clean Water Act Methods Update Rule for the Analysis of Effluent 40 CFR part 136 and many of the regulatory bodies, such as the ASTM, ISO, DIN recommends wet chemical methods wastewater analysis.
Wastewater surveillance of COVID-19 is a rapidly evolving area of research that holds great promise as an early, cost-effective, unbiased community-level indicator of the presence of COVID-19. Persistence of the genetic signal will be impacted by: pH, ammonia, phosphate and other nutrients. Other water quality parameters can provide information about the operational status of the water resource recovery facility. This can be useful in identifying the occurrence of events that may impact the strength of the genetic signal.
Manufacturing industries consuming large volumes of water produce both hazardous and non-hazardous wastewater. Treating this wastewater and reusing it is a key challenge for companies seeking to maintain a profitable operation while protecting the environment. Reclaimed or recycled water is produced by conversion of wastewater into water that can be reused for other purposes. These waters are adequately treated to meet “fit-for-purpose specifications” for next use. Wastewater testing serves two purposes:
- To indicate that treatment in place is working for re-use purposes.
- Industrial wastewater is safe before it’s discharged into surface waters in accordance with regulations.
EPA sets legal limits on over 90 contaminants in drinking water. National primary drinking water regulations (NPDWR) includes inorganic and organic chemicals, disinfection byproducts and disinfectants determination by wet chemical and ion chromatography methods.
Automated nutrient analysis and water quality monitoring
Traditional water testing involves – multiple wet chemical instruments for multiple parameters such as Nutrient analysis, pH, Conductivity, alkalinity and hardness etc. Each test requires a separate sample and often multiple sequential steps which extends the testing process. With slow processes and the need for specialist staff to run and monitor equipment, water quality testing is labor-intensive, time-consuming and inefficient. Routine water testing can be costly and time consuming for a laboratory that performs routine nutrients, ground water, drinking water and wastewater analysis while experiencing increased demand for the routine analytical services.
Traditional wet chemistry methods like titration or ISE sequentially measure individual test parameters. For each parameter, dedicated ISE are necessary and have limited shelf lives. Each titration parameter needs a dedicated burette and sensor, as the number of parameters per sample increases, the overall system complexity increases, resulting in long hands-on sample and response times.
Consolidated high throughput water analysis workflow
Consolidated multiparameter discrete analysis now offers a solution to these difficulties and offers high throughput wet chemical analysis workflow. Discrete analyzers, which consolidate and simultaneously test for up to 20 parameters — using a single instrument with a single operator. Gallery and Gallery Plus discrete analyzers are easy-to-use, automated systems that allow laboratories to simplify their testing with dual benefits: time and cost savings. All necessary analysis steps are automated, providing true walkaway time for the operator. Both Gallery and Gallery Plus instruments provide an integrated platform for two measurement techniques, photometric and electrochemical (pH and conductivity), which can be run simultaneously. Parallel determination of several analytes from a single sample as well as the presence of several automated features ensures analytical efficiency.
pH and conductivity for 100 samples can be tested simultaneously in less than 90 minutes, while the system is measuring alkalinity, hardness and other parameters. While traditional analysis requires multiple wet chemistry methods and, therefore, multiple samples, the Gallery discrete analyzer consumes a maximum of 300 μL of sample per test, can test for up to 20 parameters per sample, and runs up to 200 tests per hour.
Our discrete analyzers automate photometric and electrochemical analysis provide fast, reproducible results that allow laboratories to measure multiple analytes simultaneously, while reducing total analysis and operator time.
- Integrated platform for photometric and electrochemical analysis can be operated simultaneously
- Ability to achieve detection of very low levels (ppb).
- Contamination-free analysis using disposable cuvettes.
- Continuous access to samples, reagents, and cuvettes without interruption of routine workflows.
- Large concentration range measurement, with excellent reproducibility.
Recommended products
The Thermo Scientific Disc-IC system is a package of two independent techniques from Thermo Fisher Scientific—discrete analysis and ion chromatography for comprehensive water analysis. Unique and fully automated, the Disc-IC system combines the power of direct measurement, discrete analysis with ion chromatography to support a comprehensive water analysis workflow that offers the highest flexibility and sample throughput to reduce overall cost per analysis.
The Thermo Scientific Gallery discrete analyzer provides an integrated platform for two measurement techniques, photometric and electrochemical (ECM), which can be run in parallel. Discrete cell technology allows for simultaneous measurement of several different tests for the same sample, eliminating method changeover time. The Gallery system can achieve very low detection levels, and its sophisticated dilution features help to manage a wide concentration range without user intervention.
Gallery Discrete Analyzer
The Thermo Scientific Gallery Discrete Analyzer includes a combined sample and reagent disk for a maximum capacity of 90 samples and 30 reagents, with the capacity to perform up to 200 tests per hour.
Gallery Plus Discrete Analyzer
Accommodate 108 samples and 42 reagents in its separated sample and reagent disks, with the capacity to perform up to 350 tests per hour with the Thermo Scientific Gallery Plus Discrete Analyzer.
ECM is an integrated unit into the Gallery discrete analyzers and supports the automated pH and conductivity measurements. It is capable to simultaneously measure both pH and conductivity alongside the photometric testing. ECM is suitable for raw water, ground water, sea water, rainwater, municipal water, drinking water and wastewater.
Ready to use reagents for environmental analysis
Save time by eliminating reagent preparation with the Thermo Scientific Gallery system reagents for environmental testing, such as water (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use gallery system reagents for environmental analysis, the kits include 1 to 3 reagents in 20 mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries. Save time and reduce errors by simplifying your analysis with the Gallery system reagents for environmental analysis.
Thermo Scientific reagents are ready to go, saving the technician’s time and reducing errors. The unique low volume cuvette design guarantees small reagent volumes, minimizes reagent waste, and as a result, reduces reagent costs. Optimized kit sizes and on-board stability further minimize the amount of waste produced and increase cost efficiency.
Ensuring confidence in the quality of results, the methods used for analysis in the Thermo Scientific discrete analyzers are well known enzymatic and colorimetric chemistries optimized to the result levels established in international reference methods, US EPA, ISO, DIN, ASTM and other standard methods.
Thermo Scientific water analysis system reagents kits offers
- Accurate and reproducible results
- Requires less reagent usage
- Reduces preparation errors
- Reduces hands-on time
- Ease of use
Regulatory Methods: U.S. EPA-Approved Discrete Analyzer Methods
Thermo Scientific Gallery discrete analyzer with ready to use system reagents are optimized for speed, flexibility, and precision for drinking water and wastewater. The unique discrete cell technology allows laboratories to measure multiple analytes simultaneously while reducing total analysis and operator time. The unique low volume cuvette design accommodates small reagent volumes, minimizes reagent waste, and as a result, reduces reagent costs.
Thermo Scientific system reagents for automated discrete analyzers covers over 50 analytes. Ready to use reagents are easy to work with eliminating extra preparation steps and saving time and money.
The table below summarizes U.S. EPA, ASTM, DIN and other international regulatory approved discrete analyzer and Ion chromatography methods.
Gallery discrete analyzer
U.S. EPA and international reference methods
Regulatory methods | Analytes |
---|---|
EPA 310.2 ISBN 0117516015 SMWW 2320 B | Alkalinity* |
SM 3500-AI B | Aluminum* |
EPA 350.1 ISO 7150 ISO 15923-1 DIN 38406 ISBN 0117516139 | Ammonia |
ISBN 0117515833 | Boron* |
EPA 410.4 | COD* |
EPA 325.2 EN ISO 15682 ISO 15923-1 ISBN 0117516260 SM 4500-Cl-E | Chloride |
EPA 330.5 SM4500-Cl G | Chlorine* |
EPA 120.1 | Conductivity |
SM 3500-Cu C | Copper |
ASTM D2036-09(B) | Cyanide |
EPA 335.4 | Cyanide (Total) |
EPA 340.3 SM 3500-F D | Fluoride |
EPA 130.1 | Hardness (Total) |
Regulatory methods | Analytes |
---|---|
ISO 11083 DIN EN ISO 23913:2009 ASTM D1687-12(A) SM 3500 Cr-B SW 7196 A | Hexavalent chromium |
ISO 6332-1988 SM 3500-Fe B | Iron |
SM 3500-Fe B | Iron (Ferrous), Iron (Total) |
ASTM D7781-14 ASTM D7781-14 (NECi) Nitrate Reductase method for drinking water (USGS I-2547-11 (USGS I-2548-11 (NECi) Method N07-0003 | Nitrate (TON ENz) |
EPA 353.1 ISO 15923-1 ISBN 0117515930 SM 4500-NO3 H | Nitrate (TON Hyd) |
NEMI (Nitrate via manual Vanadium (III) reduction) | Nitrate (TON Vanadium) |
EPA 354.1 ISO 13395:1996 ISO 15923-1 DIN EN 26777 ISBN 0117515930 SM 4500 NO2-B | Nitrite |
EPA 150.2 | pH |
Regulatory methods | Analytes |
---|---|
EPA 420.1 ISBN 0117516171 | Phenols (Total) |
EPA 365.1 EN ISO 6878 ISO 15923-1 ISBN 0117515825 SM 4500 P-E | Phosphate |
EPA 365.4 ISBN 0117518883 | Phosphorus (Total) |
EPA 370.1 ISO 15923-1 SM 4500 SiO2-C USGS I-2700-85 | Silica |
EPA 375.4 ISO 15923-1 DIN 38405-D 5-2 ASTM D516-11 ISBN 0117533406 SM 4500 SO4-E | Sulfate |
EPA 376.2 ISBN 011751718 SM 4500-S2 D HMSO SCA Blue Book | Sulfide* |
SMWW 4500-CN-M | Thiocyanate* |
EPA 351.2 ASTM D3590-11 (B) ISBN 0117521299 SMWW 4500-N(Org) | Total Kjeldahl Nitrogen (TKN) |
Ion chromatograph
U.S. EPA and international reference methods
Regulatory methods | Analytes |
---|---|
EPA 302 EPA 317 EPA 321.8 EPA 326 ISO 11206 | Bromate |
ASTM D6581 | Chlorite, bromate, bromide, chlorate |
ASTM D7237 | Cyanide |
EPA 300.0 ASTM D4327 | Fluoride, chloride, nitrite, bromide, nitrate, ortho-phosphate, sulfate |
EPA 300.1 | Fluoride, chloride, nitrite, bromide, nitrate, ortho-phosphate, sulfate, bromate, chlorite, chlorate |
EPA 557 | HAAs, BrO3, dalapon |
EPA 218.7 ASTM D5257 | Hexavalent chromium |
ISO 14911 ASTM D6919 | Lithium, sodium, potassium, ammonium, magnesium, calcium |
ASTM D6994 | Metal cyanide |
EPA 314.0 EPA 314.2 EPA 332 ISO 19340 | Perchlorate |
ASTM D8001 | TKN, total phosphorus |
Resources
Fast, safe, and easy nutrient analysis
Discover Thermo Scientific Gallery and Gallery Plus discrete analyzers for complete nutrient analysis of drinking water, wastewater, and soil samples.
Wastewater surveillance for SARS‑CoV‑2 biomarkers
Learn how the National Laboratory Service achieves high-throughput biomarker testing for SARS-CoV-2 in wastewater.
Fast and accurate Gallery discrete industrial analyzers
Thermo Scientific Gallery analyzers are sophisticated discrete photometric systems for the precise and easily adaptable analysis of water and environmental samples.
Solution for routine and comprehensive water analysis
Discrete analyzers using proved to be the solution needed for easy soil testing for a South African Fertilizer Manufacturer. Automated operation, easy operation, and time savings proved most beneficial.
Increasing analytical efficiencies in the City of Atlanta
The City of Atlanta used the Thermo Scientific Gallery Plus discrete analyzer with electrochemical (ECM) unit for methods that enable them to reliably test and supply safe drinking water to the citizens of Atlanta.
Environmental & industrial applications compendium
Discover how Gallery Discrete Analyzers offer simultaneous, multi-parameter nutrient analysis for drinking water, wastewater, sewage water, soil and fertilizer and plant extract samples.
Environmental analysis webinars
Browse our searchable database of on-demand webinars about discrete industrial analyzers for environmental analysis to find methods and applications to accelerate the work in your lab.
No records were found matching your criteria