It’s Christmas! Again! A time of year to celebrate and reflect on the year gone by, and to catch up with friends or family (or both). What better way to do it, than over your favourite drink (alcoholic or not). Coming from Scotland, my go-to festive tipple has always been Whisky (yes, that’s the way we spell it). A drink with such popularity, complexity and let’s be honest – some high price tags! Walk through any airport and marvel at the choice on offer and of course some eye watering prices. It’s a revered spirit with a history as rich as its flavor, has always been synonymous with tradition, craftsmanship, and quality. However, the increasing demand for premium whisky has led to a surge in adulteration practices, posing significant challenges for consumers and producers alike. In this blog, we delve into the intricacies of whisky adulteration and explore how cutting-edge technology, specifically the Thermo Scientific Orbitrap Exploris GC 240 mass spectrometer, is revolutionizing the fight against counterfeit whisky and ensuring your Christmas cheer is genuine.
Understanding whisky adulteration
Whisky adulteration can take several forms, each compromising the integrity of the spirit.
- Artificial Whisky Creation: This involves mixing various chemicals to mimic the flavor and aroma of genuine whisky. The absence of safety controls over the quality and concentration of these chemicals raises health concerns.
- Mislabelling and False Aging Claims: Bottles may be falsely labelled with more expensive brands or incorrect maturation periods. This type of adulteration often involves adding artificial colours or using heat to accelerate the aging process, giving the appearance of a well-aged whisky without the time investment.
For the consumer its hard, or even impossible, to distinguish between a budget blended whisky and a genuinely aged single malt whisky. Furthermore, how could you tell if a whisky had been aged for 10 years or 25 years (apart from the higher price). You can see the economic motivation for the whisky fraudster.
The role of advanced technology
To combat these sophisticated adulteration techniques, a robust and precise analytical approach is essential. The Orbitrap Exploris GC 240, combined with headspace solid-phase microextraction (SPME Arrow) via the Thermo Scientific TriPlus RSH SMART Autosampler, offers a powerful solution for the comprehensive characterization of whisky.
Workflow for finding the differences
The workflow shown in figure 1 for differential analysis of whisky using the Orbitrap Exploris GC 240 mass spectrometer is straightforward yet highly effective:
- Sample Preparation: Whisky samples (100 µl) are aliquoted directly into vials and mixed with water (900 µl). After heating, SPME Arrow was used to sample from the vial headspace, which allows for efficient extraction of volatile compounds.
- Chemical Profiling: The samples are analyzed in EI full scan at 120,000 mass resolving power (FWHM m/z 200) using the Orbitrap Exploris GC 240. This provides comprehensive high-resolution and accurate mass data that enables the detection of subtle differences in the chemical profiles of different whisky samples.
- Data Analysis: The data is processed using Thermo Scientific Compound Discoverer software, which incorporates a high-resolution accurate mass flavor and fragrance library. This facilitates the identification of compounds and comparison between samples.
Figure 1. Workflow from left to right showing how from data acquisition in full scan we move from peak detection, to identifying unique components in a sample, to finally making a compound identification.
Case study: identifying adulteration
In the study reported in application note 003094: “Non-targeted analysis of whisky using SPME Arrow and Orbitrap Exploris GC 240 mass spectrometer”, the differential analysis of whisky samples revealed significant insights (figure 2). The principal component analysis plot for whiskies of different age statement shows clear chemical differences between the samples. We can dive deeper into the data to see what compounds drive that difference. For instance, comparing sample 2 and sample 3 the results highlighted compounds unique to sample 2, such as higher levels of 2,3 – methyl-1-butanol and Furfural. These compounds are typically associated with Scotch malt whisky and indicate a longer and different maturation process, suggesting that sample 2 is a genuine aged whisky compared to sample 3.
Figure 2. Principal component analysis loading plot of whisky samples based on identified chemical profile from Compound Discoverer software.
Ensuring quality and authenticity
As the whisky market continues to grow, the importance of ensuring quality and authenticity cannot be overstated. Advanced analytical technologies like the Orbitrap GC-MS provide a powerful tool in the fight against whisky adulteration. By leveraging these technologies, producers can uphold the tradition and integrity of whisky, ensuring that every bottle delivers the genuine experience that consumers expect and deserve.
Whether you are a whisky enthusiast or a producer, understanding and utilizing these advanced technologies is key to preserving the heritage and quality of this beloved spirit.
So cheers, or as we say in Scotland “Slàinte Mhath” to authenticity and innovation in the world of whisky!