Are your columns not lasting as long as you want? Are you struggling to identify what kills your column or how you can increase its lifetime? A new white paper describes what actions you can take to prolong the lifetime of your HPLC column.
There are many preventative measures you can take to increase column lifetime, in addition to systems you can implement to improve your data when you do need to troubleshoot a problem with your column.
The majority of challenges with column lifetime come from your samples. Dirty samples — either full of particulates or chemical contaminants — will deteriorate your column faster if not effectively removed. Your lab can extend column lifetime through proper sample preparation, column protection or by implementing effective cleaning procedures.
For more information on how to implement these preventative measures, see the white paper: How to prolong the lifetime of your HPLC column.
It is also important to implement a system to investigate what causes the deterioration of a column so that when a problem arises, it takes less time to get to the route cause. For tips on what systems to implement, see the aforementioned white paper.
AAV-SEC Analysis: Top 3 Takeaways From the Latest Article
What is the best column for separating AAV size variants? Th...
Read More
New Column for Fast Analysis of Haloacetic Acids, Bromate and Dalapon Using Ion Chromatography Coupled with Mass Spectrometry
Haloacetic acids (HAAs) are disinfection byproducts (DBPs) f...
Read More
Automated LC-MS Sample Cleanup Using Turbulent Flow Chromatography
Too much interfering matrix? Discover how to streamline your...
Read More
5 Key Takeaways from LC Columns Tips and Tricks Webinar
The webinar, hosted by Hai Ning, Regional Marketing Manager ...
Read More