Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Food and Beverage / Trace Origin of Food with Strontium Isotope Analysis Using MC-ICP-MS
Trace Origin of Food with Strontium Isotope Analysis Using MC-ICP-MS
By AnalyteGuru Staff, 08.21.2023
The apple never falls far from the tree. … But where is the tree?
Tracing the origin of food is important for both producers and consumers.
No one wants to pay more than usual for an Italian Golden Delicious apple only to find out that it’s actually a crab apple from Scunthorpe, U.K.. Equally, growers want to be fairly compensated for maintaining traditional, sustainable farming practices. Food labeling provides a guarantee for both producers and consumers, ensuring quality for the consumer and protection for the regional producers from cheaper imitations.
Food labeling, however, can be manipulated and used fraudulently. Objective analytical methods are required for reliable food tracing. Isotopes provide a powerful means to trace the origin of food products. The basic idea is that crops inherit their isotopic fingerprints from the soil, water and air from which they are grown, and this isotopic signature passes into the products made from those raw ingredients.
There are many different isotope systems that we can choose to look at, but one of the most powerful for food origin is strontium isotopes. The strontium isotopic signature of rocks (and therefore soil) is highly variable and often can be different region to region, or even valley to valley.  What’s more, it doesn’t change with the season, and is not altered when it is taken up by plants. As such, strontium isotopes provide a very useful tool for spatially pinpointing the origin of food.
Read our new application note to find out how strontium isotopes measured on our multicollector ICP-MS (MC-ICP-MS) has been used to identify apples and cereal crops within northern Italy.
Mushroom image
Deciphering Selenium Nutrition Using Speciation Analysis with ICP-MS
Many elements, such as chromium, zinc or selenium are consid...
Read More
Cutting-Edge Triple Quad ICP-MS for Clinical Trace Element Analysis
Cutting-Edge Triple Quad ICP-MS for Clinical Trace Element Analysis
Unlocking the power of triple quadrupole ICP-MS for clinical...
Read More
Advancements in Thorium Isotope Analysis: The Role of Neoma MS/MS MC-ICP-MS
Thorium (Th) isotope analysis is important across various sc...
Read More
Ensuring Baby Food Safety: Tackling Toxic Metals with Advanced Technology
Ensuring Baby Food Safety: Tackling Toxic Metals with Advanced Technology
In recent years, the spotlight on the safety of baby food ha...
Read More
AnalyteGuru Staff
Single-Cell Sensitivity and Low-Load Proteomics: Q&A for the Novice
Elemental Analysis in the Battery Recycling Sector: Learn More at the Battery Show