Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Pharmaceutical / Understanding Nitrosamine Drug Substance-Related Impurities (NDSRIs)
Understanding Nitrosamine Drug Substance-Related Impurities (NDSRIs)
By Aaron Lamb, Vertical Marketing Manager, Pharma and Biopharma, Thermo Fisher Scientific 06.26.2025
Introduction
Nitrosamine drug substance-related impurities (NDSRIs) have emerged as a significant concern in the pharmaceutical industry. These impurities are associated with potential carcinogenic risks, making their detection and control crucial for ensuring drug safety and efficacy. This blog aims to provide an in-depth understanding of NDSRIs, their sources, regulatory guidelines, and methods for their detection and control.
tablet image
What are Nitrosamines?
Nitrosamines are chemical compounds containing the nitroso functional group. While some nitrosamines are formed naturally and can be found in foods, beverages, and the environment, others can be synthetically produced. The concern arises from the fact that many nitrosamines have been identified as carcinogenic in animal studies, with implications for human health.
Sources of NDSRI in pharmaceuticals
NDSRI can form during the manufacturing process of pharmaceuticals through various pathways, including:
  1. Reaction of secondary or tertiary amines with nitrosating agents: Secondary or tertiary amines present in drug substances or excipients can react with nitrosating agents such as nitrites, leading to the formation of NDSRIs.
  2. Degradation pathways: Some drug substances may degrade over time, leading to the formation of NDSRIs.
  3. Contaminated raw materials: Raw materials contaminated with NDSRIs or nitrosating agents can introduce these impurities into the final product.
  4. Cross-contamination: Cross-contamination during manufacturing processes can also lead to the presence of NDSRIs in drug products.
Regulatory guidelines
Regulatory agencies like the U.S. Food and Drug Administration (FDA), European Medicines Agency (EMA), and others have issued guidelines to control and limit the presence of NDSRIs in drug products. Key points include:
  1. Risk assessment: Manufacturers must conduct a thorough risk assessment to identify potential sources of NDSRIs.
  2. Analytical testing: Use of validated analytical methods to detect and quantify NDSRIs in drug products.
  3. Control strategies: Implementation of control strategies to mitigate the formation of NDSRIs during manufacturing.
  4. Acceptable Intake (AI) limits: Adherence to specified acceptable intake limits for NDSRIs to ensure patient safety. This is a challenge for NDSRIs as specific toxicology information is often missing so establishing AIs is a problem.
Detection and analysis
Detecting and analyzing NDSRIs requires sophisticated analytical techniques. The most used methods include:
  1. Gas Chromatography-Mass Spectrometry (GC-MS): GC-MS is highly effective for separating and identifying volatile nitrosamines.
  2. Liquid Chromatography-Mass Spectrometry (LC-MS): LC-MS is suitable for non-volatile nitrosamines and provides high sensitivity and specificity.
  3. High-Performance Liquid Chromatography (HPLC): HPLC can be used in combination with other detectors for the analysis of nitrosamines.
  4. Ion Chromatography: Useful for detecting nitrite and nitrate ions, which are precursors to nitrosamine formation.
Control and mitigation strategies
To control and mitigate the formation of NDSRIs, manufacturers can implement several strategies:
  1. Process optimization: Optimizing manufacturing processes to minimize the use of nitrosating agents and amines. This is also a challenge as many APIs contain secondary and tertiary amine functionality.
  2. Raw material quality control: Ensuring raw materials are free from NDSRIs.
  3. Environmental controls: Implementing environmental controls to prevent cross-contamination.
  4. Stability studies: Conducting stability studies to understand the degradation pathways and potential for NDSRIs formation.
  5. Formulation adjustments: Adjusting formulations to replace or reduce the use of amines or nitrosating agents.
Conclusion
NDSRIs present a significant challenge for the pharmaceutical industry, with implications for drug safety and regulatory compliance. By understanding the sources of NDSRIs, adhering to regulatory guidelines, and employing robust analytical and control strategies, manufacturers can effectively manage these impurities and ensure the safety and efficacy of their drug products. Continuous vigilance and advancements in analytical technology will be crucial in addressing the evolving landscape of nitrosamine impurities.
To learn more, download our comprehensive solution guide on nitrosamine impurities analysis now: Nitrosamine Impurities Analysis Solution Guide and visit our website on Nitrosamines Impurity Analysis.
Visit us on LinkedIn: #Nitrosamines #NDSRI
Temperature regulation graphic image
How to Get More Accurate HPLC Quantitation of Semi-Volatiles
Struggling to accurately measure semi-volatile analytes? Str...
Read More
Innovation in Biopharma analysis image
Scaling Up Biopharmaceutical Characterization with the Orbitrap Ascend BioPharma Tribrid Mass Spectrometer
Biopharmaceutical pipelines are expanding rapidly, with incr...
Read More
The Benefits of Charged Aerosol Detection (CAD) in High-Performance Liquid Chromatography (HPLC)
Episode 1: The Benefits of Charged Aerosol Detection (CAD) in High-Performance Liquid Chromatography (HPLC)
High-Performance Liquid Chromatography (HPLC) is a cornersto...
Read More
Advantage of Orbitrap Technology for NDSRI Quantitation: An N-Nitroso Duloxetine Case Study 
In October 2024, thousands of bottles of the popular antidep...
Read More
Aaron Lamb
Vertical Marketing Manager, Pharma and Biopharma, Thermo Fisher Scientific
Why Compound Discoverer 3.4 Software is Worth Your Attention!
New European Wine Scandal on PFAS in Wine