Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Uncategorized / Understanding the Significance of 6:2 FTAB in the World of PFAS
Understanding the Significance of 6:2 FTAB in the World of PFAS
By Vanessa Wright, Global Market Development Manager, Environmental and Food Safety, Chromatography and Mass Spectrometry, Thermo Fisher Scientific 02.27.2024
In the intricate landscape of per- and polyfluoroalkyl substances (PFAS), one compound has been asserting its presence — 6:2 FTAB. 2024 is a year of exploration and discovery, so let’s unravel the mysteries surrounding this specific PFAS compound and delve into its environmental implications, health considerations, and the technological prowess of instruments like the Thermo Scientific Orbitrap Exploris 120 in decoding its secrets.
What is 6:2 FTAB?
6:2 fluorotelomer sulfonamidoalkyl betaine (6:2 FTAB), is a member of the PFAS family, characterized by its perfluorinated structure. The 6:2 designation indicates the length of the carbon chain in the compound, specifically six carbons with two fluorine atoms on either side. As one of the major alternative PFOS (perfluorooctane sulfonate) components in firefighting foams, this seemingly innocuous molecular structure belies potential risks that have garnered attention in scientific circles.
One of the defining characteristics of 6:2 FTAB is its persistence in the environment. The compound’s resistance to degradation poses challenges in natural attenuation processes, leading to concerns about its long-term impact on ecosystems and wildlife.
Similar to other PFAS compounds, 6:2 FTAB exhibits a propensity for bioaccumulation. As it traverses through the food chain, it could accumulate in organisms, potentially reaching concentrations that pose risks to aquatic life and, subsequently, to those who rely on these ecosystems for sustenance.
While research on 6:2 FTAB is still evolving, there are emerging concerns about its potential health effects. Studies have suggested links between PFAS exposure and adverse health outcomes, raising questions about the compound’s impact on human health over time.
The Analytical Edge: Thermo Scientific Orbitrap Exploris 120
To confront the challenges posed by 6:2 FTAB and its PFAS counterparts, advanced analytical tools become indispensable. The Thermo Scientific Orbitrap Exploris 120 emerges as a beacon of precision and sensitivity in the realm of mass spectrometry. The Orbitrap Exploris 120’s high-resolution capabilities enable scientists to discern subtle differences in PFAS compounds, including 6:2 FTAB. This level of detail is essential for accurate identification and quantification.
PFAS compounds are often present in trace amounts, especially in environmental samples. The Orbitrap Exploris 120’s sensitivity allows for the detection of even minuscule concentrations, providing a comprehensive understanding of the compound’s distribution.
Navigating the Future
The combination of scientific inquiry and technological innovation, exemplified by instruments like the Orbitrap Exploris 120, propels us toward a future where we can mitigate the impact of PFAS and safeguard our ecosystems and well-being.
In conclusion, the story of 6:2 FTAB is one of complexity, requiring a multidisciplinary approach for a comprehensive understanding. Through continued research, technological advancements, and collaborative efforts, we pave the way for informed decision-making and a more sustainable coexistence with the chemicals that shape our modern world.
Learn more about use of the Thermo Scientific Exploris 120 Orbitrap HRAM-LC-MS for a sensitive, quantitative and robust HRAM method for the detection and quantitation of known PFAS and unknown PFAS compounds in waters.
Register for the live/on-demand PFAS webinar series, where we cover all aspect of PFAS analytical testing and future-proofing your lab for PFAS detection.
From Complexity to Clarity: Smarter PFAS Identification Workflows in Compound Discoverer Software
Per- and polyfluoroalkyl substances (PFAS) have become a maj...
Read More
pfas image
Understanding PFAS in Pharmaceutical Manufacturing
Introduction to PFAS Per- and polyfluoroalkyl substances (PF...
Read More
Food packaging
Enhancing PFAS Analysis in Food Contact Materials Using Combustion Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) have been used in...
Read More
US EPA Method 1621_1200x600
Understanding PFAS and Combustion-Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) comprise more tha...
Read More
Vanessa Wright
Vanessa Wright is Global Market Development Manager for Environmental and Food Safety, in the Chromatography and Mass Spectrometry group at Thermo Fisher Scientific, creating marketing campaigns showcasing complete workflow solutions for PFAS, pesticides and water testing for environmental and food analytical testing laboratories and research institutions.
CAD vs ELSD: Which HPLC Detector Is Your Better Option?
Analyzing Natural Gas? A New Way to Determine Alkanolamines With Improved Separation Technology