Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Food and Beverage / Unveiling the Advanced Workflow for PAH Analysis in Smoke Flavored Foods
Unveiling the Advanced Workflow for PAH Analysis in Smoke Flavored Foods
By Petra Gerhards, Regional Marketing Manager GC and GC/MS, EMEA, Thermo Fisher Scientific 02.17.2025
Introduction
Polycyclic aromatic hydrocarbons (PAHs) are a prominent class of organic compounds that have raised significant concerns due to their presence in various environmental and food matrices. These compounds are composed of multiple aromatic rings and are primarily formed during the incomplete combustion of organic matter. Sources of PAHs range from natural processes like forest fires to human activities such as vehicle emissions, tobacco combustion, and food preparation techniques like grilling or smoking.
Unveiling the Advanced Workflow for PAH Analysis in Smoke Flavored Foods
In the food industry, PAHs are particularly relevant in the context of smoke-flavored products. While smoke flavorings enhance the sensory properties of foods, they also pose potential risks due to the formation of PAHs during the smoking process. As such, stringent analytical methods are essential for detecting and quantifying PAHs to ensure consumer safety. In this blog, we explore a comprehensive GC-MS/MS workflow specifically designed for PAH analysis in smoke-flavored foods, with a focus on innovation in sample preparation, instrumental sensitivity, and regulatory compliance.
Understanding smoke flavors and PAHs
Smoke flavorings are derived from the controlled combustion of specific wood types, which are subsequently condensed, fractionated, and purified. These flavorings are used in foods such as fish, meat, sauces, and even snacks to replicate traditional smoking processes. The primary flavoring components include phenols and carbonyl compounds (aldehydes and ketones).
However, PAHs can also be introduced during this process. Examples include naphthalene (two aromatic rings), anthracene, and phenanthrene (three aromatic rings), which are considered lower molecular weight PAHs, alongside heavier compounds such as benzo(a)pyrene and coronene. Regulatory frameworks in the European Union (e.g., Regulation (EC) No. 2065/2003) ensure that only authorized smoke flavoring products are used, with maximum allowable levels for PAHs.
The challenges of PAH analysis in food matrices
Analyzing PAHs in food matrices, particularly those with high-fat content, presents unique challenges. These include:
  1. Complexity of matrices: Foods like smoked trout and tofu contain fats and proteins that can interfere with analytical detection.
  2. Stringent detection requirements: Regulatory limits demand detection of PAHs at extremely low concentrations (picogram to femtogram levels).
  3. Sample throughput: Laboratories need workflows that are both sensitive and efficient to handle routine analysis of multiple samples daily.
A comprehensive workflow for PAH analysis
1. Sample preparation: The foundation of accurate results
Sample preparation is a critical step in PAH analysis, especially for fat-containing matrices. A robust and systematic approach minimizes matrix interferences and enhances analyte recovery. The workflow includes:
  • Homogenization: A 5-gram sample is weighed and homogenized in a glass tube. Internal standards are added, followed by equilibration for 10 minutes.
  • Liquid extraction: Samples are extracted using hexane in an ultrasonic bath. This process is repeated five times to ensure thorough extraction.
  • Evaporation and cleanup: Extracts are evaporated to 1 mL and subjected to Solid Phase Extraction (SPE). Two SPE steps are employed:
    • Primary SPE (HyperSep Florisil): Removes major interferences.
    • Secondary SPE (HyperSep Phenyl): Further purifies the sample for precise GC-MS/MS analysis.
2. Advanced instrumentation: The role of GC-MS/MS
Instrumentation plays a pivotal role in achieving the sensitivity required for PAH detection. The Thermo Scientific TSQ 9610 GC-MS/MS system incorporates cutting-edge technologies to address the demands of PAH analysis:
  • Advanced electron ionization (AEI): This ion source provides superior sensitivity through highly efficient ionization and a tightly focused ion beam. This ensures robustness even with challenging matrices.
  • NeverVent technology: Unique design features allow vent-free column and ion source exchanges, maximizing instrument uptime.
  • Dual filament design: Extends the operational lifespan and reduces maintenance frequency.
3. Importance of glass quality in analytical accuracy
The quality of glass materials used in vials and liners significantly impacts analytical outcomes. Thermo Fisher Scientific’s LinerGold technology addresses the challenges posed by active glass surfaces that can adsorb PAHs, leading to signal loss or contamination. By providing highly inert glass materials, the workflow ensures consistent and accurate results.
Results and key findings
The GC-MS/MS workflow for PAH analysis has been rigorously tested on various food matrices, demonstrating its robustness and reliability. Key findings include:
  • Ultra-low detection limits: The system achieves femtogram-level detection, with examples such as chrysene (150 fg in smoked trout) and coronene (320 fg in smoked trout).
  • Precision and reproducibility: Calibration curves for compounds like pyrene and benzo(k)fluoranthene exhibit excellent linearity, ensuring reliable quantification.
  • Wide applicability: The workflow performs well across diverse matrices, from smoked trout to tofu, maintaining compliance with regulatory limits.
Regulatory compliance and consumer safety
Ensuring compliance with EU regulations is a cornerstone of PAH analysis in smoke-flavored foods. The presented workflow not only meets these standards but also provides a scalable solution for routine laboratories. By integrating advanced sample preparation, state-of-the-art instrumentation, and high-quality consumables, laboratories can confidently deliver accurate and reliable results.
Conclusions and future perspectives
The detection and quantification of PAHs in smoke-flavored foods require a synergistic approach that combines meticulous sample preparation with innovative GC-MS/MS technology. The Thermo Scientific TSQ 9610 system, with its AEI source and NeverVent technology, sets a new benchmark in analytical sensitivity and robustness.
This workflow underscores the importance of quality at every step—from sample preparation to vial selection—to achieve the necessary detection limits and regulatory compliance. As analytical demands evolve, these advancements pave the way for safer food production and enhanced consumer protection.
Additional information:
  • TSQ 9610 Triple Quadrupole GC-MS
  • Chromatography Columns and Consumables Now Available Online
Join our free webinars:
20.3.2025 English, 11 am CET, 1 hour
Advanced workflow for PAH analysis of smoke flavored foods
25.3.2025 German, 11 am CET, 1 hour
Innovativer Workflow für die PAK-Analyse in Lebensmitteln mit Raucharoma
After the actual dates they are available on demand using the same links.
Visit us on LinkedIn: #SmokeFlavors #Pah #GCMS #Consumables
MOSH MOAH image
MOSH/MOAH Analysis: Technology, Regulation, and the Road Ahead
Mineral oil hydrocarbons (MOSH) and mineral oil aromatic hyd...
Read More
Dr Araceli Rivera-Pérez
GC Orbitrap Guru Series #3: Dr Araceli Rivera-Pérez
Hey everyone! The response to my second blog with Brooklynn ...
Read More
Dr Kerstin Krätschmer
GC Orbitrap Guru Series: Dr. Kerstin Krätschmer
It’s been 10 years! Yes, in 2015 “gas chromatography...
Read More
Tackling PAH Analysis in Environmental Samples: Overcoming the Challenges with Advanced GC-MS Technology
Polycyclic aromatic hydrocarbons (PAHs) are a group of organ...
Read More
Petra Gerhards
Petra Gerhards, Dipl-Ing, is Regional Marketing Manager of GC and GC-MS for EMEA at Thermo Fisher Scientific. She has more than 29 years of experience in the fields of GC-MS, SPE and LC-MS. Since joining the regional team she has contributed to workflow solutions combining vials and closures with SPE solutions, GC-MS and LC-MS. She works with KOL's on data for regional specific marketing campaigns, organizes in-house seminars and works on customer specific solutions. Her main expertise is in the field of doping and drugs-of-abuse analysis.
Benefits of Automating Your Antibody QC Analytical Workflows
Ensuring Baby Food Safety: Tackling Toxic Metals with Advanced Technology