Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Chromatography Data System / USP Chapter <621>: Chromeleon CDS Flexibility Simplifies Calculation of Updated Values
USP Chapter <621>: Chromeleon CDS Flexibility Simplifies Calculation of Updated Values
By Crystal Welch, Global Product Marketing Manager – CMD Software, Thermo Fisher Scientific 12.13.2023
Another USP Chapter <621> update?
Starting with the obvious – the USP <621> chapter on system suitability calculations has (once again!) been updated. 
I know we were all prepared for the switch to harmonize the calculations for Resolution, Plate count, and signal-to-noise from the USP to match the EP and JP, but on Aug. 25, 2023, the USP announced there would be a return to the prior calculation for USP signal-to-noise with an official, revised, version of <621>1 being posted on Oct. 1. The rest of the harmonized calculations have not been impacted and the implementation date for methods is set for May 1, 2025.  
Chromatography calculations
For those wishing to report the harmonized Plate count and Resolution calculation in Chromeleon chromatography data system (CDS), configure your Plates and Resolution columns to report the “EP Formula” instead of the “USP Formula” as outlined by our v.2.0 support article USP 621 Harmonization – Chromeleon Guidance2 and demonstrated in the video below.
Video: Calculation options featuring USP <621> calculations
Chromeleon CDS was engineered to give flexible options and the preceding video goes through some of the ways to calculate updated values without having to upgrade or change anything. All of these skills can be applied to reports, too.
Resources
For free access to our public knowledge base2, click here.
For users with support contracts who would like to search through our support knowledge base 3, click here.
To find out more about our Chromeleon CDS service contracts, click here.
  1. You must have a registered USP account (free) to view.
  2. You must register a Thermo Scientific account (free) to see articles in our public knowledge base.
  3. You must have a support contract account to access our support knowledge base.
Illustrated scientists, sustainability icons, and arrow pointing to a laboratory instrument.
Modernizing Routine HPLC Methods: Aligning Productivity, Compliance, and Sustainability
Interested in HPLC method modernization and compliance? I an...
Read More
1975 text image
Innovations of 1975 and the Birth of the First Commercialized Suppressed Ion Chromatograph
The year 1975 was a remarkable period in the history of tech...
Read More
US EPA Method 1621_1200x600
Understanding PFAS and Combustion-Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) comprise more tha...
Read More
IC Golden Jubilee blog header image
What is the Ion Chromatography Golden Jubilee? Celebrating 50 Years of IC Innovation
The year 2025 marks the golden jubilee of ion chromatography...
Read More
Crystal Welch
Crystal Welch started her industry journey in large molecule manufacturing and quality, working through several more years in process development and contributing to projects in validation, quality system initiatives, process improvements, product characterization, and regulatory submissions. After more than a decade of being a customer, she then offered consultation and support to the same departments in the field, helping others to accomplish projects in their own facilities. Now she spends her time reading regulatory policy updates, following cutting edge informatics product development, and summarizing user feedback to provide improvement suggestions with the goal to make scientists' lives easier.
Take a Step Closer to Unlocking Full Potential of AAV Gene Therapy – Download the Poster
Label-free Quantitative Proteomics Analysis Advances Understanding of Biological Processes