Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Ion Chromatography / What Really Matters When You Purchase a New Instrument for Your Lab
What Really Matters When You Purchase a New Instrument for Your Lab
By AnalyteGuru Staff, 06.22.2020
The ongoing COVID-19 pandemic has made us stop and define for ourselves what really matters. What if we consider what really matters in our laboratory? When we purchase a new instrument, we naturally spend time reviewing that instrument’s specifications. These are certainly important, but they are rarely the reason we purchase an instrument. We purchase an instrument to run one or more applications, and that is what really matters. In the Thermo Scientific Dionex Ion Chromatography Applications Lab, we have been aware of that fact for over four decades and have created a large library of applications to help our customers be successful. These applications consist of a lot more than running a simple standard. After all, who buys an instrument to run a standard? I submit that we provide complete applications that are more detailed than you will find from other instrument companies, and often done just as you would do them.
So, what do I mean by complete? There are many components to complete and they will vary with the application and its purpose, but let’s look at two recent applications our lab developed to demonstrate. All our applications can be found in the AppsLab Library of Analytical Applications and the links will take you there so, if you would like, you can read the details. The first is the determination of ammonia impurity is potassium bitartrate. This project was done to modernize the United States Pharmacopeia’s (USP) Potassium Bitartrate monograph by replacing the colorimetric ammonia impurity test with a fast and robust ion chromatography (IC) test. Looking closely at this application note you see the components of a complete application. All the equipment, consumables, and chemicals needed for the application are listed along with their sources and catalog numbers. There are detailed instructions for preparing eluents, preparing samples, calibration, and setting up the experiment. Additionally, we show the results of our robustness studies that follow USP criteria so that you can judge if the method meets your needs. Part of that study is to test the method on second column but of a different lot. We typically test two columns and recommend that customers do so when designing a method they plan to use indefinitely, or that will be used by other labs. Key performance parameters are provided so you can check your set up.
The second application is the determination of halogens and sulfur in liquified petroleum gas (LPG). This is far from a typical IC application as it involves incremental combustion of LPG and capturing the resulting gases in a liquid that is then analyzed by IC. While that sounds complicated, in addition to having all the components discussed in our first example, this application note guides you on how to safely install, execute, and shut down this application. For all our applications, our goal is to make you successful in running the application or provide you a blueprint for designing your own similar application. In the end, that is what really matters.
Scientists are looking at data charts on a computer with test tubes beside them.
Ion Chromatography Turns 50: Chromeleon Software’s Tribute to a Lifelong Partner
This year marks a special milestone: the 50th anniversary of...
Read More
US EPA Method 1621_1200x600
Understanding PFAS and Combustion-Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) comprise more tha...
Read More
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
In industrial processes, accurately determining halogens and...
Read More
IC Golden Jubilee blog header image
What is the Ion Chromatography Golden Jubilee? Celebrating 50 Years of IC Innovation
The year 2025 marks the golden jubilee of ion chromatography...
Read More
AnalyteGuru Staff
To Edge Your View on Ion Chromatography – The new eBook on the basics of this well-established analytical method
A Fully Automated Method for MCPD- and GE-esters and the Importance of Glass Quality of the Used Autosampler Vials