Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Isotope Ratio MS / Why Care About Past Climate? Find Answers in Isotope Analysis
Why Care About Past Climate? Find Answers in Isotope Analysis
By Dr. Claudia Bouman, Product Manager, Multicollector ICP-MS and Noble Gas MS, Thermo Fisher Scientific, Bremen, Germany, 04.06.2018
Why should we care about the climate of the past?
Our climate is continuously changing and has severe impacts on society and our environment. We hear news of floods, storms and droughts almost daily. Studying our past climate helps us understand climate changes and gives scientists clues that will help us plan for future climate changes.
A great deal of climate research is focused on ensuring that climate models can simulate most aspects of the present-day climate. The study of past climate, paleoclimatology, helps scientists to improve the ability of computer models to simulate future climate.
Modern climate records from satellites and other equipment generally cover less than ~150 years. This is too short to examine the full range of climatic variability. For this reason, scientists use imprints created during past climates to interpret paleoclimate. An example is the use of fossils and other shelled organisms from foraminifera, as well as sediments in aquatic and marine environments. Once the organisms die, their shells, made from calcium carbonates, get buried in sediments on the bottom of lakes and oceans. By drilling cores into the sediment layer, scientists collect these fossils and use them to “read” past climate.
How does this work?
The stable oxygen isotope composition of the fossils can be used to infer past water temperatures. These oxygen isotopes are found naturally in both the atmosphere and dissolved in water. Warmer water tends to evaporate off more of the lighter isotopes leaving the water isotopically heavier. This means that when shells grow in warmer waters they will be enriched in the heavier isotope.
Find out more about the process in this video:
https://www.youtube.com/watch?v=oaOfeSJZ3lY
For the analysis of isotopes in (calcium) carbonates, Thermo Fisher Scientific offers dedicated instrumentation. The Thermo Scientific 253™Plus10™kV IRMS, together with the Thermo Scientific™Kiel IV™Carbonate Device, is the gold standard for carbon and oxygen isotope analysis of carbonates, producing world-class data from small foraminifera samples. With the 253 Plus IRMS and Kiel IV Carbonate Device precisions of better than 0.1 ‰ can be reached for total carbonate amounts of down to 6 µg. With this, paleoclimatologists can resolve 0.5 °C temperature changes.
Learn more >
From Complexity to Clarity: Smarter PFAS Identification Workflows in Compound Discoverer Software
Per- and polyfluoroalkyl substances (PFAS) have become a maj...
Read More
ACT platinum cover image
A Greener HPLC Guide: How ACT Labels Help Promote Lab Sustainability
Labs use up to 10 times more energy than offices. Here’s h...
Read More
ba-003411-sw-compound-discoverer-scripting-nodes-webinar-ba003411
Take Control of Your Data: An Introduction to the Scripting Node in Compound Discoverer Software
In small molecule research, your data is only as powerful as...
Read More
Advancements in Thorium Isotope Analysis: The Role of Neoma MS/MS MC-ICP-MS
Thorium (Th) isotope analysis is important across various sc...
Read More
Dr. Claudia Bouman
Claudia Bouman is Product Manager at Thermo Fisher Scientific in Bremen, Germany, responsible for the Multicollector ICP-MS and Noble Gas MS product line. Claudia graduated from the University of Utrecht and completed her Ph.D. in Isotope Geochemistry at the VU in Amsterdam, Netherlands. In 2002, Claudia joined the Bremen factory to work as an Application Specialist. Since then, she had various roles in Product Management, Sales Support and Marketing.
Can You Really Get Twice as Much Done When Running Samples on Your LC System?
Science Taking Lead on Cannabis to Accelerate Research