Mpox cases
Learn more about our infectious disease testing solutions to support your research here.
With public health officials reporting more than 15,600 cases of mpox infections and 537 deaths in Central and East African countries, the WHO Director-General has “determined that the upsurge of mpox in the Democratic Republic of the Congo (DRC) and a growing number of countries in Africa constitutes a public health emergency of international concern.”1
The clade 1 mpox variant, responsible for the majority of current infections, has been detected in neighboring DRC countries, and in an increasing number of non-African countries where this variant is not known to be endemic.2,3 To date, only one case outside the endemic region has been confirmed, suggesting limited transmission to-date outside of Africa.
Public health labs respond to mpox virus
The CDC has issued an updated Health Alert Network advisory recommending that clinicians and laboratories “send clinical specimens collected from patients who traveled from DRC or its neighboring countries, or had close or intimate contact with symptomatic people from these countries, to a laboratory that can perform clade-specific testing as quickly as possible”.2
For the most up-to-date information, please see the CDC Mpox outbreak website.
WHO prefers detection of viral DNA by PCR as antigen- and antibody-based methods typically do not distinguish between Orthopoxviruses. Specimens collected directly from the rash—skin, fluid or crusts, or biopsy where feasible—are preferred.4
What is Mpox virus (MPX or MPXV)?
- Mpox is a DNA virus endemic to Central and West African countries.
- An Orthopoxvirus, mpox is part of the genus of viruses that cause diseases like smallpox. It is associated with skin lesions and lymph node swelling among other symptoms.
- Tropical rainforest animals including rodents and non-human primates are the primary carriers of mpox.
- There are two distinct clades of MPXV, a West African clade and a Congo Basin clade. People infected by virus from the Congo Basin/Central African clade experience approximately 10% case fatality rate while those infected by the West African clade experience a <4% case fatality rate.3
[1] If RNA and DNA analyses are performed on the same plate using a common thermal cycling protocol, the TaqPath™ 1‑Step Multiplex Master Mix (No ROX™) (Cat. No. A28522) can be used instead of TaqPath™ BactoPure™ Microbial Detection Master Mix (No ROX™).
For research use only. Not for use in diagnostic procedures.
Sources
- https://www.who.int/news/item/14-08-2024-who-director-general-declares-mpox-outbreak-a-public-health-emergency-of-international-concern
- https://emergency.cdc.gov/han/2024/han00513.asp
- https://www.folkhalsomyndigheten.se/the-public-health-agency-of-sweden/communicable-disease-control/disease-information-about-mpox/one-case-of-mpox-clade-i-reported-in-sweden/
- https://www.who.int/publications/i/item/WHO-MPX-Laboratory-2024.1
© 2022 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. TaqMan is a registered trademark of Roche Molecular Systems, Inc., used under permission and license.
Leave a Reply