The importance of biomedical image segmentation
Researchers studying whole bones utilize volumetric imaging techniques such as micro-computed tomography (micro-CT) to understand the complex 3D structures underlying bone morphology. Biomedical image segmentation is often an essential part of this analysis, helping to reveal the finer details of bone composition. The process of manually segmenting thousands of micro-CT slices can, however, slow the path from imaging to biological insights. To address this challenge, Professor Andrew Lee of Midwestern University has recently developed BONe, a set of deep-learning tools that run within Thermo Scientific Amira and Avizo Software, which support bone segmentation at scale. In this workflow, Amira-Avizo Software brings model-based segmentation together with 3D visualization and 2D inspection to streamline data processing while the researcher subsequently applies their anatomical expertise to check and refine results.
“I wanted to reduce segmentation as a major workflow bottleneck while prioritizing usability, scalability, and reproducibility.”
— Professor Andrew Lee, Midwestern University
Why does whole-bone segmentation become a bottleneck?
Professor Lee’s research explores how diet and exercise affect the robusticity of mouse limb bones. By analyzing whole bones rather than a single cross section, his lab can examine the intricate geometrical variations that occur along each bone. Before such biological measurements could begin, thousands of image slices needed accurate segmentation.
For example, one of their studies, highlighted in this blog, included 74 bones collected over 20 scans. Manual segmentation of such data can serve as an excellent reference, but can be time consuming, and is dependent on the expertise of the person doing the work. Automatic thresholding is a common, alternate approach that can quickly segment datasets based on image contrast, but it can miss thin trabeculae, confuse background with internal spaces, and respond inconsistently to artifacts or changes in brightness. Across many specimens, the shortcomings and challenges of these two methods can make segmentation a limiting step in the micro-CT analysis of bone.
Deep-learning-based image analysis is a comparatively new method that can readily meet the challenges posed by traditional manual segmentation and automatic thresholding. Instead of relying on one intensity cutoff, the model learns from a set of reference segmentations, using image features and surrounding spatial patterns to distinguish bone, medullary spaces, and background.
How does BONe work within Amira-Avizo Software?
The BONe deep-learning tools developed by Professor Lee support three parts of the segmentation process:
- BONe DLFit trains or fine-tunes a model
- BONe DLPred applies a trained model to new scans
- BONe IOU compares a predicted segmentation with a reference mask
These modules are available within Amira 3D and Avizo 3D Software, as well as in standalone versions that feature a similar interface. The models can also be moved between the different software implementations.
BONe brings model-based segmentation into the Amira-Avizo Software workflow. Typically, a researcher will use BONe DLPred to apply a compatible pretrained model to a scan in the project (e.g., BP-2D-03a, which is trained to distinguish bone and medullary pores in micro-CT scans of mammalian bone). The resulting multiclass segmentation is returned to the project for 2D inspection, 3D visualization, and, where needed, manual refinement. If the scan differs substantially from the model’s training data, the researcher can fine-tune the model with BONe DLFit and evaluate it against reference masks with BONe IoU.
What should researchers check after deep-learning-based image segmentation?
While a 3D, deep-learning-based segmentation may look convincing, it still needs to be reviewed by the researcher performing the analysis. In the example shown here, the model appropriately identified bones and medullary spaces, but the results also included small background artifacts and areas where segmentation extended from the medullary spaces into the background. These areas may need to be cleaned manually, depending on the pervasiveness of these errors.
Professor Lee recommends quality-control review for every segmentation, especially when a scan differs from the data used to train the model. Visualization and inspection tools in Amira-Avizo Software support this review as part of the same workflow as prediction.
“BONe does not remove the need for anatomical expertise, it just redirects the expertise from tracing every slice to training, evaluating, and refining a scalable segmentation workflow.”
— Professor Andrew Lee, Midwestern University
When should researchers fine-tune a pretrained segmentation model?
Direct inference is a practical starting point when new scans resemble the anatomy and imaging conditions represented by the model’s training data. Researchers can load the pretrained model, run BONe DLPred, and inspect the result.
If scans differ in resolution, contrast, anatomy, or artifact profile, the initial performance of the segmentation may need to be improved. In such cases, Professor Lee recommends using a representative set of reference segmentations to fine-tune the pretrained model with BONe DLFit. The resulting task-specific model can then be applied with BONe DLPred; BONe IOU can also quantify overlap when reference masks are available. The benefit of this approach is that researchers can adapt an existing model rather than train a new one from scratch.
How should researchers evaluate model performance?
A segmentation model’s practical value depends not only on benchmark performance but also on how well its training data represent the scans it will encounter. For the examples shown in this blog, the training data included multiple skeletal elements and variation in voxel size, noise, specimen tilt, brightness, beam hardening, as well as drill holes and broken surfaces. The fitting process added further variation through rotations, flips, and cropping, as well as changes in brightness and contrast.
In five-fold cross-validation, the resulting revised BP-2D-03a model achieved a mean intersection over union (IoU) of approximately 91%. However, mean performance varied from about 85% to 97% across folds, with some lower results associated with unusual or out-of-domain scans. These results show why a benchmark score alone cannot establish how a model will perform on every new dataset. Researchers should assess performance on scans that reflect their specific anatomy and imaging conditions, reviewing the predictions before they are used for downstream measurements.
A more scalable image-analysis workflow
Professor Lee’s work shows how Amira-Avizo Software can support deep-learning-based segmentation workflows that include prediction, visualization, inspection, and refinement. BONe DLPred can take on repetitive segmentation across large scans, and researchers can subsequently use Amira-Avizo Software to review the results and decide whether manual cleanup or model fine-tuning is needed.
For labs facing a growing volume of micro-CT data, this approach serves as a practical way to ease the burden of segmentation and analysis; instead of spending time tracing every slice, teams can focus on understanding what the images reveal. With deep-learning-based segmentation, researchers can test a pretrained model on representative scans, review its output in 2D and 3D, and adapt it further to meet the specific needs of their data.
Explore more features and capabilities of Amira Software at thermofisher.com/amira >
Leave a Reply