Twin-Screw Granulation in Oral Solid Dosage Manufacturing – Process, Benefits & Optimization
Oral solid dosage forms, such as tablets and capsules, are among the most common and preferred methods of drug delivery due to their convenience, stability, and patient compliance.
The production of these pharmaceuticals requires precise control and optimization of several processes, from raw material preparation to final packaging. Understanding these stages is crucial to help ensure efficacy, safety, and quality. In this article, we explore the various stages of manufacturing oral solid dosage pharmaceuticals, highlighting twin-screw granulation.
Key processes in the production of oral solid dosage pharmaceuticals
  1. Raw material preparation: The first step in the production process is the preparation of raw materials, including active pharmaceutical ingredients (APIs) and excipients. These materials must be accurately weighed, milled, and blended to achieve uniformity. Proper handling and storage are essential to maintain their stability and prevent contamination.
  2. Granulation: Granulation is a critical process that involves the aggregation of powder particles to form granules. This can be achieved through various methods, such as wet granulation, dry granulation, and twin-screw granulation. Granulation improves the flow properties of the powder, enhances compressibility, and helps ensure uniform distribution of the API.
    • Dry granulation: Involves compressing powder into slugs or sheets and then milling them into granules.
    • Wet granulation: Uses a liquid binder to agglomerate powder particles, followed by drying.
    • Twin-screw granulation: Utilizes twin-screw extruders to continuously mix and granulate materials.
  3. Blending: After granulation, the granules are blended with excipients to help ensure uniform distribution of the API and achieve the desired properties. Blending must be controlled to prevent segregation and achieve homogeneity.
  4. Compression: The blended granules are compressed into tablets using a tablet press, forming tablets of specific shape, size, and hardness. Compression parameters must be optimized for integrity and dissolution properties.
  5. Coating: Tablets may be coated to enhance appearance, mask taste, protect the API, and control drug release. Common techniques include film and sugar coating. Coating solutions are sprayed and dried to form a uniform layer.
  6. Encapsulation: For capsules, granules or powder are filled into gelatin or HPMC shells, ensuring precise dosage and protecting the API from degradation.
  7. Quality Control: Rigorous quality control measures help ensure the final product meets standards, including testing for uniformity, potency, dissolution, and stability.
  8. Packaging: The final step is packaging tablets and capsules in blister packs, bottles, or other containers to protect them and help ensure patient convenience, maintaining the integrity and stability of the dosage forms.
Introduction to twin-screw granulation
As noted, granulation is a critical process in pharmaceutical manufacturing, essential for producing tablets and capsules with the desired properties.
The twin-screw granulator consists of two screws rotating within a barrel, creating a mixing and granulating zone. The screws’ configuration and speed, along with the feed rate of materials, are critical parameters that influence the granulation process. By optimizing these parameters, manufacturers can achieve the desired granule characteristics. This method contrasts with traditional batch granulation, offering key advantages, including continuous processing and better scalability
Key advantages of twin-screw granulation
  1. Enhanced process control: Twin-screw granulation allows for precise control over the granulation process, leading to consistent product quality. The continuous nature of the process enables real-time monitoring and adjustments, helping to ensure optimal granule size and distribution.
  2. Improved product quality: The uniform mixing and granulation achieved with twin-screw technology result in high-quality granules with excellent flow properties. This consistency translates to better tablet formation and reduced variability in the final product.
  3. Efficiency and scalability: Twin-screw granulation is highly efficient, reducing processing times and material wastage. Its continuous nature makes it easily scalable, accommodating varying production demands without significant changes to the equipment or process.
Twin-screw granulation is particularly beneficial in the production of oral solid dosage forms because it can granulate a wide range of materials, from active pharmaceutical ingredients (APIs) to excipients, helping to ensure uniformity and consistency in the final product.
While twin-screw granulation offers numerous benefits, it also presents certain challenges and considerations. These include the need for specialized equipment and expertise, as well as potential issues with material handling and process optimization. Manufacturers must carefully consider these factors to fully leverage the advantages of twin-screw granulation.
Conclusion
The production of oral solid dosage pharmaceuticals is a complex and meticulously controlled process that involves multiple stages, from raw material preparation to final packaging. By understanding and optimizing these processes, manufacturers can help ensure the consistent quality, efficacy, and safety of their products.
Twin-screw granulation represents a major advancement in pharmaceutical manufacturing, offering improved process control, product quality, and efficiency. Its applications in producing oral solid dosage forms make it a valuable technique for manufacturers seeking to optimize their production processes. As technology continues to evolve, twin-screw granulation is poised to become even more integral to the pharmaceutical industry.
Additional resources
Dirk Leister
Written by:
Dirk Leister
Manager Applications & Customer Support , Thermo Fisher Scientific
Dirk Leister is a marketing manager at Thermo Fisher Scientific focused on extruder and compounder technology used in the pharmaceutical, food, and polymer industries.
Read more Leister, Dirk