Scalability in Bioprocessing with the DynaDrive 5 L Single-Use Bioreactor
Reimagining Scalability in Bioprocessing: Advancing Process Development with the DynaDrive 5 L Single-Use Bioreactor
Experience the innovation of DynaDrive S.U.B. now available at 5L for process development
In the rapidly evolving field of bioprocessing, scalability and efficiency are paramount. The introduction of the 5 L benchtop Single-Use Bioreactor (S.U.B) marks a significant advancement in process development, offering numerous advantages for laboratories. This innovative bioreactor is designed to be compact yet powerful, facilitating scalability and offering enhanced performance that can help transform your research and development processes.
Excellent scalability
One of the standout features of the DynaDrive 5 L Single-Use Bioreactor is its excellent scalability. Researchers can now experience the same reactor design and film from 1 L to 5,000 L, facilitating consistent and reliable scale-up. This consistency is crucial for maintaining the integrity of bioprocesses as they transition from small-scale experiments to large-scale production.
Enhanced performance and efficiencies
Efficiency is a key factor in bioprocessing, and the DynaDrive 5 L Single-Use Bioreactor is designed to deliver on this front. Compared to traditional glass bioreactors, this innovative system offers up to a 30% increase in workflow efficiency. This significant boost in efficiency can help accelerate your development process, reduce time-to-market, and ultimately enhance the overall productivity of your laboratory operations.
Environmental impact
Various volumes of DynaDrive Single-Use Bioreactors range from 1 L to 5,000 L
In addition to its performance benefits, the DynaDrive 5 L Single-Use Bioreactor also supports environmental sustainability. Traditional glass bioreactors consume large amounts of water, which can be a significant environmental burden. In contrast, the DynaDrive 5 L Single-Use Bioreactor can save up to 18,000 liters of water annually, making it a more sustainable choice for modern laboratories.
Versatile performance
Flexibility is another key advantage of the DynaDrive 5 L Single-Use Bioreactor. With a 5:1 turndown ratio, this system offers versatility that can adapt to various bioprocessing needs. Whether you are conducting small-scale research or scaling up to commercial production, the DynaDrive 5 L Single-Use Bioreactor is designed to accommodate your requirements effectively.
Elevate your research
The DynaDrive 5 L Single-Use Bioreactor is not just a piece of equipment; it is a gateway to elevating your research and transforming your process development. Its compact size, combined with its powerful performance, makes it an ideal choice for laboratories looking to enhance their bioprocessing capabilities. By integrating this innovative technology into your workflow, you can aim to achieve greater efficiency and scalability.
Are you ready to elevate your process development?
The DynaDrive 5 L Single-Use Bioreactor represents an exciting leap forward in bioprocessing technology. Its excellent scalability, enhanced performance, and water savings make it a game-changer for process development laboratories. As the demand for efficient and sustainable bioprocessing solutions continues to grow, the DynaDrive 5 L Single-Use Bioreactor stands out as a versatile and powerful tool that can help researchers achieve their goals.
See the new DynaDrive 5 L Bioreactor
Connect with a technical specialist to discuss more
Explore the DynaDrive 3D product animation
Download DynaDrive technical note
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.