Phenol red is commonly found in many cell culture media formulations, offering a convenient way for researchers to monitor cell health. It enables a quick and easy visual indicator of culture conditions. However, it is important to be aware of its potential interactions in certain experimental setups. Understanding the role of phenol red can help researchers make informed decisions to ensure the success of their cell culture experiments.
What is Phenol Red?
Phenol red is an organic dye that serves as a pH indicator commonly used in cell culture media. It visibly changes color as the pH of the medium shifts. It turns yellow in acidic conditions (pH ~6.8 or lower), red in neutral pH ranges (pH ~7.0–7.4), and pink in basic environments (pH >8.2). This helps provide a quick way to monitor the condition of cell cultures without any additional equipment.
Phenol red is a weak acid (pKa ≈ 8.0). It either gains or loses protons depending on the pH of the environment, which in turn changes its color. It’s considered stable in standard media conditions and is included basal media such as DMEM and RPMI 1640.
What is Phenol Red used for?
The main function of phenol red in cell culture media is to serve as a pH indicator in real time. As cells grow, they produce metabolic waste such as CO₂ and organic acids, which lower the pH of the media. When this happens, phenol red shifts from a red color towards a yellow color, which gives the user a clear visual cue that the medium may be depleted or that the culture environment may not be at optimal levels. On the other hand, a color change towards a pink color can signal a high pH environment.
Phenol red also acts as an indicator for contamination. Bacteria and fungi can acidify the culture media as they grow, which causes the phenol red to change color before turbidity or other visible signs of contamination appear. This makes it a useful tool for maintaining culture quality and avoiding compromised experiments.
Why is Phenol Red in cell culture media?
Phenol red is added to most commercially available culture media because it’s usually inert in standard experiments. It allows users to monitor culture health visually and quickly, which is helpful when handling multiple cultures at once.
However, phenol red isn’t always completely inert. Several studies have shown that phenol red can act as a weak estrogen mimic, especially in estrogen receptor–positive cell lines like MCF-7 breast cancer cells. Because of this, hormone and receptor-sensitive assays are typically done in phenol red-free media, to minimize background estrogenic effects.
Why would I want to include Phenol Red in cell culture media?
- Visual indicator: Phenol red can visually show the user shifts in pH, helping monitor acidification or detect contamination early.
- Adaptability: Phenol red is usually included in most media systems that you can buy. This requires no extra handling of materials.
- Low toxicity: For most cell lines and applications, phenol red is safe and doesn’t interfere with growth or viability.
- History of use with wide range of cell lines: Phenol red has been in cell culture with a wide range of cell types for decades giving it a proven track record.
Why would I not want to use Phenol Red?
- Estrogen-like activity: It can stimulate estrogen receptors, which can interfere with hormone signaling studies. For hormone-sensitive cells, researchers may consider using phenol red-free medium and supplementing with estrogen receptor agonists for greater experimental control.
- Fluorescence Imaging or Absorbance-based Assays: The color of phenol red can create background noise or affect measurements of light-based assay readouts. Researchers should employ “media only” control conditions to quantify these effects or consider use of phenol red-free systems when these assays are employed.
- Reactivity in specialized systems: In cells with enzymes like myeloperoxidase, phenol red can undergo chemical changes that alter assay outcomes
By understanding the role and impact of phenol red in cell culture media, you can make informed decisions that best suit your research needs. The Gibco Media Selection Tool can assist with selection of catalog formulations across a wide variety of formats and configurations with options for custom formulations.
Related Content:
- Find cell culture media with Phenol Red within the Gibco Media Selection Tool
- Search your cell line of interest to find cell culture resources and recommendations with the Cell Culture Select Tool
- Read more about how our media, FBS, and plastics are validated for exceptional product performance so that you can culture with confidence.
References:
- Berthois, Y., Katzenellenbogen, J. A., & Katzenellenbogen, B. S. (1986). Phenol red in tissue culture media is a weak estrogen: Implications concerning the study of estrogen-responsive cells in culture. Proceedings of the National Academy of Sciences, 83(8), 2496–2500.
- Morgan, A., Babu, D., Reiz, B., Whittal, R., Suh, L. Y. K., & Siraki, A. G. (2019). Caution for the routine use of phenol red – It is more than just a pH indicator. Chemico-Biological Interactions, 310, 108739.
- Węsierska-Gądek, J., Schreiner, T., Maurer, M., Waringer, A., & Ranftler, C. (2007). Phenol red in the culture medium strongly affects the susceptibility of human MCF-7 cells to roscovitine. Cellular & Molecular Biology Letters, 12(2), 280–293.
- Weiskirchen, S., Schröder, S. K., Buhl, E. M., & Weiskirchen, R. (2023). A beginner’s guide to cell culture: Practical advice for preventing needless problems. Cells, 12(5), 682.
© 2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.