Genetic research and human health industries largely rely on valuable information and new obtained from recombinant DNA (rDNA) technologies. Restriction enzymes and DNA ligase are used to cut and re-combine genetic materials which can then be inserted into small DNA molecules and left to replicate as a DNA vector. This article explores the concept of recombinant DNA, how new technologies are enabling this level of genetic manipulation, and its modern applications in gene cloning, gene expression studies, and genetic engineering.
In this article we will touch on:
- Recombinant DNA applications
- Gene cloning
- Gene expression
- Genetic engineering
Modern examples of recombinant DNA applications
- With rDNA technologies, scientists have successfully inserted foreign, manipulated DNA into bacterial cells, producing human insulin in pharmaceutical quality at industrial scales.
- . In October 1982, the U.S. Food and Drug Administration (FDA) approved Humulin, the first human insulin created with recombinant DNA technology and used for diabetes treatment. (gov)
- Plasma-derived vaccines were discovered in 1982 and by 1986 the Recombinant HBV DNA vaccine was expressed in yeast and cultivated (nlm.nih.gov)
- Enzymes both aid in the production of recombinant DNA and can be use this technology to create novel enzymes that can be valuable for industries like biopharmaceutical and food. Scientists can manipulate, optimize, and recombine enzymes from different sources in test tubes at the genetic level, enhancing their activities, selectivity or substrate scope (nlm.nih.gov).
Molecular cloning workflow in 5 steps
- Step 1: Fragment generation
- Step 2: Cloning
- Step 3: Transformation
- Step 4: Plasmid preparation
- Step 5: Construct analysis
Learn more about molecular cloning essentials.
Gene cloning
Gene cloning as a technique is fundamental to recombinant DNA technologies. Techniques involved in gene cloning include: isolation of plasmid DNA carrying the desired gene, digestion of the DNA with restriction enzymes, ligation of the DNA to reform with DNA ligase, and introduction of the plasmid into bacteria with transformation.
Gene expression studies
Understanding gene expression is crucial to helping unraveling the mysteries of cellular processes. Gene expression studies may include understanding transcriptional regulation and promoters, reporter genes, and testing of inducible expression systems. By investigating gene expression, scientists gain insights into regulatory networks and disease mechanisms which can help drive development of therapeutics and vaccines.
Gene expression analysis methods include:
- Microarray Analysis
- RNA sequencing (RNA-Seq)
- Serial analysis of gene expression (SAGE)
- PCR
Genetic engineering
Redefining the boundaries of genetic engineering empowers scientists to create novel biological functions and pathways with DNA engineering and manipulation. This section focuses on the tools and techniques employed in genetic engineering, such as site-directed mutagenesis and DNA assembly methods. We explore the applications of genetic engineering, including creating knockout or knockdown models, designing synthetic DNA constructs, and engineering novel biological functions.
There are several tools and techniques employed to help advance the field of genetic engineering including:
- Site-directed mutagenesis: This technique allows researchers to introduce specific changes in DNA sequences at precise locations. Site-directed mutagenesis is used to create mutations in genes, study gene function, and engineer novel biological functions.
- DNA assembly methods: Combine multiple DNA fragments into a larger DNA molecule. This technique can be used to create synthetic genes, gene circuits, and whole genomes.
- CRISPR-Cas9: Genome editing tool that allows researchers to precisely modify DNA sequences. This technique has revolutionized genetic engineering by enabling the creation of genetically modified organisms with exceptional precision.
- Synthetic biology: Combines engineering principles with biology to design and construct new biological systems. This interdisciplinary field involves the creation of novel genetic circuits, metabolic pathways, and organisms with new biological functions.
- Gene synthesis: Provides the process of creating an artificial DNA sequence from scratch allowing researchers to design and create custom DNA sequences, including genes that do not exist in nature.
- RNA interference: Mechanism which allows researchers to selectively silence genes by introducing small RNA molecules that target specific mRNA molecules. Applications in gene therapy, drug discovery, and functional genomics are supported with RNA interference techniques.
GeneArt Gene and Protein Synthesis Services
From sequence to DNA or protein without lifting a pipette. Order plasmids.
Recent advancements and emerging technologies
Here, we discuss the launch of automated plasmid purification technology, and the precision gene editing capabilities of CRISPR-Cas9 and its applications in biomedical research and therapeutics development. Additionally, we explore the potential of synthetic biology and DNA synthesis technologies in designing and building custom DNA constructs for diverse applications.
Automated plasmid DNA isolation has been made possible recently with new instruments including the KingFisher PlasmidPro Maxi Processor. Manual plasmid DNA isolation can be a time-consuming and repetitive process with risk of human error. By automating plasmid DNA isolation, it can help researchers can save time and improve the overall efficiency of their experiments.
CRISPR/Cas9 gene editing technology utilizes a small guide RNA molecule to direct the Cas9 enzyme to a specific target DNA sequence, where it introduces precise changes, such as insertions, deletions, or replacements. This precision gene editing capability of CRISPR-Cas9 has opened new avenues in biomedical research and biomedical research that has turned into therapeutics. By selectively editing genes in cellular and animal models, researchers can investigate the role of specific genes in disease development, evaluate potential drug targets, and unravel complex biological processes.
Challenges working with rDNA and future perspectives
With immense potential in pursuing evolution in the fields of human health, biopharmaceuticals, food safety and beyond, recombinant DNA technologies still face challenges in ethical and regulatory frameworks at macro and molecular levels. Ethical considerations can arise in the public over genetic modification requiring care and complexity.
The future holds exciting prospects as we enhance efficiency and explore new avenues in these dynamic fields. Through the power of recombinant DNA technologies, scientists continue to and unlock new possibilities for biomedical research, therapeutics development and beyond.
© 2024 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.