Detecting Defects in Electrode Coatings to Reduce the Risk of Downstream Failures
Overview
Electrode coatings play a crucial role in the performance and safety of batteries. In manufacturing, it is essential to detect defects in these coatings early on to reduce the risk of downstream failures.
One of the battery industry’s key challenges is dealing with catastrophic and performance failures. Catastrophic failures, such as fires or thermal runaways, can be life-threatening and make headlines. On the other hand, performance failures, such as reduced capacity or slow charge rates, can also significantly impact the end-user’s experience.
The battery manufacturing process begins with mixing the electrode materials and depositing the active material onto the substrate. An oven process and the addition of the separator film follow this. The battery is then slit and put together in a corkscrew fashion.
During the coating process, defects in the electrode coating can occur, which can lead to both catastrophic and performance failures. In-line metrology, which is the process of measuring and analyzing materials during the manufacturing process, can identify these defects early on and segregate the material before it is shipped or made into a final product.
Advanced battery technology and battery material analysis services can help reduce the risk of downstream failures and optimize production processes.
Detecting defects in electrode coatings is a crucial step in the battery manufacturing process to ensure product quality and reduce the risk of downstream failures. By using measurement and control systems and working with experts in advanced battery technology, manufacturers can improve their processes and ensure a reliable and high-performing product.
Battery Manufacturing Stages
Battery manufacturing is a complex process that involves several stages, including electrode manufacturing, packaging, and testing. One of the most critical aspects of this process is the use of metrology, which measures physical properties and materials used in battery production. Metrology plays a crucial role in ensuring the uniform construction of batteries, and it is essential for catching defects early in the process.
The electrode manufacturing process, for example, involves three layers of material: the anode material, which is made of copper and coated with graphite, the separator film, and the aluminum foil coated with active lithium compounds. These three layers all require the use of metrology to ensure that they are well-constructed and of the right thickness and density. The application of the battery also plays a role in the selection of materials, as different applications may require different charge densities or charge rates. 
In-line Metrology
In-line metrology is used during the manufacturing process to ensure that the electrode is homogenous and free of defects. This includes monitoring the coating material to ensure that it is not too thick or too thin and that the anode and cathode are balanced. Additionally, in-line metrology checks for defects such as scratches, voids, and delamination in the coating.
In the anode and cathode production process, for example, metrology is used at several points, including the top and bottom surfaces, to check for uniform distribution of the electro-material. This is important to ensure that the final battery shape is consistent and that it will fit properly in the casing.
Metrology plays a vital role in electrode manufacturing, as it helps to ensure that the batteries are well-constructed, uniform, and free of defects. In-line metrology is particularly useful, as it enables operators to make corrections in real-time, saving time and energy and reducing manufacturing costs.
Electrodes
In the production of lithium-ion batteries, the width of the strip used in the battery electrode is critical to the overall performance and safety of the battery. To ensure that the strip is of uniform width, two approaches can be used. The first is to use an in-line measurement system with slot beam geometry, which allows for higher resolution and better signal to noise ratio when looking for defects. The second is a measurement system with round beam geometry, which is more common but can miss smaller defects.
One example of this is in the detection of scratches on the strip. With a round beam, the change in the signal is small and the scratch may not be detected. However, with a slot beam, the scratch can be clearly identified, and the material can be quickly segregated.
Separator Film
The same technology can also be used in the production of separator film, which is a critical component in the safety of the battery. Infrared-based sensor technologies can be used to measure the thickness of the separator film and ensure it is homogenous from one edge to the other. This is particularly important for wide separator film, which can be up to eight or nine meters wide.
Overall, the use of in-line metrology with slot beam geometry is essential for ensuring a homogenous thickness and an optimum quality of the materials used in lithium-ion batteries, and ultimately, for the safety and performance of the battery itself.
Calendering
One of the key steps in the battery manufacturing process is calendering, which involves compressing the coated electrode to ensure it has the desired dimensions for use in the battery. Defects or inconsistencies in the electrode can negatively impact the performance and reputation of the final product.
One way to identify these defects early on is through laser technology, specifically confocal laser technology. This technology allows for real-time, precise thickness measurement and identification of defects on the electrode sheet.
Early Defect Identification
Identifying defects early in the manufacturing process is important, as later steps such as slitting, cell construction, winding, and testing require additional energy and time. In-line metrology, such as x-ray, beta and laser technology, can aid in identifying defects and allow operators to segregate problematic materials before they become an issue.
This technology can help improve the final product quality and performance, which ultimately benefits both the manufacturer and the customer.