EDXRF Vs WDXRF: Choosing the Right Chemical Analysis Method
In the realm of lab-based X-ray Fluorescence (XRF) analysis, there are two primary systems: energy dispersive X-ray fluorescence (EDXRF) and wavelength dispersive X-ray fluorescence (WDXRF). Each has unique capabilities and applications, making the choice between them essential for accurate chemical analysis.
Understanding EDXRF
EDXRF is renowned for its accessibility, rapid analysis, and minimal sample preparation requirements. It is adept at simultaneously processing groups of elements for both qualitative and quantitative analysis, making it a versatile front-end tool.
An energy-dispersive detector in EDXRF instruments separates the characteristic X-rays of different elements into a complete fluorescence energy spectrum. Filters positioned between the sample and detector are used to improve signal, background reduction, and focus on certain regions of the spectra.
Delving into WDXRF
WDXRF distinguishes itself through high-resolution analysis. This is particularly effective for lower atomic mass elements and rare materials. It employs crystals, such as synthetic thin-film multi-layers, to disperse the fluorescence spectrum into individual wavelengths, providing accurate elemental concentration determinations.
WDXRF systems operate based on Bragg’s Law, reflecting X-rays at specific wavelengths and angles. Two types of detectors, sealed or flow gas for lower energies and scintillation for higher energies, are used and are complemented by goniometers for sequential analysis and monochromators for the simultaneous analysis of fixed elements.
The Use of EDXRF and WDXRF in Different Industries
Applying EDXRF
EDXRF is preferred for its front-end analysis and its versatility when handling various sample types. Its applications include:
  • Cement and Building Materials: Analyzing the composition of materials
  • Mining and Exploration: Including nickel ore analysis
  • Environmental Monitoring: Assessing pollution and contamination levels
  • Pharmaceuticals: Verifying ingredients and quality control
  • Petrochemicals: Monitoring chemical processes and products
  • Forensics: Analyzing materials for crime scene investigations
  • Art and Archaeology: Uncovering historical compositions without damaging the artifacts.
Applying WDXRF
WDXRF is valued for its high-resolution analysis, particularly in:
  • Geological sample analysis: Studying earth materials
  • Cement and building materials: Assessing material quality and consistency
  • Metals and mining: Analyzing the elements in ores and alloys
  • Environmental analysis: Examining environmental samples
  • Research and academia: Researching material sciences.
Both EDXRF and WDXRF are integral to various industries. EDXRF is best for quick and versatile analysis across diverse samples, while WDXRF provides a detailed and high-resolution analysis of specific elements, especially about more complex matrices.
The choice between these techniques hinge on the analysis requirements, such as elemental interest, sample matrix, and detection limits.
Choosing the Right Method: EDXRF Vs WDXRF
When deciding between EDXRF and WDXRF, you must consider the following:
  • EDXRF for quick analysis: Ideal for scenarios requiring a fast and straightforward analysis. It can process groups of elements simultaneously and requires minimal sample preparation. EDXRF’s versatility extends to both portable and lab-based formats. These are suitable for a broad range of applications.
  • WDXRF for detailed resolution: Best suited for high-solution applications, especially for analyzing lower atomic mass elements and rare earth materials, its ability to separate signals into individual wavelengths result in highly accurate elemental concentration determinations.
  • Application-specific considerations: Your choice depends on the specific needs of the analysis, including the elements of interest, the desired resolution level and time-to-result.
  • Mining applications: Both EDXRF and WDXRF are effective for analyzing soil and sediment samples in mining and highlighting their adaptability in different industrial contexts.
With this in mind, the decision between EDXRF and WDXRF hinges on the specific demands of the analysis. The factors of your analysis must be balanced, such as speed, resolution, and the nature of the sample.
Both EDXRF and WDXRF play crucial roles in modern chemical analysis. Understanding their distinct functionalities and applications is critical to selecting the right method for your specific analytical goals and ensuring accurate and reliable results.
Additional Resources
If you are interested in learning more about these innovative methodologies, refer to our XRF Academy for more on the fundamentals and applications.
Written by:
Christina Drathen
Product Manager Leader - Metals & Minerals, Thermo Fisher Scientific
Christina Drathen leads the Product Management team for Metals and Minerals at Thermo Fisher Scientific, bringing over 10 years of experience in X-ray analytical tools and a PhD in Chemistry from the University of Edinburgh, UK.
Read more Drathen, Christina
Comments
  1. Lotfi Monser says
    Dear sir
    I want to know which one is better for the following sample matrix:
    Cathodic copper
    Copper concentrates (copper & other trace metals)
    Cobalt concentrates (cobalt & other trace metals )
    Other mining rocks or ores
  2. Md. Nure Alam Siddik says
    Dear sir,
    Have a good day.
    I want to know which one is better type (EDXRF) vs (WDXRF) for the following sample matrix:
    Glassware and tableware raw materials.
    Other mining rocks or ores test purpose.