How to Determine the Degree of Graphitization in Battery Anodes
Graphitization in battery manufacturing refers to the process of transforming amorphous carbon materials into crystalline graphite structures. This process involves subjecting the carbon-based materials, typically in the form of anodes, to high temperatures in an inert atmosphere.
The purpose of graphitization is to enhance the structural stability and electrochemical performance of the battery by increasing the graphite content in the anode. This allows for better lithium-ion intercalation and de-intercalation during the charging and discharging processes, resulting in improved battery capacity, efficiency, and cycle life.
The Making of a Battery Anode
A battery anode is one of the two electrodes in a battery, along with the cathode. It is the electrode where oxidation reactions occur during the battery’s discharge or usage phase.
In a typical rechargeable battery, such as a lithium-ion battery, the anode is typically made of a material that can intercalate or store ions, such as lithium ions. During the discharge process, lithium ions migrate from the anode to the cathode through an electrolyte, creating a flow of electrons that can be harnessed as electrical energy.
Common materials used for battery anodes include graphite in lithium-ion batteries, zinc in zinc-carbon batteries, and lithium metal in lithium-metal batteries. While natural graphite is directly obtained from geological sources via mining and has variable properties, processed graphite undergoes various treatments to enhance its characteristics, making it suitable for more specialized and high-performance applications. Whether the material is natural or processed, it is analyzed for quality. The choice of anode material depends on factors such as quality, energy density, cycling stability, safety, and cost.
The design and optimization of the anode material play a crucial role in determining the battery’s overall performance, including its capacity, efficiency, and cycle life. Researchers continue to explore new materials and technologies to improve anode performance and address challenges such as capacity degradation and safety concerns.
Assessing the Degree of Graphitization with XRD Analysis
To optimize the electrochemical performance and energy storage capabilities of carbon-based battery anodes, it is important to accurately characterize the degree of graphitization. One effective method for precisely assessing the degree of graphitization is X-ray diffraction (XRD) analysis.
X-ray diffraction (XRD) analysis is a technique used to study the atomic and molecular structure of materials. It involves directing a beam of X-rays onto a sample, which causes the X-rays to interact with the crystal lattice of the material. The interaction results in the scattering of the X-rays at different angles, producing a diffraction pattern. The diffraction pattern contains information about the arrangement of atoms within the sample.
By analyzing the diffraction pattern, XRD analysis can determine various properties of the material, such as crystal structure, lattice parameters, unit cell dimensions, and the presence of impurities or defects. It is widely used in a range of fields, including materials science, chemistry, geology, pharmaceuticals, and metallurgy.
An Example of XRD Analysis of Graphite Anode Material
We conducted an experiment to see if it was possible to determine the degree of graphitization for battery anodes using an advanced benchtop X-ray diffractometer.  By determining the location of the (002) reflection, the graphitization degree can be quantitatively evaluated. This quantity must be >90%, but the optimal value depends on the cathode as well as the anode, and therefore the value of graphitization needs to be monitored for each specific type of battery as part of regular quality control by operators. Such analysis is ideally automated using software which streamlines the analysis process and provides rapid and reliable results. Additionally, adherence to the Chinese norm GB/T 24522-2019 ensures compliance with industry standards.
The insights gained from XRD analysis, combined with automated analysis capabilities, enable researchers and manufacturers to optimize the analytical workflow and ensure expected battery anode performance.
You can get more details about the study in an application note, including material type and preparation, instruments and technologies used, measurement spectra, and results.
Summary
It was found that the XRD instrument and its associated software were well-suited to determine the degree of graphitization for battery anode material. The results could be obtained in a one-click analysis in accordance with the GB/T 24522-2019 norm, with the goal to optimize cathode/anode interaction by finding the most suitable g value.
Using XRD analysis in battery anodes manufacturing can provide valuable insights into the crystallographic properties of materials, helping researchers understand their composition, phase identification, and structural characteristics – possibly leading to better battery life and performance.
Additional Resources
Written by:
Simon Welzmiller
Global Application Specialist XRD, Thermo Fisher Scientific
Simon Welzmiller has been working as the Global Application Specialist for XRD at Thermo Fisher Scientific since 2017. He has more than 15 years of experience in various XRD applications and techniques. Simon holds a Ph.D. in solid-state chemistry from Leipzig University and a Master’s degree in Chemistry from LMU Munich.
Read more Welzmiller, Simon