Thin Film Applications in Energy
This is part of our New Materials are Getting Thinner and Thinner series. In case you missed it, start by reading part 1.
Thin film technology
As the photovoltaics industry continues to innovate, considerable progress has been made in eliminating solar panels that use heavy and inflexible silicon substrates. Using thin film technologies, these advances enable easier installation and contoured form factors. An important challenge to alternative energy sources is how energy can be stored once it is collected. And here too thin film technologies play an important role.
GIXRD
GIXRD
Thin film solar panels are typically made with one of the following four material types:
Cadmium Telluride
Cadmium Telluride (CdTe) is the most widely used thin film technology. CdTe contains significant amounts of cadmium, which is relatively toxic.
Amorphous Silicon
Amorphous Silicon (a-Si), the non-crystalline form of silicon, is a second popular thin film option. With similar technology to that of a standard silicon wafer panel, a-Si is a better option in terms of toxicity and durability, but it is less efficient and is typically used for small load requirements like consumer electronics.
Copper Indium Gallium Selenide CIGS
Copper Indium Gallium Selenide (CIGS) cells have reached efficiency highs of 22.4%. Despite considerable interest and research, these performance metrics are not yet possible at scale. In fact, two of the major CIGS manufacturing companies are now bankrupt, including the infamous Solyndra.
Gallium Arsenide GaAs
Gallium Arsenide (GaAs) is a very expensive technology but holds a world record 28.9% efficiency for all single-junction solar cells. GaAs is primarily used on spacecrafts and is meant for versatile, mass-scale installments for energy collection in unusual environments.
Thin film lithium battery research
Thin film lithium batteries are an increasingly important field of energy storage, solving the problem of what to do when the sun goes down or the wind stops. Instead of liquid or polymer gel materials, solid-state battery technology uses solid electrodes and a solid electrolyte. Safer and with higher-energy densities, solid-state batteries show promise for pacemakers, wearable batteries, RFID devices and other applications.
In a thin film lithium battery, the electrolyte is solid, and the other components are deposited in layers on a substrate. The solid electrolyte may also serve as a separator material. These materials create flexible batteries cells that are only a few microns thick.
As the demands for safety, higher energy density, and other performance metrics increase, research into anode, cathode, and electrolyte materials has been rapidly progressing. Cathode materials are often complex lithium-oxides such as LiCoO2, LiMn2O4, and LiFePO4, and anode materials are typically comprised of carbon-based materials such as graphite, lithium metal or other metallic materials.
In Part 3, we’ll look at how thin film materials are made.
To learn how thin films can be measured using X-ray diffraction, read our published application note, Investigation of Ni on Si thin film with ARL EQUINOX 100 X-ray Diffractometer.
Want to speak to someone? Need a quote? Want a demo?
Contact us
Opt in for email communications on this and other topics.
Sign up for email
Nathalie Martin, Thermo Fisher Scientific
Written by:
Nathalie Martin
Sr Market Development Manager, Thermo Fisher Scientific
Nathalie Martin supports scientific audiences through clear, data-driven communication. Her work focuses on translating complex analytical technologies into practical insights that enable informed decision-making across global markets.
Read more Martin, Nathalie
Comments
  1. Richard Green says
    Dear Sir/Madam,
    Greetings!
    We take this opportunity to express our gratitude for Companies like you to collaborate and participate in the event
    Join us at Materials Congress 2020 webinar and gain critical insights in the field of Materials Science & Engineering.
    This webinar offers a wide range of critically important scientific sessions from basic research to latest advances in the field.
    Sessions:
    Materials Science and Engineering Electronic, Optical and Magnetic Materials Materials Science and Materials Chemistry
    Biomaterials and Medical Devices Emerging Smart Materials Applied Materials Chemistry
    Nanomaterials and Nanotechnology Physics and Chemistry of Materials Green Polymers & Green Materials
    Polymer Science and Technology Graphene and 2D Materials Advanced Energy Materials
    For insights: https://materialsscience.insightconferences.com/webinar
    It gives us great pleasure to invite your team to take part in the event.
    Eagerly looking forward to your participation
    Thanks & Regards
    Richard Green
    Conference Director
    Materials Congress 2020
    materialscongress@brainstormingmeetings.com