Using XRD to Determine Crystallite Sizes and Phase Composition of Molybdate-Type Catalysts
Molybdenum catalysts
Similar to molybdenum-bearing enzymes which are common bacterial catalysts that break down the chemical bond of molecular nitrogen in the atmosphere in the process of biological nitrogen fixation, which is critical to plant life and agriculture, molybdates are molybdenum compounds in high oxidation states. Used as catalysts in selective oxidation of hydrocarbons, molybdates are precursor compounds for catalysts used in the hydrodesulfurization of refined petroleum products and natural gas. Hydrodesulfurization is a critical step in refining fossil and biofuels to remove sulfur, a known pollutant responsible for smog. The process is also used to create ultra-low sulfur diesel fuel and to upgrade the octane rating of intermediate hydrocarbons in the production of petroleum.
Molybdate catalyst
Molybdate catalyst
Types of molybdate catalysts
Nickel molybdate (NiMoO4) has attractive structures, higher specific capacitance, higher electrochemical activity and magnetic properties compared to other molybdates (Zn, Co, Mg, etc.) because it displays high density of states near the top of the valence band. It is used in the oxidative dehydrogenation of light alkanes such as propane and has been studied for use as a high-performance NiMoO4xH2O nanorod electrodes for supercapacitors. It can be found in three crystalline forms under atmospheric pressure, α-NiMoO4 at low temperature, β-NiMoO4 at high temperature, NiMoO4nH2O hydrate, and another allotrope (NiMoO4-II) at high pressure. (ref. https://www.ncbi.nlm.nih.gov/pmc/articles/PMC6017352/) X-ray diffraction (XRD) is a method that is very sensitive to changes in the crystallographic structure of solid materials and commonly used to distinguish different crystalline forms.
Cobalt molybdate heterostructures (CoMoO4 and CoO2MoO3O8) create a high-performance hydrogen evolution reaction, which has been suggested as a promising clean and renewable energy source. These molybdate heterostructures were found to create efficient and inexpensive electrocatalysts for catalytic reactions that produced energy with high voltage efficiency, excellent long-term stability, and superior conductivity. The materials were shown to reduce hydrogen adsorption energy and significantly enhance electrocatalytic activity (ref. https://pubag.nal.usda.gov/catalog/6539770).
XRD analysis of catalyst particles
The catalytic activity of such molybdenum materials strongly depends on the particle size and the ratio to the support material. For these types of materials, Al2O3 is often used as a support material and appears as a mixture of amorphous and crystalline fractions. The most commonly used method to determine both the crystallite size of nanoparticles and the quantitative composition of solid mixtures is XRD.
One of our XRD application specialists, Dr. Simon Welzmiller, conducted XRD measurements of NiMoO4 and CoMoO4 powders to determine quantitative phase analysis and crystallite size of these candidate molybdate catalysts. Using our ARL EQUINOX 100 benchtop X-ray diffractor, the qualitative phase analysis revealed that both samples contain a significant amount of Al2O3, which acts as a support for the actual β-CoMoO4 and β-NiMoO4 phases. The analysis was performed in 45 minutes. This analysis revealed significant amounts of active molybdate catalysts with crystallite size of 10 nm for both molybdates.
For more information on this, you can download our whitepaper. Download “Characterization of molybdate type catalysts using benchtop ARL EQUINOX 100 XRD”.
Visit XRD Academy to learn about XRD basics and fundamentals, as well as its main applications, and watch XRD how-to videos.
Want to speak to someone? Need a quote? Want a demo?
Contact us
Opt in for email communications on this and other topics.
Sign up for email
Round gray circle with a human silhouette. Dark gray shadow on the bottom. Isolated on a light background. EPS 10.
Written by:
Carl D. Millholland
Read more Millholland, Carl