  • Contact
Accelerating ScienceAnalyzing Metals / Positive Material Identification / What is LIBS and What Does It Have to Do With PMI?
What is LIBS and What Does It Have to Do With PMI?
Written by Brian Dalton | Published: 04.07.2020
What is LIBS and What Does it Have to Do With PMI?Laser Induced Breakdown Spectroscopy (LIBS) is the analytical technique using a high-focused laser to determine the chemical composition of materials. LIBS has been around for many years and is a technique used primarily in laboratory equipment. With recent advances in technology, the technique has now been developed into a portable handheld analyzer capable of measuring elements, including carbon, in the field for material identification.
LIBS is an important technology used in the oil and gas industry for positive material identification (PMI) of piping, pressure vessels, valves, pumps, and finished welds, or to grade unknown materials to regain traceability. Changing the amount of carbon in these products can change the properties of the steel including tensile strength, hardness, weldability, ductility, and corrosion resistance.
Fatal accidents and injuries, as well as leaks, premature pipe replacements, loss of property, and unplanned outages at refineries, chemical plants and gas processing facilities often can be traced back to equipment failures due to faulty or counterfeit metal building components or because piping is made from material that does not meet specifications.
LIBS analysis is vital to calculating carbon equivalency prior to welding to determine heat affected zone hardenability (HAZ).
The LIBS analysis process
The LIBS technique utilizes a high-focused laser that interacts with the surface of a material and forms a plasma in which the material is broken down into single elements. A laser pulse is produced by the analyzer and pointed at the sample surface. The surface is ablated and enters the plasma.
The plasma atomizes the samples, and the excited atoms emit light as they return to ground state. The emitted light is transferred through fiber optics and enters the spectrometer through a slit. The light interacts with a diffraction grating where it is split into single wavelengths/ colors. The single wavelengths/ colors hit the detector and produce spectral data.
The central processing unit (CPU) analyzes the spectral data and determines the concentration of each element present in the sample. Composition data and identified alloy grade are displayed and stored via memory for later recall or download to an external PC.
If you would like to know more about LIBS, we have outlined the process and explained the science behind this technology in our easy-to-understand eBook: LIBS technology for non-scientists.  This free eBook also explains the components that make up a handheld LIBS analyzer and offers 10 features to look for when purchasing one.
  • Download the ebook: LIBS technology for non-scientists
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Share this article
FacebookLinkedinTwitterMail
radiation symbol for xrf safety
I Use XRF Analyzers on Metals… Is it Safe?
Do you use portable x-ray fluorescence (XRF) analyzers for... by Esa Nummi / 06.17.2025
Read More
gold metallic number 5
5 Precious Metals Analysis Methods
We’ve previously written about the four primary precious m... by Juan Pablo Zuluaga / 05.15.2025
Read More
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
The Connection Between Scrap Metal Recycling and Drug Abuse
Steel Production Content at Your Fingertips
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook