Best Practices for Accurate In Situ Geochemical Analysis
In situ geochemical analysis refers to elemental testing performed directly on geological materials in their natural location, avoiding the delays and costs associated with laboratory testing.  While in situ geochemical analysis delivers immediate elemental data, the reliability of those results depends heavily on proper field methodology. Portable analytical technologies used in mining and mineral exploration are powerful, but like any scientific tool, they require disciplined sampling and measurement techniques to produce defensible data.
Understanding why best practices matter begins with recognizing one key principle: the instrument can only analyze what is directly in front of it. If the sample is inconsistent, contaminated, or poorly positioned, the results will reflect those issues.
Below is a deeper look at the technology used for analysis, and the four core best practices that help significantly improve data quality.
Portable XRF Technologies
Handheld X-ray fluorescence (XRF) analyzers facilitate and provide rapid, non-destructive elemental analysis of geological samples.
XRF is an acronym for X-ray fluorescence spectroscopy. XRF is a non-destructive analytical technique used to determine the elemental composition of materials. Handheld XRF analyzers work by measuring the fluorescent (or secondary) X-rays emitted from a sample when excited by a primary X-ray source.
Each of the elements present in a sample produces a set of characteristic fluorescent X-rays, or “unique fingerprints”. These “fingerprints” are distinct for each element, making handheld XRF analysis an excellent tool for quantitative and qualitative measurements.
These portable devices interpret these ‘fingerprints’ and enable immediate, on-site analysis, supporting quick decision-making and more efficient exploration activities. By delivering accurate elemental composition data, portable XRF technology enhances the speed and precision of geological surveys, making it an indispensable tool for prospecting, exploration, and optimizing mining operations.
1. Ensure Samples Are Representative
A representative sample accurately reflects the overall material being evaluated. In geological and environmental contexts, materials are rarely uniform. Elemental concentrations can vary within centimeters due to mineralization patterns, grain size differences, or contamination hotspots.
If a measurement is taken on an unrepresentative surface:
  • Results may overestimate or underestimate true concentrations
  • Ore boundaries may be misidentified
  • Contamination mapping may become inaccurate
  • Follow-up laboratory assays may not correlate
To improve representativeness:
  • Remove weathered or altered surface layers when possible
  • Avoid measuring visibly heterogeneous areas
  • Take multiple readings across a defined area
  • Average results when appropriate
Representative sampling reduces variability and increases confidence in field-based elemental analysis. Practically, handheld XRF measurements performed directly on rocks or soils are at best semi-quantitative and provide only ballpark figures in terms concentrations. Those results remain useful to locate pollution hotspots or ore-waste boundaries.
2. Prepare Powdered or Homogenized Materials for Quantitative Analysis
In situ testing can be performed directly on rock faces, soil surfaces, or drill core. However, for quantitative geochemical analysis, sample preparation significantly enhances accuracy.
Why? Because surface texture, particle size, localized heterogeneities and mineralogy can influence X-ray interaction and signal detection.
Grinding and homogenizing a sample:
  • Reduces particle size effects
  • Minimizes localized mineral concentration bias
  • Improves repeatability
  • Produces more consistent matrix density
Powdered samples provide a more uniform analytical surface, which improves measurement precision and considerably reduces statistical noise. For screening purposes, minimal preparation may suffice. For reporting quantitative concentrations, homogenization is strongly recommended.
3. Use Stable Measurement Setups
Measurement geometry plays a critical role in portable elemental analysis. Inconsistent positioning, movement during analysis, or uneven contact between the analyzer and sample can introduce variability.
Common stability issues include:
  • Gaps between instrument window and sample
  • Shifting during measurement
  • Uneven sample surfaces
  • Variable angle positioning
Using stable setups — such as flat sample cups, test stands, or firm surface placement — helps ensure consistent X-ray path length and detector exposure.
Stability improves:
  • Signal consistency
  • Reproducibility
  • Detection limits
  • Overall data reliability
Even small inconsistencies in positioning can affect results, especially when analyzing trace-level elements.
4. Apply Appropriate Calibration Adjustments for Specific Matrices
A “matrix” refers to the overall composition and physical characteristics of a sample.  Geochemical samples all have different matrix properties that influence X-ray absorption and enhancement effects.
Without proper calibration:
  • Elemental concentrations may be biased
  • Light elements may be under- or over-reported
  • Results may not correlate with laboratory assays
Modern portable analyzers use often fundamental parameter (FP) models combined with empirical adjustments to compensate for matrix effects. Selecting the correct analytical mode or calibration profile for the sample type is essential.
Moreover, applying type standardization (matrix-specific calibration adjustments) improves:
  • Accuracy of quantification
  • Correlation with lab data
  • Traceability and data defensibility in regulatory or reporting contexts
Calibration is not just a software setting — it is a process that helps ensure the instrument interprets fluorescence signals correctly. We recommend that you monitor drifts and verify accuracy of calibration by running relevant certified reference materials.
Integrating Field Screening and Laboratory Confirmation
In situ geochemical analysis is most powerful when integrated into a broader analytical workflow.
Field screening provides high density of measurement for:
  • Immediate decision support
  • Rapid anomaly detection
  • Real-time drill targeting
  • Efficient contamination mapping
Laboratory confirmation provides:
  • High-precision validation
  • Regulatory-grade reporting
  • Advanced multi-method analysis
When combined strategically:
  • Field data reduces unnecessary laboratory submissions
  • Laboratory data validates and refines field calibration
  • Overall project timelines shrink
  • Analytical budgets are optimized
Rather than replacing laboratory analysis, in situ testing enhances it. The two approaches work best as complementary tools within a data-driven field strategy.
Summary
In situ geochemical analysis delivers speed and flexibility. However, accuracy depends on disciplined sampling, preparation, measurement stability, and proper calibration.
Organizations that apply these best practices achieve:
  • Stronger correlation with laboratory assays
  • Lower analytical uncertainty
  • More defensible reporting
  • Greater operational confidence
In modern field operations, data quality is not determined by the technology alone — it is determined by the accuracy of the analyzer and how the technology is used.
Read more about in situ analysis for mining and minerals in our Application Note: Exploring New Frontiers of In Situ Geochemical Analysis
Resources
Written by:
Mathieu Bauer
Senior Application Scientist, Thermo Fisher Scientific
Mathieu has almost 25 years of experience with various analytical technologies including NIR, FT-IR, ICP-OES, MS, XRF and LIBS. Mathieu is the Senior Application Scientist for the Thermo Scientific Niton Analyzers product line and provides his expertise in handheld XRF and handheld LIBS to our commercial team and end-users. He also supports the development of new markets and applications for both technologies. Mathieu is fluent in French, English and German and is based out of Europe.
Read more Bauer, Mathieu