How Do I Use Inline Raman Spectroscopy in Downstream Oil & Gas Applications?
Inline Raman spectroscopy is a powerful analytical technique with numerous applications in the downstream oil and gas industry, including quality assurance. One of the key advantages of Raman spectroscopy is its ability to perform non-destructive analysis, making it an ideal technique for real-time monitoring of various processes. Inline Raman also provides valuable information about the chemical composition and properties of samples, making it useful for monitoring processes and identifying contaminants.
Inline Raman spectroscopy is particularly useful in this regard, as it allows continuous monitoring of the process stream without sample preparation. This blog post will discuss using inline Raman spectroscopy in downstream oil and gas and outline the steps required to add this technique to your process.
Step 1: Determine Your Requirements
Determining your requirements is the first step in adding inline Raman to your downstream oil and gas applications. This includes understanding the type of samples you will be analyzing, the desired measurement parameters, and the environmental conditions in which the Raman system will operate.
Some key considerations include:
  • Sample type: Raman spectroscopy is suitable for analyzing liquids and gases. Site integration considerations, such as fluid process conditions and piping modifications, should be defined to allow for the selection of probe interface.
  • Measurement parameters: Consider the specific parameters you need to measure, such as chemical composition, concentration, and physical properties. This will help determine the appropriate Raman system configuration and spectral range.
  • Environmental conditions: Inline Raman systems may be exposed to harsh environmental conditions, such as high temperatures and pressures, corrosive fluids, and hazardous gases. Ensure that the Raman system is designed to withstand these conditions.
Step 2: Select a suitable Raman system
Once you have determined your requirements, the next step is to select a suitable inline Raman system.
When selecting a system, consider the following factors:
  • Spectral range: Ensure the system can measure the spectral range required for your analysis.
  • Sampling interface: Inline Raman systems typically use a fiber-optically connected probe to interface with the process stream. Consider the size and design of the probe to ensure it is compatible with your process conditions.
  • Sensitivity and resolution: These parameters determine the detection limit and the ability to resolve closely spaced spectral features. Consider the expected concentrations and complexity of your samples.
  • Robustness and reliability: Inline Raman systems must be designed to withstand the harsh environmental conditions of oil and gas applications. Consider the construction materials, sealing mechanisms, and protection against shock and vibration. Also, consider whether the Raman analyzer is factory-calibrated, which can save significant time and money in the long run.
Step 3: Integrate the Inline Raman system into your process
The final step is to integrate the Raman system into your process. This involves installing the fiber-optic probe into the process stream and connecting the Raman system to the control system.
Some key considerations include:
  • Installation location: The probe should be installed in a representative location of the process stream and accessible for maintenance.
  • Site integration: Power and applicable site utilities will need to be available to integrate the Raman system. Communications cabling will need to be connected between your DCS/SCADA network and the Raman system.
  • Calibration: Raman systems requiring recalibration cannot continue to take measurements while recalibrating. A factory-calibrated Raman system can help you get started immediately by enabling out-of-the-box installation directly in your process.
  • Data analysis: The Raman system should be configured to provide real-time data analysis and feedback to the control system. This may involve developing customized software or algorithms to interpret the Raman spectra.
Summary
Adding inline Raman to downstream oil and gas applications can provide valuable insights into process monitoring and control. Consider an analyzer that features a robust design, is high sensitivity, provides real-time feedback, and is easy to integrate and customize to ensure it meets your specific requirements.
Additional Resources
Written by:
John Richmond
Sr. Manager, Sales, Thermo Fisher Scientific
Before retiring John Richmond spent several years as a Sales Specialist focusing on analytical and process instruments.
Read more Richmond, John