Is Portable XRF Technology Useful for Mining Applications?
The realm of mineral exploration and mining has significantly evolved over the years, with a keen shift towards innovative technologies that improve efficiency and yield. Among these, portable X-ray fluorescence (XRF) technology has emerged as a vital tool, becoming a mainstay in the search for precious metals, rare earth elements, and in greenfield exploration.
Portable XRF is not just a peripheral gadget in the mining toolkit, but rather a comprehensive solution that amplifies value across the entire mining value chain. Traditional portables were designed to deliver the top-grade performance of a benchtop analyzer yet enable users to move lab-based analysis into the field. Although these on-site mining technologies provided significant time-savings, they lacked a certain level of practicability and user-friendliness. The next evolution of XRF technology for industrial use was the single-unit handheld XRF analyzer – sometimes abbreviated as HHXRF. Handheld systems are ergonomic XRF instruments designed to maximize productivity through ease of use in various industrial locations such as outdoors or in corners and crevices. So, how exactly do handheld XRF analyzers enhance mining operations?
Where Portables Fit into Mining & Exploration
In the broad landscape of mining, portable XRF finds applications across a spectrum of tasks. Its wide use is attributable to its immediate, non-destructive elemental analysis. To get a holistic perspective on the role of portable XRF, let’s explore its role in various mining operations.
  • Exploration: As an initial step in mining, exploration is crucial. Portable XRF supports this process by enabling fast identification of mineral anomalies for further investigation.
  • Geochemical Mapping: Geochemical mapping forms the core of many exploration programs. With portable XRF, vast areas can be mapped effectively to delineate ore deposits.
  • Grade Control: An accurate estimation of ore grade is pivotal to the profitability of mining operations. XRF analyzers contribute by providing rapid feedback on grades, thus assisting in operational decisions.
  • Process Monitoring: XRF can be used to monitor processing activities to ensure efficient recovery of precious metals.
  • Environmental Compliance: Mining activities are subject to stringent environmental regulations. XRF technology helps in ensuring compliance by analyzing soil and water samples for contaminant levels.
Using Portable XRF Analysis in Open Pits and Underground Mines
There’s a plethora of minerals present in the wide expanse of the earth’s crust, each varying in elemental composition. Portable XRF analyzers exhibit an extensive elemental range, typically from magnesium to uranium. This wide range creates opportunities for identifying not just traditional base metals and precious metals, but also rare earth elements that are increasingly sought after for their unique properties. Regardless of the sample type—whether it is drill core, bagged samples, or samples cut directly from the mine face—portable XRF can help decipher its elemental constituents.
The unique challenges of hard rock mining stem from the necessity of extracting tightly bound ores and minerals such as gold, silver, iron, copper, zinc, and precious gemstones. These operations involve working with tough igneous and metamorphic rocks. The complex mineralogical matrix within these rocks complicates the efficient extraction of valuable minerals. Amidst these challenges, portable XRF technology emerges as a standout tool. It enables real-time, in situ elemental analysis of rock samples, providing immediate and accurate data about the elemental composition and mineral concentration variability. This data empowers geologists to make prompt, informed decisions, accelerating the process of locating and assessing the profitability of ore bodies. As a result, the overall efficiency and productivity of hard rock mining operations are bolstered.
By allowing for near-instantaneous, non-destructive analysis, portable XRF enables mining geologists to make immediate, informed decisions without having to send samples to a laboratory and wait for results. This on-site decision-making capability contributes to an increase in productivity and, ultimately, profitability. It can facilitate precise ore deposit modeling and ore grade control, which contributes to a well-optimized mining operation. Handheld XRF analysis can add significant gains beyond the mining industry too.
To summarize: portable XRF technology is changing the landscape of the mining industry, enabling explorers to optimize their efforts and maximize the value of their mining operations.
Additional Resources:
Written by:
Mathieu Bauer, PhD
Senior Application Scientist, Thermo Fisher Scientific
Mathieu has almost 25 years of experience with various analytical technologies including NIR, FT-IR, ICP-OES, MS, XRF and LIBS. Mathieu is the Senior Application Scientist for the Thermo Scientific Niton Analyzers product line and provides his expertise in handheld XRF and handheld LIBS to our commercial team and end-users. He also supports the development of new markets and applications for both technologies. Mathieu is fluent in French, English and German and is based out of Europe.
Read more Bauer, Mathieu